Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15020374-15020881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC105
Entrez Gene ID
126402
Gene Name
coiled-coil domain containing 105
Gene Aliases
-
UniGene
Hs.375985
Gene Chromosome Location
Chr.19: 15010727-15023271 on Build GRCh38
Transcripts
NM_173482.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3932908 c.746A>C p.N249T Chr.19: 15020531-15020531 on Build GRCh38
3282757 c.702G>A p.L234L Chr.19: 15020487-15020487 on Build GRCh38
3849578 c.874G>A p.A292T Chr.19: 15020659-15020659 on Build GRCh38
6414570 c.759C>A p.D253E Chr.19: 15020544-15020544 on Build GRCh38
1129970 c.727C>A p.Q243K Chr.19: 15020512-15020512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781131148 Chr.19: 15020661-15020661 on Build GRCh38
rs35352238 Chr.19: 15020518-15020518 on Build GRCh38
rs369331488 Chr.19: 15020567-15020567 on Build GRCh38
rs944093017 Chr.19: 15020726-15020726 on Build GRCh38
rs187938143 Chr.19: 15020700-15020700 on Build GRCh38
rs745767305 Chr.19: 15020675-15020675 on Build GRCh38
rs956265571 Chr.19: 15020725-15020725 on Build GRCh38
rs540706591 Chr.19: 15020548-15020548 on Build GRCh38
rs34375549 Chr.19: 15020528-15020528 on Build GRCh38
rs201032992 Chr.19: 15020598-15020598 on Build GRCh38
rs202152474 Chr.19: 15020588-15020588 on Build GRCh38
rs747426926 Chr.19: 15020485-15020485 on Build GRCh38
rs951260385 Chr.19: 15020426-15020427 on Build GRCh38
rs752860423 Chr.19: 15020561-15020561 on Build GRCh38
rs182391995 Chr.19: 15020697-15020697 on Build GRCh38
rs879848708 Chr.19: 15020729-15020729 on Build GRCh38
rs575230957 Chr.19: 15020477-15020477 on Build GRCh38
rs920612714 Chr.19: 15020803-15020803 on Build GRCh38
rs76535067 Chr.19: 15020683-15020683 on Build GRCh38
rs1013279928 Chr.19: 15020850-15020850 on Build GRCh38
rs573133985 Chr.19: 15020814-15020814 on Build GRCh38
rs117318288 Chr.19: 15020464-15020464 on Build GRCh38
rs372493151 Chr.19: 15020590-15020590 on Build GRCh38
rs770702173 Chr.19: 15020659-15020659 on Build GRCh38
rs767495731 Chr.19: 15020541-15020541 on Build GRCh38
rs763129327 Chr.19: 15020613-15020613 on Build GRCh38
rs34616225 Chr.19: 15020438-15020439 on Build GRCh38
rs779397868 Chr.19: 15020497-15020497 on Build GRCh38
rs773220791 Chr.19: 15020531-15020531 on Build GRCh38
rs35827059 Chr.19: 15020501-15020502 on Build GRCh38
rs201745447 Chr.19: 15020625-15020625 on Build GRCh38
rs569608268 Chr.19: 15020703-15020703 on Build GRCh38
rs767909552 Chr.19: 15020631-15020631 on Build GRCh38
rs768611128 Chr.19: 15020413-15020413 on Build GRCh38
rs200013323 Chr.19: 15020507-15020507 on Build GRCh38
rs370743090 Chr.19: 15020645-15020645 on Build GRCh38
rs202195475 Chr.19: 15020624-15020624 on Build GRCh38
rs201115065 Chr.19: 15020593-15020593 on Build GRCh38
rs778523539 Chr.19: 15020639-15020639 on Build GRCh38
rs567174748 Chr.19: 15020448-15020448 on Build GRCh38
rs752449861 Chr.19: 15020644-15020644 on Build GRCh38
rs562182355 Chr.19: 15020566-15020566 on Build GRCh38
rs772353273 Chr.19: 15020443-15020443 on Build GRCh38
rs141552724 Chr.19: 15020494-15020494 on Build GRCh38
rs373445003 Chr.19: 15020606-15020606 on Build GRCh38
rs545950246 Chr.19: 15020754-15020754 on Build GRCh38
rs141714050 Chr.19: 15020517-15020517 on Build GRCh38
rs754052984 Chr.19: 15020653-15020653 on Build GRCh38
rs778359354 Chr.19: 15020481-15020481 on Build GRCh38
rs925328953 Chr.19: 15020680-15020680 on Build GRCh38
rs760818941 Chr.19: 15020451-15020451 on Build GRCh38
rs776563275 Chr.19: 15020545-15020545 on Build GRCh38
rs747029802 Chr.19: 15020599-15020599 on Build GRCh38
rs754555236 Chr.19: 15020637-15020637 on Build GRCh38
rs763293071 Chr.19: 15020687-15020687 on Build GRCh38
rs761895947 Chr.19: 15020436-15020436 on Build GRCh38
rs7249373 Chr.19: 15020856-15020856 on Build GRCh38
rs765450342 Chr.19: 15020559-15020559 on Build GRCh38
rs896136220 Chr.19: 15020829-15020829 on Build GRCh38
rs1048333446 Chr.19: 15020826-15020826 on Build GRCh38
rs917380660 Chr.19: 15020733-15020733 on Build GRCh38
rs921328897 Chr.19: 15020450-15020450 on Build GRCh38
rs748997369 Chr.19: 15020506-15020506 on Build GRCh38
rs7248834 Chr.19: 15020401-15020401 on Build GRCh38
rs559269063 Chr.19: 15020799-15020799 on Build GRCh38
rs540119872 Chr.19: 15020728-15020728 on Build GRCh38
rs567180451 Chr.19: 15020421-15020421 on Build GRCh38
rs770294323 Chr.19: 15020441-15020443 on Build GRCh38
rs369426784 Chr.19: 15020470-15020470 on Build GRCh38
rs73929534 Chr.19: 15020783-15020783 on Build GRCh38
rs145380555 Chr.19: 15020649-15020649 on Build GRCh38
rs558410878 Chr.19: 15020730-15020730 on Build GRCh38
rs761244031 Chr.19: 15020707-15020707 on Build GRCh38
rs540786013 Chr.19: 15020785-15020785 on Build GRCh38
rs753056711 Chr.19: 15020476-15020476 on Build GRCh38
rs141762895 Chr.19: 15020632-15020632 on Build GRCh38
rs929115040 Chr.19: 15020768-15020768 on Build GRCh38
rs747751880 Chr.19: 15020592-15020592 on Build GRCh38
rs762471697 Chr.19: 15020412-15020412 on Build GRCh38
rs754133085 Chr.19: 15020461-15020461 on Build GRCh38
rs754881095 Chr.19: 15020843-15020844 on Build GRCh38
rs768755763 Chr.19: 15020689-15020689 on Build GRCh38
rs374386622 Chr.19: 15020811-15020811 on Build GRCh38
rs1027347717 Chr.19: 15020591-15020591 on Build GRCh38
rs200108697 Chr.19: 15020536-15020536 on Build GRCh38
rs770773192 Chr.19: 15020600-15020600 on Build GRCh38
rs566310857 Chr.19: 15020629-15020629 on Build GRCh38
rs766331643 Chr.19: 15020455-15020455 on Build GRCh38
rs4405675 Chr.19: 15020449-15020449 on Build GRCh38
rs748798959 Chr.19: 15020578-15020578 on Build GRCh38
rs548822665 Chr.19: 15020664-15020664 on Build GRCh38
rs758787413 Chr.19: 15020564-15020564 on Build GRCh38
rs369689058 Chr.19: 15020605-15020605 on Build GRCh38
rs760501981 Chr.19: 15020534-15020534 on Build GRCh38
rs374935812 Chr.19: 15020685-15020685 on Build GRCh38
rs774562123 Chr.19: 15020691-15020691 on Build GRCh38
rs1048902461 Chr.19: 15020779-15020779 on Build GRCh38
rs897776766 Chr.19: 15020762-15020762 on Build GRCh38

View Full Product Details