Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15226550-15227094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHX3
Entrez Gene ID
79852
Gene Name
epoxide hydrolase 3
Gene Aliases
ABHD9, EH3
UniGene
Hs.156457
Gene Chromosome Location
Chr.19: 15226919-15233047 on Build GRCh38
Transcripts
NM_001142886.1 NM_024794.2

SNP Detail

SNP ID Chromosome Location
rs893261742 Chr.19: 15227021-15227021 on Build GRCh38
rs139694209 Chr.19: 15227008-15227008 on Build GRCh38
rs940443240 Chr.19: 15226897-15226898 on Build GRCh38
rs987644523 Chr.19: 15226582-15226582 on Build GRCh38
rs373884884 Chr.19: 15226811-15226811 on Build GRCh38
rs930708120 Chr.19: 15226686-15226686 on Build GRCh38
rs958337015 Chr.19: 15226844-15226844 on Build GRCh38
rs972336449 Chr.19: 15226898-15226898 on Build GRCh38
rs143104329 Chr.19: 15226601-15226601 on Build GRCh38
rs749687698 Chr.19: 15226967-15226967 on Build GRCh38
rs1019642019 Chr.19: 15226963-15226963 on Build GRCh38
rs1054459581 Chr.19: 15227000-15227000 on Build GRCh38
rs572412654 Chr.19: 15227017-15227017 on Build GRCh38
rs562696462 Chr.19: 15226652-15226652 on Build GRCh38
rs531570884 Chr.19: 15226660-15226660 on Build GRCh38
rs940824944 Chr.19: 15226612-15226612 on Build GRCh38
rs901792135 Chr.19: 15226580-15226580 on Build GRCh38
rs953025382 Chr.19: 15226875-15226875 on Build GRCh38
rs775771887 Chr.19: 15226697-15226697 on Build GRCh38
rs902322072 Chr.19: 15226818-15226818 on Build GRCh38
rs1010862612 Chr.19: 15226696-15226696 on Build GRCh38
rs923110656 Chr.19: 15226959-15226959 on Build GRCh38
rs554566312 Chr.19: 15226813-15226813 on Build GRCh38
rs1009116078 Chr.19: 15226895-15226895 on Build GRCh38
rs542407167 Chr.19: 15226627-15226627 on Build GRCh38
rs571589021 Chr.19: 15226728-15226728 on Build GRCh38
rs191668412 Chr.19: 15226769-15226769 on Build GRCh38
rs547771084 Chr.19: 15226780-15226780 on Build GRCh38
rs533509644 Chr.19: 15227045-15227045 on Build GRCh38
rs150605028 Chr.19: 15226861-15226861 on Build GRCh38
rs184215124 Chr.19: 15226843-15226843 on Build GRCh38
rs867044979 Chr.19: 15226757-15226757 on Build GRCh38
rs909004870 Chr.19: 15226891-15226891 on Build GRCh38
rs766951093 Chr.19: 15227041-15227041 on Build GRCh38
rs1033059145 Chr.19: 15226622-15226622 on Build GRCh38
rs937262080 Chr.19: 15226727-15226727 on Build GRCh38
rs1033995401 Chr.19: 15226812-15226812 on Build GRCh38
rs1050332358 Chr.19: 15226966-15226966 on Build GRCh38
rs972677550 Chr.19: 15226991-15226991 on Build GRCh38
rs528988783 Chr.19: 15226606-15226606 on Build GRCh38
rs905155699 Chr.19: 15226724-15226724 on Build GRCh38
rs920716883 Chr.19: 15226671-15226671 on Build GRCh38
rs764714504 Chr.19: 15226766-15226766 on Build GRCh38
rs757291 Chr.19: 15226974-15226974 on Build GRCh38
rs1057408244 Chr.19: 15226734-15226734 on Build GRCh38
rs551680936 Chr.19: 15226683-15226683 on Build GRCh38
rs773928122 Chr.19: 15227046-15227046 on Build GRCh38
rs186616189 Chr.19: 15226873-15226873 on Build GRCh38
rs998430934 Chr.19: 15226824-15226824 on Build GRCh38
rs1028616264 Chr.19: 15226871-15226871 on Build GRCh38
rs972087847 Chr.19: 15226797-15226797 on Build GRCh38
rs770128478 Chr.19: 15226575-15226575 on Build GRCh38
rs1020446355 Chr.19: 15226758-15226758 on Build GRCh38
rs980829045 Chr.19: 15227065-15227065 on Build GRCh38
rs371030528 Chr.19: 15226626-15226626 on Build GRCh38
rs952111342 Chr.19: 15227049-15227049 on Build GRCh38

View Full Product Details