Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15537538-15538036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F22
Entrez Gene ID
126410
Gene Name
cytochrome P450 family 4 subfamily F member 22
Gene Aliases
ARCI5, INLNE, LI3
UniGene
Hs.156452
Gene Chromosome Location
Chr.19: 15508487-15552317 on Build GRCh38
Transcripts
NM_173483.3 XM_011527692.2 XM_011527693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524715 c.565C>A p.L189M Chr.19: 15537887-15537887 on Build GRCh38
3283844 c.534C>T p.S178S Chr.19: 15537647-15537647 on Build GRCh38
7159511 c.645C>G p.V215V Chr.19: 15537967-15537967 on Build GRCh38
3283845 c.538G>A p.D180N Chr.19: 15537651-15537651 on Build GRCh38
6524716 c.622C>A p.L208M Chr.19: 15537944-15537944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531670218 Chr.19: 15537986-15537986 on Build GRCh38
rs774213936 Chr.19: 15537677-15537677 on Build GRCh38
rs768585238 Chr.19: 15537675-15537675 on Build GRCh38
rs770695228 Chr.19: 15538010-15538010 on Build GRCh38
rs577229558 Chr.19: 15537891-15537891 on Build GRCh38
rs576705212 Chr.19: 15537814-15537814 on Build GRCh38
rs750743002 Chr.19: 15537606-15537606 on Build GRCh38
rs751335732 Chr.19: 15537559-15537559 on Build GRCh38
rs745717138 Chr.19: 15537999-15537999 on Build GRCh38
rs534242787 Chr.19: 15537704-15537704 on Build GRCh38
rs199641250 Chr.19: 15537583-15537583 on Build GRCh38
rs777866266 Chr.19: 15538004-15538004 on Build GRCh38
rs755471652 Chr.19: 15537634-15537634 on Build GRCh38
rs868030805 Chr.19: 15537629-15537629 on Build GRCh38
rs759543514 Chr.19: 15537845-15537845 on Build GRCh38
rs147748244 Chr.19: 15537974-15537974 on Build GRCh38
rs78418003 Chr.19: 15537976-15537976 on Build GRCh38
rs758503533 Chr.19: 15537687-15537688 on Build GRCh38
rs935078250 Chr.19: 15537834-15537834 on Build GRCh38
rs372867266 Chr.19: 15537709-15537709 on Build GRCh38
rs374464817 Chr.19: 15537596-15537596 on Build GRCh38
rs946071699 Chr.19: 15537761-15537761 on Build GRCh38
rs760550354 Chr.19: 15537917-15537917 on Build GRCh38
rs143598221 Chr.19: 15537799-15537799 on Build GRCh38
rs750472966 Chr.19: 15537561-15537561 on Build GRCh38
rs753177417 Chr.19: 15537946-15537946 on Build GRCh38
rs747690640 Chr.19: 15537996-15537996 on Build GRCh38
rs114980833 Chr.19: 15537560-15537560 on Build GRCh38
rs752264797 Chr.19: 15537829-15537829 on Build GRCh38
rs573750979 Chr.19: 15537785-15537785 on Build GRCh38
rs761697748 Chr.19: 15537691-15537691 on Build GRCh38
rs756321971 Chr.19: 15537562-15537562 on Build GRCh38
rs868549151 Chr.19: 15537923-15537923 on Build GRCh38
rs759653194 Chr.19: 15537925-15537925 on Build GRCh38
rs1005088726 Chr.19: 15537713-15537713 on Build GRCh38
rs374190832 Chr.19: 15537979-15537979 on Build GRCh38
rs141755148 Chr.19: 15537605-15537605 on Build GRCh38
rs780704118 Chr.19: 15537563-15537563 on Build GRCh38
rs370400762 Chr.19: 15537670-15537670 on Build GRCh38
rs778308984 Chr.19: 15537986-15537986 on Build GRCh38
rs373223970 Chr.19: 15537613-15537613 on Build GRCh38
rs370285007 Chr.19: 15537854-15537854 on Build GRCh38
rs1015071704 Chr.19: 15537972-15537972 on Build GRCh38
rs770500550 Chr.19: 15537579-15537579 on Build GRCh38
rs368781436 Chr.19: 15537882-15537882 on Build GRCh38
rs766540379 Chr.19: 15537826-15537826 on Build GRCh38
rs141382984 Chr.19: 15537576-15537576 on Build GRCh38
rs556566002 Chr.19: 15537807-15537807 on Build GRCh38
rs775270460 Chr.19: 15537586-15537586 on Build GRCh38
rs772155840 Chr.19: 15537907-15537907 on Build GRCh38
rs148063219 Chr.19: 15537804-15537804 on Build GRCh38
rs776771681 Chr.19: 15537924-15537924 on Build GRCh38
rs753883685 Chr.19: 15537621-15537621 on Build GRCh38
rs758343094 Chr.19: 15537998-15537998 on Build GRCh38
rs946591987 Chr.19: 15537873-15537873 on Build GRCh38
rs199892192 Chr.19: 15537989-15537989 on Build GRCh38
rs1003060629 Chr.19: 15537578-15537578 on Build GRCh38
rs749840377 Chr.19: 15537566-15537566 on Build GRCh38
rs16980531 Chr.19: 15537645-15537645 on Build GRCh38
rs982261190 Chr.19: 15537818-15537818 on Build GRCh38
rs758816247 Chr.19: 15537648-15537648 on Build GRCh38
rs371097842 Chr.19: 15537981-15537981 on Build GRCh38
rs780816103 Chr.19: 15537649-15537649 on Build GRCh38
rs776879263 Chr.19: 15538011-15538011 on Build GRCh38
rs79603814 Chr.19: 15537909-15537909 on Build GRCh38
rs990606848 Chr.19: 15537990-15537990 on Build GRCh38
rs758728427 Chr.19: 15537856-15537856 on Build GRCh38
rs11666601 Chr.19: 15537904-15537904 on Build GRCh38
rs1005392453 Chr.19: 15537919-15537919 on Build GRCh38
rs142364533 Chr.19: 15537881-15537881 on Build GRCh38
rs778065865 Chr.19: 15537865-15537865 on Build GRCh38
rs187004457 Chr.19: 15537598-15537598 on Build GRCh38
rs192304348 Chr.19: 15537757-15537757 on Build GRCh38
rs1055995688 Chr.19: 15537811-15537811 on Build GRCh38
rs368813073 Chr.19: 15537590-15537590 on Build GRCh38
rs201743364 Chr.19: 15537798-15537798 on Build GRCh38
rs746123334 Chr.19: 15538014-15538014 on Build GRCh38
rs779288178 Chr.19: 15537574-15537574 on Build GRCh38
rs771126991 Chr.19: 15537918-15537918 on Build GRCh38
rs562637818 Chr.19: 15537957-15537957 on Build GRCh38
rs140657557 Chr.19: 15537840-15537840 on Build GRCh38
rs539896338 Chr.19: 15537900-15537900 on Build GRCh38
rs973011727 Chr.19: 15537558-15537558 on Build GRCh38
rs776275777 Chr.19: 15537580-15537580 on Build GRCh38
rs201717667 Chr.19: 15537927-15537927 on Build GRCh38
rs571757470 Chr.19: 15537656-15537656 on Build GRCh38
rs761196649 Chr.19: 15537582-15537582 on Build GRCh38
rs748818073 Chr.19: 15537889-15537889 on Build GRCh38
rs766528561 Chr.19: 15537614-15537614 on Build GRCh38
rs1010835839 Chr.19: 15537627-15537627 on Build GRCh38
rs764977269 Chr.19: 15537805-15537805 on Build GRCh38
rs746783510 Chr.19: 15538007-15538007 on Build GRCh38
rs146036982 Chr.19: 15537571-15537571 on Build GRCh38
rs772035186 Chr.19: 15537694-15537694 on Build GRCh38
rs755273482 Chr.19: 15537573-15537573 on Build GRCh38
rs151269464 Chr.19: 15537903-15537903 on Build GRCh38
rs754310081 Chr.19: 15537736-15537736 on Build GRCh38
rs374509524 Chr.19: 15537866-15537866 on Build GRCh38
rs569166154 Chr.19: 15537597-15537597 on Build GRCh38
rs751954643 Chr.19: 15537876-15537876 on Build GRCh38
rs148454445 Chr.19: 15537652-15537652 on Build GRCh38
rs377373101 Chr.19: 15537647-15537647 on Build GRCh38

View Full Product Details