Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15652405-15652937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F3
Entrez Gene ID
4051
Gene Name
cytochrome P450 family 4 subfamily F member 3
Gene Aliases
CPF3, CYP4F, LTB4H
UniGene
Hs.106242
Gene Chromosome Location
Chr.19: 15640897-15660760 on Build GRCh38
Transcripts
NM_000896.2 NM_001199208.1 NM_001199209.1 XM_005259911.4 XM_011528014.2 XM_017026814.1 XM_017026815.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5590456 c.1020C>T p.V340V Chr.19: 15652857-15652857 on Build GRCh38
6200758 c.942C>T p.S314S Chr.19: 15652592-15652592 on Build GRCh38
5555316 c.1006G>A p.G336S Chr.19: 15652843-15652843 on Build GRCh38
243947 c.953T>C p.I318T Chr.19: 15652603-15652603 on Build GRCh38
2155100 c.955A>T p.R319* Chr.19: 15652605-15652605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779126203 Chr.19: 15652888-15652888 on Build GRCh38
rs894429239 Chr.19: 15652731-15652731 on Build GRCh38
rs201829745 Chr.19: 15652880-15652880 on Build GRCh38
rs772515507 Chr.19: 15652816-15652816 on Build GRCh38
rs993402841 Chr.19: 15652520-15652521 on Build GRCh38
rs149389323 Chr.19: 15652635-15652635 on Build GRCh38
rs894140945 Chr.19: 15652467-15652467 on Build GRCh38
rs746070418 Chr.19: 15652602-15652602 on Build GRCh38
rs532340633 Chr.19: 15652863-15652863 on Build GRCh38
rs762429530 Chr.19: 15652642-15652642 on Build GRCh38
rs757921130 Chr.19: 15652567-15652567 on Build GRCh38
rs755949961 Chr.19: 15652775-15652775 on Build GRCh38
rs112083753 Chr.19: 15652917-15652917 on Build GRCh38
rs1045821807 Chr.19: 15652733-15652733 on Build GRCh38
rs768858084 Chr.19: 15652634-15652634 on Build GRCh38
rs977638859 Chr.19: 15652721-15652721 on Build GRCh38
rs748123808 Chr.19: 15652894-15652894 on Build GRCh38
rs775008796 Chr.19: 15652539-15652539 on Build GRCh38
rs768805172 Chr.19: 15652805-15652805 on Build GRCh38
rs756991110 Chr.19: 15652456-15652456 on Build GRCh38
rs767299743 Chr.19: 15652860-15652860 on Build GRCh38
rs894594037 Chr.19: 15652427-15652427 on Build GRCh38
rs984571381 Chr.19: 15652709-15652709 on Build GRCh38
rs897766243 Chr.19: 15652518-15652518 on Build GRCh38
rs2683038 Chr.19: 15652460-15652460 on Build GRCh38
rs59790611 Chr.19: 15652543-15652544 on Build GRCh38
rs531235045 Chr.19: 15652541-15652541 on Build GRCh38
rs763564905 Chr.19: 15652846-15652846 on Build GRCh38
rs760885195 Chr.19: 15652826-15652826 on Build GRCh38
rs752351655 Chr.19: 15652659-15652659 on Build GRCh38
rs764743105 Chr.19: 15652657-15652657 on Build GRCh38
rs760713878 Chr.19: 15652916-15652916 on Build GRCh38
rs762588767 Chr.19: 15652905-15652905 on Build GRCh38
rs755821342 Chr.19: 15652877-15652877 on Build GRCh38
rs374530263 Chr.19: 15652675-15652675 on Build GRCh38
rs376212056 Chr.19: 15652636-15652636 on Build GRCh38
rs755288193 Chr.19: 15652887-15652887 on Build GRCh38
rs576785602 Chr.19: 15652442-15652442 on Build GRCh38
rs775472639 Chr.19: 15652534-15652534 on Build GRCh38
rs2733750 Chr.19: 15652767-15652767 on Build GRCh38
rs957841219 Chr.19: 15652557-15652557 on Build GRCh38
rs928797264 Chr.19: 15652436-15652436 on Build GRCh38
rs751134520 Chr.19: 15652849-15652849 on Build GRCh38
rs761159834 Chr.19: 15652909-15652909 on Build GRCh38
rs773519059 Chr.19: 15652648-15652648 on Build GRCh38
rs937918916 Chr.19: 15652712-15652712 on Build GRCh38
rs763547195 Chr.19: 15652536-15652536 on Build GRCh38
rs751477800 Chr.19: 15652670-15652670 on Build GRCh38
rs757864219 Chr.19: 15652664-15652664 on Build GRCh38
rs950123735 Chr.19: 15652732-15652732 on Build GRCh38
rs767898542 Chr.19: 15652644-15652644 on Build GRCh38
rs1055840450 Chr.19: 15652724-15652724 on Build GRCh38
rs919104752 Chr.19: 15652688-15652688 on Build GRCh38
rs552131266 Chr.19: 15652666-15652666 on Build GRCh38
rs201402323 Chr.19: 15652900-15652900 on Build GRCh38
rs767082417 Chr.19: 15652910-15652910 on Build GRCh38
rs1018794284 Chr.19: 15652867-15652868 on Build GRCh38
rs759217793 Chr.19: 15652653-15652653 on Build GRCh38
rs1037401626 Chr.19: 15652769-15652769 on Build GRCh38
rs764754837 Chr.19: 15652538-15652538 on Build GRCh38
rs777021261 Chr.19: 15652903-15652903 on Build GRCh38
rs113293324 Chr.19: 15652531-15652531 on Build GRCh38
rs1024482581 Chr.19: 15652439-15652439 on Build GRCh38
rs762644240 Chr.19: 15652841-15652841 on Build GRCh38
rs368774644 Chr.19: 15652815-15652815 on Build GRCh38
rs756287959 Chr.19: 15652728-15652728 on Build GRCh38
rs28371484 Chr.19: 15652710-15652710 on Build GRCh38
rs746982348 Chr.19: 15652589-15652589 on Build GRCh38
rs772202060 Chr.19: 15652895-15652895 on Build GRCh38
rs929118395 Chr.19: 15652707-15652707 on Build GRCh38
rs754630520 Chr.19: 15652544-15652544 on Build GRCh38
rs1805041 Chr.19: 15652881-15652881 on Build GRCh38
rs749548251 Chr.19: 15652788-15652788 on Build GRCh38
rs548001447 Chr.19: 15652752-15652752 on Build GRCh38
rs770349262 Chr.19: 15652625-15652626 on Build GRCh38
rs773787198 Chr.19: 15652817-15652817 on Build GRCh38
rs775696171 Chr.19: 15652621-15652621 on Build GRCh38
rs1005856472 Chr.19: 15652473-15652473 on Build GRCh38
rs777621665 Chr.19: 15652575-15652575 on Build GRCh38
rs573272995 Chr.19: 15652493-15652493 on Build GRCh38
rs780899134 Chr.19: 15652593-15652593 on Build GRCh38
rs562238744 Chr.19: 15652540-15652540 on Build GRCh38
rs28371483 Chr.19: 15652592-15652592 on Build GRCh38
rs771316362 Chr.19: 15652832-15652832 on Build GRCh38
rs761794826 Chr.19: 15652850-15652850 on Build GRCh38
rs774066427 Chr.19: 15652908-15652908 on Build GRCh38
rs185622630 Chr.19: 15652740-15652740 on Build GRCh38
rs750243030 Chr.19: 15652774-15652774 on Build GRCh38
rs543666112 Chr.19: 15652483-15652483 on Build GRCh38
rs749322639 Chr.19: 15652626-15652626 on Build GRCh38
rs564762942 Chr.19: 15652604-15652604 on Build GRCh38
rs199825874 Chr.19: 15652906-15652906 on Build GRCh38
rs780352932 Chr.19: 15652779-15652779 on Build GRCh38
rs1055481125 Chr.19: 15652736-15652736 on Build GRCh38
rs764803164 Chr.19: 15652550-15652550 on Build GRCh38
rs141822395 Chr.19: 15652901-15652901 on Build GRCh38
rs775074714 Chr.19: 15652836-15652836 on Build GRCh38
rs752265414 Chr.19: 15652556-15652556 on Build GRCh38
rs371375686 Chr.19: 15652671-15652671 on Build GRCh38
rs746214273 Chr.19: 15652567-15652567 on Build GRCh38
rs781034491 Chr.19: 15652677-15652677 on Build GRCh38
rs1805042 Chr.19: 15652911-15652911 on Build GRCh38
rs779179698 Chr.19: 15652811-15652811 on Build GRCh38
rs975770423 Chr.19: 15652640-15652640 on Build GRCh38
rs2733749 Chr.19: 15652438-15652438 on Build GRCh38

View Full Product Details