Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16190780-16191275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM32A
Entrez Gene ID
26017
Gene Name
family with sequence similarity 32 member A
Gene Aliases
OTAG-12, OTAG12
UniGene
Hs.631614
Gene Chromosome Location
Chr.19: 16185184-16192046 on Build GRCh38
Transcripts
NM_014077.3 XM_011527898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5464413 c.292A>C p.T98P Chr.19: 16190908-16190908 on Build GRCh38
6283510 c.300G>A p.T100T Chr.19: 16190916-16190916 on Build GRCh38
5004241 c.271-1G>T p.? Chr.19: 16190886-16190886 on Build GRCh38
7367368 c.280A>G p.R94G Chr.19: 16190896-16190896 on Build GRCh38
4543234 c.333G>A p.T111T Chr.19: 16190949-16190949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761385716 Chr.19: 16190910-16190910 on Build GRCh38
rs766123731 Chr.19: 16190949-16190949 on Build GRCh38
rs1023333678 Chr.19: 16190809-16190809 on Build GRCh38
rs768229802 Chr.19: 16190885-16190885 on Build GRCh38
rs11541648 Chr.19: 16191031-16191031 on Build GRCh38
rs943035582 Chr.19: 16191143-16191143 on Build GRCh38
rs10412228 Chr.19: 16190874-16190874 on Build GRCh38
rs934846062 Chr.19: 16191227-16191227 on Build GRCh38
rs563514710 Chr.19: 16190811-16190811 on Build GRCh38
rs549016714 Chr.19: 16191087-16191087 on Build GRCh38
rs771423861 Chr.19: 16190987-16190987 on Build GRCh38
rs1012884653 Chr.19: 16191240-16191240 on Build GRCh38
rs974816076 Chr.19: 16191148-16191148 on Build GRCh38
rs373088103 Chr.19: 16190925-16190925 on Build GRCh38
rs771832203 Chr.19: 16190843-16190843 on Build GRCh38
rs199874860 Chr.19: 16191237-16191238 on Build GRCh38
rs372809693 Chr.19: 16191004-16191004 on Build GRCh38
rs547790142 Chr.19: 16190922-16190922 on Build GRCh38
rs558776473 Chr.19: 16191007-16191007 on Build GRCh38
rs1022882268 Chr.19: 16191009-16191009 on Build GRCh38
rs192940284 Chr.19: 16191228-16191228 on Build GRCh38
rs778800385 Chr.19: 16190853-16190853 on Build GRCh38
rs57818321 Chr.19: 16190992-16190992 on Build GRCh38
rs34796192 Chr.19: 16191253-16191254 on Build GRCh38
rs550101685 Chr.19: 16190984-16190984 on Build GRCh38
rs752026877 Chr.19: 16191193-16191193 on Build GRCh38
rs755088587 Chr.19: 16190849-16190849 on Build GRCh38
rs764548328 Chr.19: 16191187-16191187 on Build GRCh38
rs754102824 Chr.19: 16190957-16190957 on Build GRCh38
rs183916631 Chr.19: 16191051-16191051 on Build GRCh38
rs987167784 Chr.19: 16191124-16191124 on Build GRCh38
rs187684892 Chr.19: 16190863-16190863 on Build GRCh38
rs188468975 Chr.19: 16191194-16191194 on Build GRCh38
rs529177823 Chr.19: 16191057-16191057 on Build GRCh38
rs147062774 Chr.19: 16190867-16190867 on Build GRCh38
rs749463021 Chr.19: 16190848-16190848 on Build GRCh38
rs375817075 Chr.19: 16191059-16191059 on Build GRCh38
rs767484189 Chr.19: 16190916-16190916 on Build GRCh38
rs911674993 Chr.19: 16191130-16191130 on Build GRCh38
rs541582307 Chr.19: 16191178-16191178 on Build GRCh38
rs777583121 Chr.19: 16190978-16190978 on Build GRCh38
rs1026583152 Chr.19: 16191057-16191057 on Build GRCh38
rs8102293 Chr.19: 16190854-16190854 on Build GRCh38
rs56084066 Chr.19: 16190982-16190982 on Build GRCh38
rs768262940 Chr.19: 16190991-16190991 on Build GRCh38
rs554917433 Chr.19: 16191034-16191034 on Build GRCh38
rs547586683 Chr.19: 16190958-16190958 on Build GRCh38
rs758527349 Chr.19: 16190968-16190968 on Build GRCh38
rs746234491 Chr.19: 16190989-16190989 on Build GRCh38
rs368543360 Chr.19: 16191002-16191002 on Build GRCh38
rs148445686 Chr.19: 16190896-16190896 on Build GRCh38
rs115233434 Chr.19: 16191159-16191159 on Build GRCh38
rs771524580 Chr.19: 16191003-16191003 on Build GRCh38
rs778443813 Chr.19: 16190969-16190969 on Build GRCh38
rs375373984 Chr.19: 16190864-16190864 on Build GRCh38
rs370495221 Chr.19: 16190855-16190855 on Build GRCh38
rs755873790 Chr.19: 16190839-16190839 on Build GRCh38
rs1043208815 Chr.19: 16191195-16191195 on Build GRCh38
rs201483276 Chr.19: 16190964-16190964 on Build GRCh38
rs566190835 Chr.19: 16191014-16191014 on Build GRCh38
rs200056983 Chr.19: 16190845-16190845 on Build GRCh38
rs543664468 Chr.19: 16191131-16191131 on Build GRCh38
rs779190046 Chr.19: 16190961-16190961 on Build GRCh38
rs895808694 Chr.19: 16191005-16191005 on Build GRCh38
rs781645378 Chr.19: 16190988-16190988 on Build GRCh38
rs947640731 Chr.19: 16191166-16191166 on Build GRCh38
rs903333883 Chr.19: 16191206-16191206 on Build GRCh38
rs200327546 Chr.19: 16190878-16190878 on Build GRCh38

View Full Product Details