Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16234118-16234610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1M1
Entrez Gene ID
8907
Gene Name
adaptor related protein complex 1 mu 1 subunit
Gene Aliases
AP47, CLAPM2, CLTNM, MU-1A
UniGene
Hs.71040
Gene Chromosome Location
Chr.19: 16197854-16235345 on Build GRCh38
Transcripts
NM_001130524.1 NM_032493.3 XM_017027420.1 XM_017027421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6601554 c.1229G>A p.R410H Chr.19: 16234254-16234254 on Build GRCh38
5486006 c.1278G>T p.Q426H Chr.19: 16234267-16234267 on Build GRCh38
73689 c.1224G>C p.W408C Chr.19: 16234249-16234249 on Build GRCh38
5589681 c.1191C>T p.I397I Chr.19: 16234216-16234216 on Build GRCh38
3787573 c.1216C>T p.L406L Chr.19: 16234241-16234241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758037347 Chr.19: 16234280-16234280 on Build GRCh38
rs751077992 Chr.19: 16234436-16234436 on Build GRCh38
rs368068984 Chr.19: 16234417-16234417 on Build GRCh38
rs903539831 Chr.19: 16234510-16234510 on Build GRCh38
rs917694915 Chr.19: 16234516-16234516 on Build GRCh38
rs756260692 Chr.19: 16234538-16234538 on Build GRCh38
rs200986619 Chr.19: 16234168-16234168 on Build GRCh38
rs751075343 Chr.19: 16234302-16234302 on Build GRCh38
rs780328990 Chr.19: 16234369-16234369 on Build GRCh38
rs201926045 Chr.19: 16234308-16234308 on Build GRCh38
rs762285943 Chr.19: 16234267-16234267 on Build GRCh38
rs915198403 Chr.19: 16234541-16234541 on Build GRCh38
rs1024378201 Chr.19: 16234584-16234584 on Build GRCh38
rs535215936 Chr.19: 16234501-16234501 on Build GRCh38
rs546425522 Chr.19: 16234464-16234464 on Build GRCh38
rs774363558 Chr.19: 16234381-16234381 on Build GRCh38
rs990552113 Chr.19: 16234489-16234489 on Build GRCh38
rs527394286 Chr.19: 16234484-16234484 on Build GRCh38
rs375678399 Chr.19: 16234298-16234298 on Build GRCh38
rs920618600 Chr.19: 16234397-16234397 on Build GRCh38
rs897774 Chr.19: 16234288-16234288 on Build GRCh38
rs751351568 Chr.19: 16234455-16234455 on Build GRCh38
rs201411972 Chr.19: 16234204-16234204 on Build GRCh38
rs745625654 Chr.19: 16234182-16234182 on Build GRCh38
rs374061291 Chr.19: 16234400-16234400 on Build GRCh38
rs201201130 Chr.19: 16234388-16234388 on Build GRCh38
rs375321256 Chr.19: 16234459-16234459 on Build GRCh38
rs202192686 Chr.19: 16234293-16234293 on Build GRCh38
rs781252643 Chr.19: 16234309-16234309 on Build GRCh38
rs937678224 Chr.19: 16234487-16234487 on Build GRCh38
rs771138980 Chr.19: 16234395-16234395 on Build GRCh38
rs755934381 Chr.19: 16234316-16234316 on Build GRCh38
rs886291636 Chr.19: 16234263-16234263 on Build GRCh38
rs148649169 Chr.19: 16234379-16234379 on Build GRCh38
rs781199249 Chr.19: 16234174-16234174 on Build GRCh38
rs770189489 Chr.19: 16234201-16234201 on Build GRCh38
rs1022352649 Chr.19: 16234499-16234499 on Build GRCh38
rs745678992 Chr.19: 16234315-16234315 on Build GRCh38
rs536580317 Chr.19: 16234290-16234290 on Build GRCh38
rs374418377 Chr.19: 16234292-16234292 on Build GRCh38
rs768010248 Chr.19: 16234284-16234284 on Build GRCh38
rs755987562 Chr.19: 16234445-16234445 on Build GRCh38
rs967340210 Chr.19: 16234171-16234171 on Build GRCh38
rs767593182 Chr.19: 16234439-16234439 on Build GRCh38
rs1054728151 Chr.19: 16234589-16234589 on Build GRCh38
rs771462229 Chr.19: 16234478-16234478 on Build GRCh38
rs1046008236 Chr.19: 16234360-16234360 on Build GRCh38
rs748197219 Chr.19: 16234382-16234382 on Build GRCh38
rs34850342 Chr.19: 16234264-16234264 on Build GRCh38
rs1045957204 Chr.19: 16234540-16234540 on Build GRCh38
rs527247445 Chr.19: 16234231-16234231 on Build GRCh38
rs567076354 Chr.19: 16234463-16234463 on Build GRCh38
rs998891779 Chr.19: 16234173-16234173 on Build GRCh38
rs976353852 Chr.19: 16234456-16234456 on Build GRCh38
rs761029566 Chr.19: 16234393-16234393 on Build GRCh38
rs753665081 Chr.19: 16234458-16234458 on Build GRCh38
rs1014453661 Chr.19: 16234559-16234559 on Build GRCh38
rs950273234 Chr.19: 16234354-16234354 on Build GRCh38
rs369847063 Chr.19: 16234474-16234474 on Build GRCh38
rs763840006 Chr.19: 16234303-16234305 on Build GRCh38
rs371038503 Chr.19: 16234376-16234376 on Build GRCh38
rs370959225 Chr.19: 16234219-16234219 on Build GRCh38
rs777765779 Chr.19: 16234167-16234167 on Build GRCh38
rs376948295 Chr.19: 16234207-16234207 on Build GRCh38
rs577475020 Chr.19: 16234453-16234453 on Build GRCh38
rs377712548 Chr.19: 16234444-16234444 on Build GRCh38
rs201926259 Chr.19: 16234158-16234158 on Build GRCh38
rs557581215 Chr.19: 16234404-16234404 on Build GRCh38
rs540652538 Chr.19: 16234543-16234543 on Build GRCh38
rs554903220 Chr.19: 16234551-16234551 on Build GRCh38
rs370346641 Chr.19: 16234365-16234365 on Build GRCh38
rs944577348 Chr.19: 16234467-16234467 on Build GRCh38
rs764842453 Chr.19: 16234155-16234155 on Build GRCh38
rs754695250 Chr.19: 16234149-16234149 on Build GRCh38
rs374022955 Chr.19: 16234157-16234157 on Build GRCh38
rs948762989 Chr.19: 16234140-16234141 on Build GRCh38
rs917377037 Chr.19: 16234383-16234383 on Build GRCh38
rs1055301875 Chr.19: 16234319-16234319 on Build GRCh38
rs930532373 Chr.19: 16234422-16234422 on Build GRCh38
rs566168350 Chr.19: 16234331-16234331 on Build GRCh38
rs564803280 Chr.19: 16234465-16234465 on Build GRCh38
rs534675236 Chr.19: 16234361-16234361 on Build GRCh38
rs370397966 Chr.19: 16234392-16234392 on Build GRCh38

View Full Product Details