Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16355601-16356106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15L1
Entrez Gene ID
58513
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
Gene Aliases
EPS15R
UniGene
Hs.654639
Gene Chromosome Location
Chr.19: 16355244-16472012 on Build GRCh38
Transcripts
NM_001258374.1 NR_047665.1 XM_017027086.1 XM_017027089.1 XM_017027090.1 XM_017027091.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6524725 c.2701G>T p.A901S Chr.19: 16355737-16355737 on Build GRCh38
4154074 c.2617G>A p.A873T Chr.19: 16355821-16355821 on Build GRCh38
7205586 c.2632G>A p.E878K Chr.19: 16355806-16355806 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs76169759 Chr.19: 16356036-16356036 on Build GRCh38
rs751495521 Chr.19: 16355830-16355830 on Build GRCh38
rs1005141499 Chr.19: 16356017-16356017 on Build GRCh38
rs773540636 Chr.19: 16355710-16355710 on Build GRCh38
rs757239390 Chr.19: 16355880-16355880 on Build GRCh38
rs763795002 Chr.19: 16355860-16355861 on Build GRCh38
rs762567661 Chr.19: 16355683-16355683 on Build GRCh38
rs752166419 Chr.19: 16355755-16355755 on Build GRCh38
rs914928594 Chr.19: 16355815-16355815 on Build GRCh38
rs1025327538 Chr.19: 16355923-16355923 on Build GRCh38
rs994951586 Chr.19: 16355696-16355696 on Build GRCh38
rs946855903 Chr.19: 16355978-16355978 on Build GRCh38
rs1023618792 Chr.19: 16356057-16356057 on Build GRCh38
rs981935544 Chr.19: 16355752-16355752 on Build GRCh38
rs536140146 Chr.19: 16355807-16355807 on Build GRCh38
rs762462486 Chr.19: 16355768-16355768 on Build GRCh38
rs995277146 Chr.19: 16356056-16356056 on Build GRCh38
rs780064994 Chr.19: 16355670-16355670 on Build GRCh38
rs918221089 Chr.19: 16355631-16355631 on Build GRCh38
rs993851783 Chr.19: 16355866-16355866 on Build GRCh38
rs139944694 Chr.19: 16355970-16355970 on Build GRCh38
rs780737254 Chr.19: 16356074-16356074 on Build GRCh38
rs749388034 Chr.19: 16355962-16355962 on Build GRCh38
rs955068618 Chr.19: 16355927-16355927 on Build GRCh38
rs1016736633 Chr.19: 16355759-16355759 on Build GRCh38
rs567265352 Chr.19: 16355764-16355764 on Build GRCh38
rs1042523950 Chr.19: 16356009-16356009 on Build GRCh38
rs564741577 Chr.19: 16355674-16355674 on Build GRCh38
rs369783739 Chr.19: 16355954-16355954 on Build GRCh38
rs924012978 Chr.19: 16356012-16356012 on Build GRCh38
rs1012498016 Chr.19: 16355753-16355753 on Build GRCh38
rs764162027 Chr.19: 16355773-16355773 on Build GRCh38
rs986420200 Chr.19: 16355928-16355928 on Build GRCh38
rs55712949 Chr.19: 16355633-16355633 on Build GRCh38
rs956628331 Chr.19: 16355778-16355778 on Build GRCh38
rs879016578 Chr.19: 16355868-16355869 on Build GRCh38
rs1026516331 Chr.19: 16355671-16355671 on Build GRCh38
rs943566413 Chr.19: 16355653-16355653 on Build GRCh38
rs756728581 Chr.19: 16355676-16355676 on Build GRCh38
rs558982688 Chr.19: 16356032-16356032 on Build GRCh38
rs988422900 Chr.19: 16355814-16355814 on Build GRCh38
rs551438167 Chr.19: 16355673-16355673 on Build GRCh38
rs902539657 Chr.19: 16355679-16355679 on Build GRCh38
rs1039199834 Chr.19: 16355658-16355658 on Build GRCh38
rs1051968873 Chr.19: 16355973-16355973 on Build GRCh38
rs1039357916 Chr.19: 16355949-16355949 on Build GRCh38
rs538774106 Chr.19: 16356004-16356004 on Build GRCh38
rs912132047 Chr.19: 16355652-16355652 on Build GRCh38
rs946346939 Chr.19: 16355845-16355845 on Build GRCh38
rs1036998306 Chr.19: 16356034-16356034 on Build GRCh38
rs369548061 Chr.19: 16355621-16355622 on Build GRCh38
rs750850162 Chr.19: 16355683-16355683 on Build GRCh38
rs912186212 Chr.19: 16355958-16355958 on Build GRCh38
rs964952390 Chr.19: 16355960-16355960 on Build GRCh38
rs890595462 Chr.19: 16356015-16356015 on Build GRCh38
rs781173129 Chr.19: 16355900-16355900 on Build GRCh38
rs758944442 Chr.19: 16356068-16356068 on Build GRCh38
rs895061026 Chr.19: 16356042-16356042 on Build GRCh38
rs547301069 Chr.19: 16355741-16355741 on Build GRCh38
rs527262944 Chr.19: 16355711-16355711 on Build GRCh38
rs780179829 Chr.19: 16355867-16355867 on Build GRCh38
rs762195222 Chr.19: 16355686-16355686 on Build GRCh38
rs879632075 Chr.19: 16355620-16355621 on Build GRCh38
rs890752062 Chr.19: 16355650-16355650 on Build GRCh38
rs772371052 Chr.19: 16355695-16355695 on Build GRCh38
rs749586023 Chr.19: 16355736-16355736 on Build GRCh38
rs939370956 Chr.19: 16356031-16356031 on Build GRCh38

View Full Product Details