Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16392059-16392458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15L1
Entrez Gene ID
58513
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
Gene Aliases
EPS15R
UniGene
Hs.654639
Gene Chromosome Location
Chr.19: 16355244-16472012 on Build GRCh38
Transcripts
NM_001258374.1 NM_001258375.1 NM_021235.2 NR_047665.1 NR_047666.1 XM_017027086.1 XM_017027087.1 XM_017027088.1 XM_017027089.1 XM_017027090.1 XM_017027091.1 XM_017027092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6655517 c.2008C>T p.R670C Chr.19: 16392399-16392399 on Build GRCh38
3891802 c.2066C>T p.S689L Chr.19: 16392341-16392341 on Build GRCh38
6084311 c.2068G>T p.D690Y Chr.19: 16392339-16392339 on Build GRCh38
4592301 c.2078C>T p.T693M Chr.19: 16392329-16392329 on Build GRCh38
4869517 c.2028C>T p.D676D Chr.19: 16392379-16392379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967870866 Chr.19: 16392210-16392210 on Build GRCh38
rs1014277243 Chr.19: 16392179-16392179 on Build GRCh38
rs758961656 Chr.19: 16392372-16392372 on Build GRCh38
rs371635442 Chr.19: 16392286-16392286 on Build GRCh38
rs753919062 Chr.19: 16392287-16392287 on Build GRCh38
rs371775479 Chr.19: 16392406-16392406 on Build GRCh38
rs535270269 Chr.19: 16392294-16392294 on Build GRCh38
rs910486082 Chr.19: 16392236-16392236 on Build GRCh38
rs778541349 Chr.19: 16392412-16392412 on Build GRCh38
rs774296709 Chr.19: 16392270-16392270 on Build GRCh38
rs913795292 Chr.19: 16392217-16392217 on Build GRCh38
rs957975492 Chr.19: 16392187-16392187 on Build GRCh38
rs764029598 Chr.19: 16392231-16392231 on Build GRCh38
rs779816749 Chr.19: 16392328-16392328 on Build GRCh38
rs772050964 Chr.19: 16392431-16392431 on Build GRCh38
rs143261397 Chr.19: 16392381-16392381 on Build GRCh38
rs993642899 Chr.19: 16392104-16392104 on Build GRCh38
rs564032891 Chr.19: 16392206-16392206 on Build GRCh38
rs111917473 Chr.19: 16392129-16392129 on Build GRCh38
rs140895469 Chr.19: 16392329-16392329 on Build GRCh38
rs762488898 Chr.19: 16392266-16392266 on Build GRCh38
rs761522186 Chr.19: 16392214-16392214 on Build GRCh38
rs771888913 Chr.19: 16392340-16392340 on Build GRCh38
rs367867461 Chr.19: 16392399-16392399 on Build GRCh38
rs762864737 Chr.19: 16392211-16392211 on Build GRCh38
rs897580197 Chr.19: 16392123-16392123 on Build GRCh38
rs374304767 Chr.19: 16392341-16392341 on Build GRCh38
rs768331449 Chr.19: 16392332-16392332 on Build GRCh38
rs1005839587 Chr.19: 16392165-16392165 on Build GRCh38
rs368631545 Chr.19: 16392281-16392281 on Build GRCh38
rs1037931122 Chr.19: 16392093-16392093 on Build GRCh38
rs779411366 Chr.19: 16392290-16392290 on Build GRCh38
rs769578542 Chr.19: 16392312-16392312 on Build GRCh38
rs767991057 Chr.19: 16392366-16392367 on Build GRCh38
rs759729762 Chr.19: 16392361-16392361 on Build GRCh38
rs765338193 Chr.19: 16392363-16392363 on Build GRCh38
rs1046190766 Chr.19: 16392393-16392393 on Build GRCh38
rs888641695 Chr.19: 16392293-16392293 on Build GRCh38
rs912519030 Chr.19: 16392093-16392094 on Build GRCh38
rs3786587 Chr.19: 16392168-16392168 on Build GRCh38
rs540405223 Chr.19: 16392269-16392269 on Build GRCh38
rs147174754 Chr.19: 16392291-16392291 on Build GRCh38
rs751511828 Chr.19: 16392235-16392235 on Build GRCh38
rs929036938 Chr.19: 16392146-16392146 on Build GRCh38
rs750179589 Chr.19: 16392280-16392280 on Build GRCh38
rs757647851 Chr.19: 16392392-16392392 on Build GRCh38
rs761635857 Chr.19: 16392272-16392272 on Build GRCh38
rs773705155 Chr.19: 16392333-16392333 on Build GRCh38
rs944595234 Chr.19: 16392242-16392242 on Build GRCh38
rs888613697 Chr.19: 16392158-16392158 on Build GRCh38
rs200432030 Chr.19: 16392267-16392267 on Build GRCh38
rs766615514 Chr.19: 16392356-16392356 on Build GRCh38
rs143484833 Chr.19: 16392288-16392288 on Build GRCh38
rs781674469 Chr.19: 16392398-16392398 on Build GRCh38
rs112068280 Chr.19: 16392278-16392278 on Build GRCh38
rs1033294698 Chr.19: 16392230-16392230 on Build GRCh38
rs555317936 Chr.19: 16392155-16392155 on Build GRCh38
rs544063593 Chr.19: 16392169-16392169 on Build GRCh38
rs369274991 Chr.19: 16392305-16392305 on Build GRCh38
rs751989776 Chr.19: 16392382-16392382 on Build GRCh38
rs758797263 Chr.19: 16392295-16392295 on Build GRCh38
rs752708341 Chr.19: 16392371-16392371 on Build GRCh38
rs147722989 Chr.19: 16392432-16392432 on Build GRCh38
rs747553465 Chr.19: 16392418-16392418 on Build GRCh38
rs541476937 Chr.19: 16392145-16392145 on Build GRCh38
rs747281785 Chr.19: 16392311-16392311 on Build GRCh38
rs765050854 Chr.19: 16392307-16392307 on Build GRCh38
rs976159188 Chr.19: 16392249-16392249 on Build GRCh38
rs754007911 Chr.19: 16392359-16392359 on Build GRCh38

View Full Product Details