Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16522962-16523449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHERP
Entrez Gene ID
10523
Gene Name
calcium homeostasis endoplasmic reticulum protein
Gene Aliases
DAN16, SCAF6, SRA1
UniGene
Hs.740364
Gene Chromosome Location
Chr.19: 16517889-16542452 on Build GRCh38
Transcripts
NM_006387.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6150214 c.1974C>T p.L658L Chr.19: 16523058-16523058 on Build GRCh38
4610068 c.1912G>A p.D638N Chr.19: 16523120-16523120 on Build GRCh38
3284707 c.1836C>T p.F612F Chr.19: 16523196-16523196 on Build GRCh38
1201034 c.1750C>T p.P584S Chr.19: 16523282-16523282 on Build GRCh38
3284706 c.1849delC p.H617fs*38 Chr.19: 16523183-16523183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371011161 Chr.19: 16523257-16523257 on Build GRCh38
rs780562940 Chr.19: 16523195-16523195 on Build GRCh38
rs371448176 Chr.19: 16523185-16523185 on Build GRCh38
rs1004769561 Chr.19: 16523362-16523362 on Build GRCh38
rs769014572 Chr.19: 16523125-16523125 on Build GRCh38
rs770349359 Chr.19: 16523111-16523111 on Build GRCh38
rs763921296 Chr.19: 16523299-16523299 on Build GRCh38
rs1023504427 Chr.19: 16523403-16523403 on Build GRCh38
rs577471412 Chr.19: 16523389-16523389 on Build GRCh38
rs963367176 Chr.19: 16523384-16523384 on Build GRCh38
rs947542598 Chr.19: 16523011-16523011 on Build GRCh38
rs748457104 Chr.19: 16523229-16523229 on Build GRCh38
rs199591449 Chr.19: 16523140-16523140 on Build GRCh38
rs779384365 Chr.19: 16523227-16523227 on Build GRCh38
rs1015248754 Chr.19: 16523363-16523363 on Build GRCh38
rs775106668 Chr.19: 16523258-16523258 on Build GRCh38
rs775905252 Chr.19: 16523123-16523123 on Build GRCh38
rs746418645 Chr.19: 16523010-16523010 on Build GRCh38
rs368013917 Chr.19: 16523325-16523325 on Build GRCh38
rs916314293 Chr.19: 16523117-16523117 on Build GRCh38
rs773829616 Chr.19: 16523243-16523243 on Build GRCh38
rs746139397 Chr.19: 16523300-16523301 on Build GRCh38
rs981878470 Chr.19: 16523422-16523422 on Build GRCh38
rs1030316223 Chr.19: 16523013-16523013 on Build GRCh38
rs369620500 Chr.19: 16523003-16523003 on Build GRCh38
rs776347542 Chr.19: 16523149-16523149 on Build GRCh38
rs200966843 Chr.19: 16523184-16523184 on Build GRCh38
rs759050763 Chr.19: 16523153-16523153 on Build GRCh38
rs770612415 Chr.19: 16523219-16523219 on Build GRCh38
rs374902355 Chr.19: 16523157-16523157 on Build GRCh38
rs765280437 Chr.19: 16523156-16523156 on Build GRCh38
rs376237090 Chr.19: 16523192-16523192 on Build GRCh38
rs377203890 Chr.19: 16523019-16523019 on Build GRCh38
rs900580936 Chr.19: 16523248-16523248 on Build GRCh38
rs900633087 Chr.19: 16523002-16523002 on Build GRCh38
rs772769177 Chr.19: 16523132-16523132 on Build GRCh38
rs546473562 Chr.19: 16523407-16523407 on Build GRCh38
rs758156052 Chr.19: 16523082-16523082 on Build GRCh38
rs1011624789 Chr.19: 16522989-16522989 on Build GRCh38
rs890458892 Chr.19: 16523343-16523343 on Build GRCh38
rs963980496 Chr.19: 16523426-16523426 on Build GRCh38
rs996228534 Chr.19: 16523283-16523283 on Build GRCh38
rs913516264 Chr.19: 16523206-16523206 on Build GRCh38
rs770259450 Chr.19: 16523012-16523012 on Build GRCh38
rs61749017 Chr.19: 16523178-16523178 on Build GRCh38
rs914667769 Chr.19: 16523000-16523000 on Build GRCh38
rs200431428 Chr.19: 16523037-16523037 on Build GRCh38
rs181968256 Chr.19: 16523058-16523058 on Build GRCh38
rs557288720 Chr.19: 16523226-16523226 on Build GRCh38
rs201681974 Chr.19: 16523038-16523038 on Build GRCh38
rs757394350 Chr.19: 16523099-16523099 on Build GRCh38
rs368216975 Chr.19: 16523245-16523245 on Build GRCh38
rs757443115 Chr.19: 16523182-16523183 on Build GRCh38
rs947538736 Chr.19: 16523222-16523222 on Build GRCh38
rs750510620 Chr.19: 16523188-16523188 on Build GRCh38
rs373087540 Chr.19: 16523088-16523088 on Build GRCh38
rs753367315 Chr.19: 16523048-16523048 on Build GRCh38
rs372757352 Chr.19: 16523009-16523009 on Build GRCh38
rs777195535 Chr.19: 16523061-16523061 on Build GRCh38
rs781388903 Chr.19: 16523295-16523295 on Build GRCh38
rs549724277 Chr.19: 16522999-16522999 on Build GRCh38
rs938894147 Chr.19: 16523183-16523183 on Build GRCh38
rs752665285 Chr.19: 16523240-16523240 on Build GRCh38
rs765871236 Chr.19: 16523039-16523039 on Build GRCh38
rs751489414 Chr.19: 16523009-16523010 on Build GRCh38
rs762992720 Chr.19: 16523262-16523262 on Build GRCh38
rs762517535 Chr.19: 16523168-16523168 on Build GRCh38
rs375080317 Chr.19: 16523324-16523324 on Build GRCh38
rs769385871 Chr.19: 16523251-16523251 on Build GRCh38
rs749663323 Chr.19: 16523124-16523124 on Build GRCh38
rs777883442 Chr.19: 16523087-16523087 on Build GRCh38
rs770620355 Chr.19: 16523234-16523234 on Build GRCh38
rs927577972 Chr.19: 16523429-16523429 on Build GRCh38
rs777039763 Chr.19: 16523006-16523006 on Build GRCh38
rs767078913 Chr.19: 16523033-16523033 on Build GRCh38
rs995061522 Chr.19: 16523386-16523386 on Build GRCh38
rs973296096 Chr.19: 16523100-16523100 on Build GRCh38
rs534828961 Chr.19: 16523101-16523101 on Build GRCh38
rs766830505 Chr.19: 16523018-16523018 on Build GRCh38
rs755024354 Chr.19: 16523055-16523055 on Build GRCh38
rs554627456 Chr.19: 16523109-16523109 on Build GRCh38
rs34025480 Chr.19: 16523181-16523181 on Build GRCh38
rs983791488 Chr.19: 16523057-16523057 on Build GRCh38
rs1043481 Chr.19: 16523139-16523139 on Build GRCh38

View Full Product Details