Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16659310-16659815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM7
Entrez Gene ID
79086
Gene Name
small integral membrane protein 7
Gene Aliases
C19orf42
UniGene
Hs.356467
Gene Chromosome Location
Chr.19: 16630741-16660177 on Build GRCh38
Transcripts
NM_001300925.1 NM_024104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3284803 c.109G>T p.E37* Chr.19: 16659407-16659407 on Build GRCh38
992501 c.75G>T p.K25N Chr.19: 16659441-16659441 on Build GRCh38
3668198 c.84G>T p.T28T Chr.19: 16659432-16659432 on Build GRCh38
5744497 c.76_78delAAG p.K26delK Chr.19: 16659438-16659440 on Build GRCh38
5854934 c.70A>G p.K24E Chr.19: 16659446-16659446 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs928171744 Chr.19: 16659519-16659519 on Build GRCh38
rs899713374 Chr.19: 16659602-16659602 on Build GRCh38
rs1029506203 Chr.19: 16659414-16659414 on Build GRCh38
rs991579346 Chr.19: 16659705-16659705 on Build GRCh38
rs758815872 Chr.19: 16659427-16659427 on Build GRCh38
rs532255085 Chr.19: 16659676-16659676 on Build GRCh38
rs776914235 Chr.19: 16659468-16659468 on Build GRCh38
rs888007771 Chr.19: 16659505-16659505 on Build GRCh38
rs896836575 Chr.19: 16659788-16659788 on Build GRCh38
rs1020735614 Chr.19: 16659666-16659666 on Build GRCh38
rs929644046 Chr.19: 16659789-16659789 on Build GRCh38
rs770788534 Chr.19: 16659464-16659464 on Build GRCh38
rs886965987 Chr.19: 16659640-16659640 on Build GRCh38
rs990172639 Chr.19: 16659659-16659659 on Build GRCh38
rs558413737 Chr.19: 16659472-16659472 on Build GRCh38
rs779642194 Chr.19: 16659350-16659350 on Build GRCh38
rs921294411 Chr.19: 16659780-16659780 on Build GRCh38
rs190429502 Chr.19: 16659678-16659678 on Build GRCh38
rs970020478 Chr.19: 16659699-16659699 on Build GRCh38
rs1038410523 Chr.19: 16659516-16659516 on Build GRCh38
rs755740503 Chr.19: 16659349-16659349 on Build GRCh38
rs942311378 Chr.19: 16659737-16659737 on Build GRCh38
rs545939239 Chr.19: 16659488-16659489 on Build GRCh38
rs962401116 Chr.19: 16659445-16659445 on Build GRCh38
rs915287422 Chr.19: 16659749-16659749 on Build GRCh38
rs774170037 Chr.19: 16659455-16659457 on Build GRCh38
rs1016593197 Chr.19: 16659437-16659437 on Build GRCh38
rs776940319 Chr.19: 16659383-16659383 on Build GRCh38
rs774351914 Chr.19: 16659358-16659358 on Build GRCh38
rs367784846 Chr.19: 16659506-16659506 on Build GRCh38
rs559244251 Chr.19: 16659681-16659681 on Build GRCh38
rs376538398 Chr.19: 16659459-16659459 on Build GRCh38
rs1018221711 Chr.19: 16659614-16659614 on Build GRCh38
rs199810423 Chr.19: 16659452-16659452 on Build GRCh38
rs192887889 Chr.19: 16659758-16659758 on Build GRCh38
rs946810780 Chr.19: 16659768-16659768 on Build GRCh38
rs759671882 Chr.19: 16659470-16659470 on Build GRCh38
rs373832787 Chr.19: 16659432-16659432 on Build GRCh38
rs982408613 Chr.19: 16659709-16659709 on Build GRCh38
rs1040082098 Chr.19: 16659770-16659770 on Build GRCh38
rs896811033 Chr.19: 16659549-16659549 on Build GRCh38
rs1029394994 Chr.19: 16659572-16659572 on Build GRCh38
rs748169930 Chr.19: 16659455-16659455 on Build GRCh38
rs7250904 Chr.19: 16659590-16659590 on Build GRCh38
rs751889243 Chr.19: 16659435-16659435 on Build GRCh38
rs150409915 Chr.19: 16659458-16659458 on Build GRCh38
rs928122077 Chr.19: 16659723-16659723 on Build GRCh38
rs779695843 Chr.19: 16659439-16659439 on Build GRCh38
rs998127626 Chr.19: 16659612-16659612 on Build GRCh38
rs764607449 Chr.19: 16659481-16659481 on Build GRCh38
rs893799342 Chr.19: 16659566-16659566 on Build GRCh38
rs956902991 Chr.19: 16659742-16659742 on Build GRCh38
rs574063712 Chr.19: 16659611-16659611 on Build GRCh38
rs916777274 Chr.19: 16659480-16659480 on Build GRCh38
rs370897054 Chr.19: 16659494-16659494 on Build GRCh38
rs909477071 Chr.19: 16659713-16659713 on Build GRCh38
rs370871373 Chr.19: 16659395-16659395 on Build GRCh38
rs752184656 Chr.19: 16659499-16659499 on Build GRCh38
rs767865109 Chr.19: 16659488-16659488 on Build GRCh38
rs763491200 Chr.19: 16659478-16659478 on Build GRCh38
rs761021319 Chr.19: 16659377-16659377 on Build GRCh38
rs533867372 Chr.19: 16659558-16659558 on Build GRCh38
rs73006566 Chr.19: 16659781-16659781 on Build GRCh38
rs1016730745 Chr.19: 16659675-16659675 on Build GRCh38
rs770194071 Chr.19: 16659554-16659554 on Build GRCh38
rs113967791 Chr.19: 16659448-16659448 on Build GRCh38
rs752007341 Chr.19: 16659484-16659484 on Build GRCh38
rs572109290 Chr.19: 16659492-16659492 on Build GRCh38
rs988183321 Chr.19: 16659745-16659745 on Build GRCh38
rs1039711892 Chr.19: 16659598-16659598 on Build GRCh38
rs937498969 Chr.19: 16659491-16659491 on Build GRCh38
rs767995173 Chr.19: 16659354-16659354 on Build GRCh38
rs1018643256 Chr.19: 16659359-16659359 on Build GRCh38
rs528603729 Chr.19: 16659738-16659738 on Build GRCh38
rs773123581 Chr.19: 16659368-16659368 on Build GRCh38
rs937531332 Chr.19: 16659703-16659703 on Build GRCh38
rs796234341 Chr.19: 16659416-16659416 on Build GRCh38
rs762170964 Chr.19: 16659487-16659487 on Build GRCh38
rs138204281 Chr.19: 16659790-16659790 on Build GRCh38
rs951084549 Chr.19: 16659433-16659433 on Build GRCh38
rs746818614 Chr.19: 16659463-16659463 on Build GRCh38
rs184361526 Chr.19: 16659343-16659343 on Build GRCh38
rs141398952 Chr.19: 16659385-16659385 on Build GRCh38
rs754586478 Chr.19: 16659446-16659446 on Build GRCh38
rs560563862 Chr.19: 16659593-16659593 on Build GRCh38
rs765160738 Chr.19: 16659405-16659405 on Build GRCh38
rs371641598 Chr.19: 16659644-16659644 on Build GRCh38
rs769972531 Chr.19: 16659471-16659471 on Build GRCh38
rs745577349 Chr.19: 16659348-16659348 on Build GRCh38
rs753368172 Chr.19: 16659440-16659440 on Build GRCh38
rs1003954166 Chr.19: 16659653-16659653 on Build GRCh38
rs1029889328 Chr.19: 16659624-16659624 on Build GRCh38
rs772053477 Chr.19: 16659366-16659366 on Build GRCh38
rs748852749 Chr.19: 16659351-16659351 on Build GRCh38
rs757568020 Chr.19: 16659436-16659436 on Build GRCh38
rs563235526 Chr.19: 16659657-16659657 on Build GRCh38
rs935399194 Chr.19: 16659520-16659520 on Build GRCh38
rs768225267 Chr.19: 16659348-16659349 on Build GRCh38
rs543350100 Chr.19: 16659643-16659643 on Build GRCh38

View Full Product Details