Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16888849-16889350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL3
Entrez Gene ID
9002
Gene Name
F2R like thrombin/trypsin receptor 3
Gene Aliases
PAR4
UniGene
Hs.137574
Gene Chromosome Location
Chr.19: 16889015-16892019 on Build GRCh38
Transcripts
NM_003950.2 XM_005260139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3378641 c.11G>A p.R4Q Chr.19: 16889200-16889200 on Build GRCh38
6437353 c.18C>A p.L6L Chr.19: 16889207-16889207 on Build GRCh38
6293301 c.88G>A p.G30R Chr.19: 16889277-16889277 on Build GRCh38
4650451 c.10C>T p.R4* Chr.19: 16889199-16889199 on Build GRCh38
6839850 c.104G>C p.G35A Chr.19: 16889293-16889293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773904 Chr.19: 16889320-16889320 on Build GRCh38
rs542089132 Chr.19: 16889144-16889144 on Build GRCh38
rs1016256174 Chr.19: 16889112-16889112 on Build GRCh38
rs990261402 Chr.19: 16889234-16889234 on Build GRCh38
rs755121629 Chr.19: 16889258-16889258 on Build GRCh38
rs777706484 Chr.19: 16889018-16889018 on Build GRCh38
rs201082991 Chr.19: 16889256-16889256 on Build GRCh38
rs535684787 Chr.19: 16888918-16888918 on Build GRCh38
rs566699092 Chr.19: 16888915-16888915 on Build GRCh38
rs757806648 Chr.19: 16888909-16888909 on Build GRCh38
rs914299100 Chr.19: 16889064-16889064 on Build GRCh38
rs1010384135 Chr.19: 16888919-16888920 on Build GRCh38
rs748653018 Chr.19: 16889150-16889150 on Build GRCh38
rs781513107 Chr.19: 16888911-16888911 on Build GRCh38
rs1017473946 Chr.19: 16888920-16888920 on Build GRCh38
rs865935891 Chr.19: 16889092-16889092 on Build GRCh38
rs760879099 Chr.19: 16889200-16889200 on Build GRCh38
rs748490642 Chr.19: 16889285-16889285 on Build GRCh38
rs959110116 Chr.19: 16888909-16888910 on Build GRCh38
rs1012361058 Chr.19: 16889129-16889129 on Build GRCh38
rs1045407147 Chr.19: 16888913-16888913 on Build GRCh38
rs1051578576 Chr.19: 16889282-16889282 on Build GRCh38
rs906896479 Chr.19: 16888914-16888914 on Build GRCh38
rs1004829616 Chr.19: 16889108-16889108 on Build GRCh38
rs911272232 Chr.19: 16889198-16889198 on Build GRCh38
rs149636145 Chr.19: 16889261-16889261 on Build GRCh38
rs1024193677 Chr.19: 16889139-16889139 on Build GRCh38
rs761441370 Chr.19: 16889021-16889021 on Build GRCh38
rs796268871 Chr.19: 16888910-16888910 on Build GRCh38
rs955347350 Chr.19: 16889154-16889154 on Build GRCh38
rs774010525 Chr.19: 16889294-16889294 on Build GRCh38
rs879175815 Chr.19: 16889103-16889103 on Build GRCh38
rs368402650 Chr.19: 16889161-16889161 on Build GRCh38
rs924197733 Chr.19: 16889148-16889148 on Build GRCh38
rs768596314 Chr.19: 16889142-16889142 on Build GRCh38
rs970899512 Chr.19: 16888887-16888887 on Build GRCh38
rs774953032 Chr.19: 16889223-16889223 on Build GRCh38
rs773583327 Chr.19: 16889168-16889168 on Build GRCh38
rs1020686015 Chr.19: 16888874-16888874 on Build GRCh38
rs2227343 Chr.19: 16889140-16889140 on Build GRCh38
rs186635098 Chr.19: 16888991-16888991 on Build GRCh38
rs558141292 Chr.19: 16889022-16889022 on Build GRCh38
rs1054476492 Chr.19: 16889074-16889074 on Build GRCh38
rs371477486 Chr.19: 16889202-16889202 on Build GRCh38
rs549256785 Chr.19: 16888931-16888931 on Build GRCh38
rs772570199 Chr.19: 16889267-16889267 on Build GRCh38
rs200284465 Chr.19: 16889286-16889286 on Build GRCh38
rs398059601 Chr.19: 16888918-16888919 on Build GRCh38
rs916808219 Chr.19: 16888901-16888901 on Build GRCh38
rs983459071 Chr.19: 16888983-16888983 on Build GRCh38
rs914269296 Chr.19: 16889242-16889242 on Build GRCh38
rs569256201 Chr.19: 16888978-16888978 on Build GRCh38
rs982373674 Chr.19: 16889038-16889038 on Build GRCh38
rs114514490 Chr.19: 16889055-16889055 on Build GRCh38
rs755817250 Chr.19: 16889255-16889255 on Build GRCh38
rs374965245 Chr.19: 16889227-16889227 on Build GRCh38
rs1039263407 Chr.19: 16889076-16889076 on Build GRCh38
rs773905 Chr.19: 16889086-16889086 on Build GRCh38
rs2227342 Chr.19: 16889054-16889054 on Build GRCh38
rs997468225 Chr.19: 16888958-16888958 on Build GRCh38
rs767502983 Chr.19: 16889253-16889253 on Build GRCh38
rs991393762 Chr.19: 16889013-16889013 on Build GRCh38
rs1030278325 Chr.19: 16888966-16888966 on Build GRCh38
rs1051632424 Chr.19: 16889094-16889094 on Build GRCh38
rs1005188395 Chr.19: 16889010-16889010 on Build GRCh38
rs113341502 Chr.19: 16889299-16889299 on Build GRCh38
rs750306446 Chr.19: 16889317-16889317 on Build GRCh38
rs546849168 Chr.19: 16888898-16888898 on Build GRCh38
rs747418532 Chr.19: 16889271-16889271 on Build GRCh38
rs761368804 Chr.19: 16889308-16889308 on Build GRCh38
rs949701526 Chr.19: 16888912-16888912 on Build GRCh38
rs575614575 Chr.19: 16889254-16889254 on Build GRCh38
rs769564057 Chr.19: 16889037-16889037 on Build GRCh38
rs748170502 Chr.19: 16889262-16889262 on Build GRCh38
rs963773014 Chr.19: 16889120-16889120 on Build GRCh38
rs543825591 Chr.19: 16889257-16889257 on Build GRCh38
rs543230398 Chr.19: 16888892-16888892 on Build GRCh38
rs763665257 Chr.19: 16889237-16889237 on Build GRCh38
rs767586884 Chr.19: 16889313-16889313 on Build GRCh38
rs751474384 Chr.19: 16889044-16889044 on Build GRCh38
rs749409912 Chr.19: 16889141-16889141 on Build GRCh38
rs984392645 Chr.19: 16889133-16889143 on Build GRCh38
rs35021759 Chr.19: 16888910-16888911 on Build GRCh38
rs773667119 Chr.19: 16889268-16889268 on Build GRCh38
rs752603563 Chr.19: 16888990-16888990 on Build GRCh38
rs771173733 Chr.19: 16889276-16889276 on Build GRCh38
rs775248739 Chr.19: 16889277-16889277 on Build GRCh38
rs1046022085 Chr.19: 16889089-16889089 on Build GRCh38
rs777606828 Chr.19: 16888909-16888910 on Build GRCh38
rs983530883 Chr.19: 16888885-16888885 on Build GRCh38
rs756750766 Chr.19: 16889075-16889075 on Build GRCh38
rs1001720765 Chr.19: 16889093-16889093 on Build GRCh38
rs892935798 Chr.19: 16888917-16888917 on Build GRCh38
rs757245502 Chr.19: 16889244-16889244 on Build GRCh38
rs879732220 Chr.19: 16888920-16888920 on Build GRCh38
rs1052875961 Chr.19: 16888916-16888916 on Build GRCh38
rs2227341 Chr.19: 16888889-16888889 on Build GRCh38
rs900417373 Chr.19: 16888943-16888943 on Build GRCh38
rs909813898 Chr.19: 16888911-16888911 on Build GRCh38
rs751612733 Chr.19: 16889243-16889243 on Build GRCh38
rs376137740 Chr.19: 16889159-16889159 on Build GRCh38
rs372392370 Chr.19: 16888919-16888919 on Build GRCh38
rs1045912122 Chr.19: 16888921-16888921 on Build GRCh38
rs113942373 Chr.19: 16889300-16889300 on Build GRCh38

View Full Product Details