Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16899392-16899883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPAMD8
Entrez Gene ID
27151
Gene Name
C3 and PZP like, alpha-2-macroglobulin domain containing 8
Gene Aliases
K-CAP, VIP
UniGene
Hs.631644
Gene Chromosome Location
Chr.19: 16892951-17026818 on Build GRCh38
Transcripts
NM_015692.2 XM_011527917.1 XM_011527918.1 XM_011527919.1 XM_011527920.2 XM_011527921.2 XM_011527922.1 XM_011527923.1 XM_011527924.2 XM_011527925.1 XM_017026594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
474309 c.4925A>C p.D1642A Chr.19: 16899539-16899539 on Build GRCh38
3891833 c.4920C>T p.L1640L Chr.19: 16899544-16899544 on Build GRCh38
5350480 c.4917G>T p.L1639L Chr.19: 16899547-16899547 on Build GRCh38
3285172 c.4957G>A p.A1653T Chr.19: 16899507-16899507 on Build GRCh38
5854943 c.4937G>A p.G1646E Chr.19: 16899527-16899527 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202120343 Chr.19: 16899500-16899500 on Build GRCh38
rs776683737 Chr.19: 16899546-16899546 on Build GRCh38
rs372341962 Chr.19: 16899654-16899654 on Build GRCh38
rs570854386 Chr.19: 16899517-16899517 on Build GRCh38
rs893190528 Chr.19: 16899819-16899819 on Build GRCh38
rs371481988 Chr.19: 16899430-16899430 on Build GRCh38
rs1017046397 Chr.19: 16899687-16899687 on Build GRCh38
rs774879992 Chr.19: 16899589-16899589 on Build GRCh38
rs188892031 Chr.19: 16899817-16899817 on Build GRCh38
rs765815991 Chr.19: 16899453-16899453 on Build GRCh38
rs1055061608 Chr.19: 16899848-16899848 on Build GRCh38
rs57816869 Chr.19: 16899781-16899781 on Build GRCh38
rs753438460 Chr.19: 16899498-16899498 on Build GRCh38
rs998193391 Chr.19: 16899615-16899615 on Build GRCh38
rs375943813 Chr.19: 16899462-16899462 on Build GRCh38
rs2227375 Chr.19: 16899573-16899573 on Build GRCh38
rs555592649 Chr.19: 16899746-16899746 on Build GRCh38
rs941154563 Chr.19: 16899859-16899859 on Build GRCh38
rs948648776 Chr.19: 16899477-16899477 on Build GRCh38
rs908287259 Chr.19: 16899727-16899727 on Build GRCh38
rs1045709312 Chr.19: 16899484-16899484 on Build GRCh38
rs994154924 Chr.19: 16899703-16899703 on Build GRCh38
rs763559280 Chr.19: 16899576-16899576 on Build GRCh38
rs897265790 Chr.19: 16899695-16899695 on Build GRCh38
rs748090856 Chr.19: 16899501-16899501 on Build GRCh38
rs948636152 Chr.19: 16899764-16899764 on Build GRCh38
rs200054840 Chr.19: 16899497-16899497 on Build GRCh38
rs764423028 Chr.19: 16899426-16899428 on Build GRCh38
rs572977645 Chr.19: 16899681-16899681 on Build GRCh38
rs1055326095 Chr.19: 16899631-16899631 on Build GRCh38
rs915749616 Chr.19: 16899763-16899763 on Build GRCh38
rs900678597 Chr.19: 16899855-16899855 on Build GRCh38
rs985734183 Chr.19: 16899737-16899737 on Build GRCh38
rs531568395 Chr.19: 16899760-16899760 on Build GRCh38
rs778516705 Chr.19: 16899816-16899816 on Build GRCh38
rs939321357 Chr.19: 16899616-16899616 on Build GRCh38
rs928581084 Chr.19: 16899802-16899802 on Build GRCh38
rs374966464 Chr.19: 16899431-16899431 on Build GRCh38
rs562657793 Chr.19: 16899704-16899704 on Build GRCh38
rs763473427 Chr.19: 16899463-16899463 on Build GRCh38
rs191586386 Chr.19: 16899841-16899841 on Build GRCh38
rs769984287 Chr.19: 16899569-16899569 on Build GRCh38
rs764570905 Chr.19: 16899474-16899474 on Build GRCh38
rs894358476 Chr.19: 16899466-16899466 on Build GRCh38
rs369606393 Chr.19: 16899433-16899433 on Build GRCh38
rs893262716 Chr.19: 16899534-16899534 on Build GRCh38
rs764656970 Chr.19: 16899584-16899584 on Build GRCh38
rs894957475 Chr.19: 16899857-16899858 on Build GRCh38
rs376952579 Chr.19: 16899847-16899847 on Build GRCh38
rs927652326 Chr.19: 16899843-16899843 on Build GRCh38
rs772932948 Chr.19: 16899427-16899427 on Build GRCh38
rs780041407 Chr.19: 16899413-16899413 on Build GRCh38
rs575266565 Chr.19: 16899691-16899691 on Build GRCh38
rs372544588 Chr.19: 16899438-16899438 on Build GRCh38
rs962810540 Chr.19: 16899728-16899728 on Build GRCh38
rs758264719 Chr.19: 16899502-16899502 on Build GRCh38
rs1018825222 Chr.19: 16899743-16899743 on Build GRCh38
rs997721579 Chr.19: 16899860-16899860 on Build GRCh38
rs770122772 Chr.19: 16899839-16899839 on Build GRCh38
rs759508703 Chr.19: 16899439-16899439 on Build GRCh38
rs755028686 Chr.19: 16899488-16899488 on Build GRCh38
rs112183208 Chr.19: 16899555-16899555 on Build GRCh38
rs771231458 Chr.19: 16899525-16899525 on Build GRCh38
rs373993040 Chr.19: 16899520-16899520 on Build GRCh38
rs777444675 Chr.19: 16899759-16899759 on Build GRCh38
rs894986482 Chr.19: 16899656-16899656 on Build GRCh38
rs973894860 Chr.19: 16899747-16899747 on Build GRCh38
rs747640444 Chr.19: 16899607-16899607 on Build GRCh38
rs755822674 Chr.19: 16899490-16899490 on Build GRCh38
rs957547941 Chr.19: 16899767-16899767 on Build GRCh38

View Full Product Details