Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16903620-16904124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPAMD8
Entrez Gene ID
27151
Gene Name
C3 and PZP like, alpha-2-macroglobulin domain containing 8
Gene Aliases
K-CAP, VIP
UniGene
Hs.631644
Gene Chromosome Location
Chr.19: 16892951-17026818 on Build GRCh38
Transcripts
NM_015692.2 XM_011527917.1 XM_011527918.1 XM_011527919.1 XM_011527920.2 XM_011527921.2 XM_011527922.1 XM_011527923.1 XM_011527924.2 XM_011527925.1 XM_017026594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674291 c.4548G>A p.A1516A Chr.19: 16903702-16903702 on Build GRCh38
4075241 c.4474G>A p.A1492T Chr.19: 16903776-16903776 on Build GRCh38
6634444 c.4440C>A p.S1480S Chr.19: 16903810-16903810 on Build GRCh38
4075240 c.4532T>C p.V1511A Chr.19: 16903718-16903718 on Build GRCh38
6585380 c.4549-6C>G p.? Chr.19: 16903629-16903629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376798414 Chr.19: 16903748-16903748 on Build GRCh38
rs374544793 Chr.19: 16903804-16903804 on Build GRCh38
rs773298243 Chr.19: 16903819-16903819 on Build GRCh38
rs768430988 Chr.19: 16903706-16903706 on Build GRCh38
rs951578148 Chr.19: 16903698-16903698 on Build GRCh38
rs148551153 Chr.19: 16903695-16903695 on Build GRCh38
rs773386950 Chr.19: 16903735-16903735 on Build GRCh38
rs761941572 Chr.19: 16903648-16903648 on Build GRCh38
rs79769868 Chr.19: 16903908-16903908 on Build GRCh38
rs746100689 Chr.19: 16904077-16904077 on Build GRCh38
rs192468055 Chr.19: 16903726-16903726 on Build GRCh38
rs772231683 Chr.19: 16903727-16903727 on Build GRCh38
rs770377550 Chr.19: 16903845-16903845 on Build GRCh38
rs199540793 Chr.19: 16903823-16903823 on Build GRCh38
rs776990790 Chr.19: 16903743-16903743 on Build GRCh38
rs897736899 Chr.19: 16904026-16904026 on Build GRCh38
rs201980602 Chr.19: 16903843-16903843 on Build GRCh38
rs763896569 Chr.19: 16903676-16903676 on Build GRCh38
rs183185573 Chr.19: 16903768-16903768 on Build GRCh38
rs200441202 Chr.19: 16903866-16903866 on Build GRCh38
rs1006115105 Chr.19: 16903703-16903703 on Build GRCh38
rs914131170 Chr.19: 16903902-16903902 on Build GRCh38
rs778787711 Chr.19: 16903873-16903873 on Build GRCh38
rs754265993 Chr.19: 16903672-16903672 on Build GRCh38
rs1046562723 Chr.19: 16903916-16903916 on Build GRCh38
rs367943801 Chr.19: 16903860-16903860 on Build GRCh38
rs113882468 Chr.19: 16903961-16903961 on Build GRCh38
rs997298650 Chr.19: 16903753-16903753 on Build GRCh38
rs749240432 Chr.19: 16903793-16903793 on Build GRCh38
rs199701977 Chr.19: 16903646-16903646 on Build GRCh38
rs756126378 Chr.19: 16903696-16903696 on Build GRCh38
rs779452403 Chr.19: 16903681-16903681 on Build GRCh38
rs569812859 Chr.19: 16903878-16903878 on Build GRCh38
rs990424776 Chr.19: 16903794-16903794 on Build GRCh38
rs752344460 Chr.19: 16903669-16903675 on Build GRCh38
rs375733041 Chr.19: 16903744-16903744 on Build GRCh38
rs760636884 Chr.19: 16903737-16903737 on Build GRCh38
rs915064639 Chr.19: 16904079-16904079 on Build GRCh38
rs780102176 Chr.19: 16903785-16903785 on Build GRCh38
rs760149758 Chr.19: 16903838-16903838 on Build GRCh38
rs756994505 Chr.19: 16903765-16903765 on Build GRCh38
rs1053596705 Chr.19: 16904078-16904078 on Build GRCh38
rs758809027 Chr.19: 16903864-16903864 on Build GRCh38
rs535293825 Chr.19: 16903957-16903957 on Build GRCh38
rs766972326 Chr.19: 16903738-16903738 on Build GRCh38
rs10419251 Chr.19: 16903932-16903932 on Build GRCh38
rs1007368881 Chr.19: 16904080-16904080 on Build GRCh38
rs200916574 Chr.19: 16903877-16903877 on Build GRCh38
rs754844891 Chr.19: 16903797-16903797 on Build GRCh38
rs547396856 Chr.19: 16903778-16903778 on Build GRCh38
rs942533470 Chr.19: 16904093-16904093 on Build GRCh38
rs574729012 Chr.19: 16904003-16904003 on Build GRCh38
rs780787118 Chr.19: 16903692-16903692 on Build GRCh38
rs917525127 Chr.19: 16904013-16904013 on Build GRCh38
rs951551803 Chr.19: 16903955-16903955 on Build GRCh38
rs369782880 Chr.19: 16903660-16903660 on Build GRCh38
rs773864 Chr.19: 16904061-16904061 on Build GRCh38
rs201393101 Chr.19: 16903872-16903872 on Build GRCh38
rs142251610 Chr.19: 16903830-16903830 on Build GRCh38
rs767266770 Chr.19: 16903849-16903849 on Build GRCh38
rs753308051 Chr.19: 16903871-16903871 on Build GRCh38
rs111399374 Chr.19: 16903670-16903670 on Build GRCh38
rs756900590 Chr.19: 16903687-16903687 on Build GRCh38
rs151104169 Chr.19: 16903848-16903848 on Build GRCh38
rs879824502 Chr.19: 16903923-16903923 on Build GRCh38
rs537096025 Chr.19: 16904015-16904015 on Build GRCh38
rs774077751 Chr.19: 16903722-16903722 on Build GRCh38
rs772375480 Chr.19: 16903818-16903818 on Build GRCh38
rs760320969 Chr.19: 16903861-16903861 on Build GRCh38
rs369367045 Chr.19: 16903702-16903702 on Build GRCh38
rs937046106 Chr.19: 16903942-16903942 on Build GRCh38
rs991690423 Chr.19: 16904011-16904011 on Build GRCh38
rs755576183 Chr.19: 16903783-16903783 on Build GRCh38
rs370949447 Chr.19: 16903799-16903799 on Build GRCh38
rs923680395 Chr.19: 16904069-16904069 on Build GRCh38
rs1025717388 Chr.19: 16903935-16903935 on Build GRCh38
rs949054408 Chr.19: 16904057-16904057 on Build GRCh38
rs779824102 Chr.19: 16903697-16903697 on Build GRCh38
rs375463591 Chr.19: 16903658-16903658 on Build GRCh38
rs753069410 Chr.19: 16903749-16903749 on Build GRCh38
rs770928378 Chr.19: 16903827-16903827 on Build GRCh38
rs755390274 Chr.19: 16903673-16903673 on Build GRCh38
rs986190936 Chr.19: 16903815-16903815 on Build GRCh38
rs75751772 Chr.19: 16903956-16903956 on Build GRCh38
rs777567036 Chr.19: 16903901-16903901 on Build GRCh38

View Full Product Details