Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17285549-17286044 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKLE1
Entrez Gene ID
126549
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
ANKRD41, LEM3, LEMD6
UniGene
Hs.721610
Gene Chromosome Location
Chr.19: 17281645-17287646 on Build GRCh38
Transcripts
NM_001278443.1 NM_001278444.1 NM_001278445.1 NM_152363.5 NR_103530.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661694 c.1523G>A p.R508Q Chr.19: 17285577-17285577 on Build GRCh38
3285867 c.1398T>C p.H466H Chr.19: 17285572-17285572 on Build GRCh38
6600148 c.1535C>T p.A512V Chr.19: 17285799-17285799 on Build GRCh38
3285866 c.1518T>C p.H506H Chr.19: 17285572-17285572 on Build GRCh38
1630758 c.1465A>G p.S489G Chr.19: 17285729-17285729 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778138714 Chr.19: 17285837-17285837 on Build GRCh38
rs954262797 Chr.19: 17285587-17285587 on Build GRCh38
rs767276791 Chr.19: 17285799-17285799 on Build GRCh38
rs904149753 Chr.19: 17285805-17285805 on Build GRCh38
rs566377495 Chr.19: 17285854-17285854 on Build GRCh38
rs772363082 Chr.19: 17285668-17285668 on Build GRCh38
rs754086777 Chr.19: 17285962-17285962 on Build GRCh38
rs771091949 Chr.19: 17285727-17285727 on Build GRCh38
rs909743287 Chr.19: 17285885-17285885 on Build GRCh38
rs766614836 Chr.19: 17285609-17285609 on Build GRCh38
rs138115996 Chr.19: 17285714-17285714 on Build GRCh38
rs769231913 Chr.19: 17285570-17285570 on Build GRCh38
rs781286541 Chr.19: 17285832-17285832 on Build GRCh38
rs137932498 Chr.19: 17285788-17285788 on Build GRCh38
rs200377044 Chr.19: 17285835-17285835 on Build GRCh38
rs757739817 Chr.19: 17285850-17285850 on Build GRCh38
rs939537179 Chr.19: 17285593-17285593 on Build GRCh38
rs747524943 Chr.19: 17285695-17285695 on Build GRCh38
rs779607023 Chr.19: 17285646-17285646 on Build GRCh38
rs559221710 Chr.19: 17285953-17285953 on Build GRCh38
rs760557838 Chr.19: 17285801-17285801 on Build GRCh38
rs745821293 Chr.19: 17285706-17285706 on Build GRCh38
rs148284514 Chr.19: 17285848-17285848 on Build GRCh38
rs891018 Chr.19: 17285952-17285952 on Build GRCh38
rs939918730 Chr.19: 17285949-17285949 on Build GRCh38
rs189070227 Chr.19: 17285604-17285604 on Build GRCh38
rs748574759 Chr.19: 17285667-17285667 on Build GRCh38
rs745915734 Chr.19: 17285792-17285792 on Build GRCh38
rs867351419 Chr.19: 17285571-17285571 on Build GRCh38
rs772897917 Chr.19: 17285800-17285800 on Build GRCh38
rs148174459 Chr.19: 17285741-17285741 on Build GRCh38
rs574549736 Chr.19: 17285655-17285655 on Build GRCh38
rs193021583 Chr.19: 17285628-17285628 on Build GRCh38
rs766677017 Chr.19: 17285918-17285918 on Build GRCh38
rs376579821 Chr.19: 17285569-17285569 on Build GRCh38
rs997824132 Chr.19: 17285974-17285974 on Build GRCh38
rs570245391 Chr.19: 17285915-17285915 on Build GRCh38
rs143862711 Chr.19: 17285815-17285815 on Build GRCh38
rs764710644 Chr.19: 17285824-17285824 on Build GRCh38
rs767460796 Chr.19: 17285630-17285630 on Build GRCh38
rs961905624 Chr.19: 17285934-17285934 on Build GRCh38
rs1021233625 Chr.19: 17285645-17285645 on Build GRCh38
rs182162701 Chr.19: 17285573-17285573 on Build GRCh38
rs201853418 Chr.19: 17285733-17285733 on Build GRCh38
rs779198787 Chr.19: 17285661-17285661 on Build GRCh38
rs150249470 Chr.19: 17285754-17285754 on Build GRCh38
rs372428932 Chr.19: 17285617-17285617 on Build GRCh38
rs780003665 Chr.19: 17285869-17285869 on Build GRCh38
rs138940487 Chr.19: 17285761-17285761 on Build GRCh38
rs762171122 Chr.19: 17285583-17285583 on Build GRCh38
rs754640255 Chr.19: 17285658-17285658 on Build GRCh38
rs370950297 Chr.19: 17285773-17285773 on Build GRCh38
rs766333087 Chr.19: 17285810-17285810 on Build GRCh38
rs374555149 Chr.19: 17285677-17285677 on Build GRCh38
rs556785966 Chr.19: 17285584-17285584 on Build GRCh38
rs751599 Chr.19: 17285740-17285740 on Build GRCh38
rs754095558 Chr.19: 17285612-17285612 on Build GRCh38
rs748998458 Chr.19: 17285577-17285577 on Build GRCh38
rs1014749659 Chr.19: 17285927-17285927 on Build GRCh38
rs376369211 Chr.19: 17285858-17285858 on Build GRCh38
rs202000838 Chr.19: 17285581-17285581 on Build GRCh38
rs980083526 Chr.19: 17285821-17285821 on Build GRCh38
rs141089836 Chr.19: 17285748-17285748 on Build GRCh38
rs755640662 Chr.19: 17285634-17285634 on Build GRCh38
rs777599104 Chr.19: 17285574-17285576 on Build GRCh38
rs765591299 Chr.19: 17285738-17285738 on Build GRCh38
rs746440035 Chr.19: 17285865-17285865 on Build GRCh38
rs547043530 Chr.19: 17285774-17285774 on Build GRCh38
rs143586031 Chr.19: 17285767-17285767 on Build GRCh38
rs146201418 Chr.19: 17285775-17285775 on Build GRCh38
rs765819000 Chr.19: 17285758-17285758 on Build GRCh38
rs747230110 Chr.19: 17285586-17285588 on Build GRCh38
rs149470751 Chr.19: 17285814-17285814 on Build GRCh38
rs148939227 Chr.19: 17285737-17285737 on Build GRCh38
rs374646433 Chr.19: 17285793-17285793 on Build GRCh38
rs375747890 Chr.19: 17285627-17285627 on Build GRCh38
rs145927575 Chr.19: 17285768-17285768 on Build GRCh38
rs930723554 Chr.19: 17285849-17285849 on Build GRCh38
rs146466494 Chr.19: 17285576-17285576 on Build GRCh38
rs1049880210 Chr.19: 17286005-17286005 on Build GRCh38
rs370339417 Chr.19: 17285600-17285600 on Build GRCh38
rs775617803 Chr.19: 17285739-17285739 on Build GRCh38
rs763912716 Chr.19: 17285625-17285625 on Build GRCh38
rs1027740700 Chr.19: 17285957-17285957 on Build GRCh38
rs369579504 Chr.19: 17285709-17285709 on Build GRCh38

View Full Product Details