Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17811447-17811941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT3
Entrez Gene ID
10331
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3
Gene Aliases
B3GAL-T8, B3GN-T3, B3GNT-3, HP10328, TMEM3, beta3Gn-T3
UniGene
Hs.657825
Gene Chromosome Location
Chr.19: 17794828-17813576 on Build GRCh38
Transcripts
NM_014256.3 XM_011527626.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3692512 c.787C>T p.R263W Chr.19: 17811790-17811790 on Build GRCh38
6609310 c.757_758delTA p.Y253fs*9 Chr.19: 17811760-17811761 on Build GRCh38
6557632 c.819C>T p.F273F Chr.19: 17811822-17811822 on Build GRCh38
279099 c.615C>T p.F205F Chr.19: 17811618-17811618 on Build GRCh38
224791 c.853C>T p.R285C Chr.19: 17811856-17811856 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766402937 Chr.19: 17811558-17811558 on Build GRCh38
rs746408665 Chr.19: 17811856-17811856 on Build GRCh38
rs969276177 Chr.19: 17811498-17811498 on Build GRCh38
rs982976835 Chr.19: 17811624-17811624 on Build GRCh38
rs567013684 Chr.19: 17811491-17811491 on Build GRCh38
rs112243978 Chr.19: 17811479-17811479 on Build GRCh38
rs954994035 Chr.19: 17811804-17811804 on Build GRCh38
rs774213191 Chr.19: 17811640-17811640 on Build GRCh38
rs1002782487 Chr.19: 17811673-17811673 on Build GRCh38
rs774672516 Chr.19: 17811885-17811885 on Build GRCh38
rs778541899 Chr.19: 17811725-17811725 on Build GRCh38
rs1014270822 Chr.19: 17811756-17811756 on Build GRCh38
rs1032203581 Chr.19: 17811696-17811696 on Build GRCh38
rs747823658 Chr.19: 17811727-17811727 on Build GRCh38
rs776991078 Chr.19: 17811682-17811682 on Build GRCh38
rs987400159 Chr.19: 17811523-17811523 on Build GRCh38
rs756015682 Chr.19: 17811706-17811706 on Build GRCh38
rs777983261 Chr.19: 17811592-17811592 on Build GRCh38
rs757613140 Chr.19: 17811695-17811695 on Build GRCh38
rs766057316 Chr.19: 17811811-17811811 on Build GRCh38
rs747770218 Chr.19: 17811659-17811659 on Build GRCh38
rs752915734 Chr.19: 17811593-17811593 on Build GRCh38
rs375696804 Chr.19: 17811521-17811521 on Build GRCh38
rs140504053 Chr.19: 17811691-17811691 on Build GRCh38
rs376780929 Chr.19: 17811759-17811759 on Build GRCh38
rs759940037 Chr.19: 17811773-17811773 on Build GRCh38
rs940827570 Chr.19: 17811565-17811565 on Build GRCh38
rs757970863 Chr.19: 17811832-17811832 on Build GRCh38
rs755169313 Chr.19: 17811584-17811584 on Build GRCh38
rs751714221 Chr.19: 17811796-17811796 on Build GRCh38
rs749176817 Chr.19: 17811711-17811711 on Build GRCh38
rs775504507 Chr.19: 17811789-17811789 on Build GRCh38
rs773673771 Chr.19: 17811664-17811664 on Build GRCh38
rs375344056 Chr.19: 17811854-17811854 on Build GRCh38
rs760584683 Chr.19: 17811807-17811807 on Build GRCh38
rs141805771 Chr.19: 17811699-17811699 on Build GRCh38
rs754575773 Chr.19: 17811715-17811715 on Build GRCh38
rs781401122 Chr.19: 17811763-17811763 on Build GRCh38
rs1035395615 Chr.19: 17811688-17811688 on Build GRCh38
rs756035897 Chr.19: 17811619-17811619 on Build GRCh38
rs745812614 Chr.19: 17811618-17811618 on Build GRCh38
rs769887503 Chr.19: 17811787-17811787 on Build GRCh38
rs754913088 Chr.19: 17811817-17811817 on Build GRCh38
rs753860570 Chr.19: 17811813-17811813 on Build GRCh38
rs183898310 Chr.19: 17811594-17811594 on Build GRCh38
rs370538223 Chr.19: 17811844-17811844 on Build GRCh38
rs768715556 Chr.19: 17811877-17811877 on Build GRCh38
rs113213689 Chr.19: 17811806-17811806 on Build GRCh38
rs780094147 Chr.19: 17811709-17811709 on Build GRCh38
rs763164172 Chr.19: 17811790-17811790 on Build GRCh38
rs761093965 Chr.19: 17811667-17811667 on Build GRCh38
rs1022894287 Chr.19: 17811507-17811507 on Build GRCh38
rs541535699 Chr.19: 17811819-17811819 on Build GRCh38
rs142379648 Chr.19: 17811661-17811661 on Build GRCh38
rs754338056 Chr.19: 17811788-17811788 on Build GRCh38
rs773894534 Chr.19: 17811549-17811549 on Build GRCh38
rs77134098 Chr.19: 17811857-17811857 on Build GRCh38
rs200934033 Chr.19: 17811628-17811628 on Build GRCh38
rs761220548 Chr.19: 17811551-17811551 on Build GRCh38
rs908267161 Chr.19: 17811660-17811660 on Build GRCh38
rs769731266 Chr.19: 17811882-17811882 on Build GRCh38
rs777550767 Chr.19: 17811840-17811840 on Build GRCh38
rs763075853 Chr.19: 17811866-17811866 on Build GRCh38
rs1010014830 Chr.19: 17811692-17811692 on Build GRCh38
rs773462089 Chr.19: 17811735-17811735 on Build GRCh38
rs149610635 Chr.19: 17811843-17811843 on Build GRCh38
rs761916055 Chr.19: 17811907-17811907 on Build GRCh38
rs775555728 Chr.19: 17811865-17811865 on Build GRCh38
rs371110502 Chr.19: 17811705-17811705 on Build GRCh38
rs557248120 Chr.19: 17811627-17811627 on Build GRCh38
rs45580833 Chr.19: 17811502-17811502 on Build GRCh38
rs771785757 Chr.19: 17811732-17811732 on Build GRCh38
rs529304462 Chr.19: 17811501-17811501 on Build GRCh38
rs780945247 Chr.19: 17811603-17811603 on Build GRCh38
rs767989921 Chr.19: 17811530-17811530 on Build GRCh38
rs753889116 Chr.19: 17811564-17811564 on Build GRCh38
rs933327561 Chr.19: 17811516-17811516 on Build GRCh38
rs868859279 Chr.19: 17811616-17811616 on Build GRCh38
rs865860468 Chr.19: 17811750-17811750 on Build GRCh38
rs752196790 Chr.19: 17811826-17811826 on Build GRCh38
rs139357124 Chr.19: 17811694-17811694 on Build GRCh38
rs768037922 Chr.19: 17811639-17811639 on Build GRCh38
rs369707732 Chr.19: 17811544-17811544 on Build GRCh38
rs150461366 Chr.19: 17811739-17811739 on Build GRCh38
rs138588406 Chr.19: 17811920-17811920 on Build GRCh38
rs922162227 Chr.19: 17811514-17811514 on Build GRCh38
rs771198396 Chr.19: 17811753-17811753 on Build GRCh38
rs766851725 Chr.19: 17811672-17811672 on Build GRCh38

View Full Product Details