Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18531679-18532182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP8
Entrez Gene ID
23770
Gene Name
FK506 binding protein 8
Gene Aliases
FKBP38, FKBPr38
UniGene
Hs.173464
Gene Chromosome Location
Chr.19: 18531752-18544077 on Build GRCh38
Transcripts
NM_001308373.1 NM_012181.4 XM_011527863.1 XM_011527864.1

SNP Detail

SNP ID Chromosome Location
rs1029647474 Chr.19: 18531801-18531801 on Build GRCh38
rs867183065 Chr.19: 18531829-18531829 on Build GRCh38
rs889260108 Chr.19: 18531749-18531749 on Build GRCh38
rs1014560091 Chr.19: 18532130-18532130 on Build GRCh38
rs1029266225 Chr.19: 18532086-18532086 on Build GRCh38
rs907632119 Chr.19: 18531871-18531871 on Build GRCh38
rs1033980340 Chr.19: 18532095-18532095 on Build GRCh38
rs1021712309 Chr.19: 18531752-18531752 on Build GRCh38
rs768161236 Chr.19: 18532134-18532134 on Build GRCh38
rs762259240 Chr.19: 18532097-18532097 on Build GRCh38
rs1003272101 Chr.19: 18531706-18531706 on Build GRCh38
rs981963306 Chr.19: 18531818-18531818 on Build GRCh38
rs767900697 Chr.19: 18532139-18532139 on Build GRCh38
rs919231346 Chr.19: 18531939-18531939 on Build GRCh38
rs955102268 Chr.19: 18532119-18532119 on Build GRCh38
rs1058853 Chr.19: 18531892-18531892 on Build GRCh38
rs906182814 Chr.19: 18532061-18532061 on Build GRCh38
rs58564298 Chr.19: 18531958-18531958 on Build GRCh38
rs149127944 Chr.19: 18531928-18531928 on Build GRCh38
rs1047144306 Chr.19: 18531802-18531802 on Build GRCh38
rs938714407 Chr.19: 18531992-18531992 on Build GRCh38
rs947862094 Chr.19: 18531947-18531947 on Build GRCh38
rs1003418015 Chr.19: 18532065-18532065 on Build GRCh38
rs373905025 Chr.19: 18532154-18532154 on Build GRCh38
rs940388440 Chr.19: 18531905-18531905 on Build GRCh38
rs997714881 Chr.19: 18531731-18531731 on Build GRCh38
rs975060453 Chr.19: 18531793-18531793 on Build GRCh38
rs906423081 Chr.19: 18531989-18531989 on Build GRCh38
rs1009187321 Chr.19: 18531814-18531814 on Build GRCh38
rs938725450 Chr.19: 18531824-18531824 on Build GRCh38
rs969271612 Chr.19: 18532148-18532148 on Build GRCh38
rs1035801462 Chr.19: 18531708-18531708 on Build GRCh38
rs3179087 Chr.19: 18531770-18531770 on Build GRCh38
rs1058849 Chr.19: 18531922-18531922 on Build GRCh38
rs1058799 Chr.19: 18532075-18532075 on Build GRCh38
rs893529751 Chr.19: 18532034-18532034 on Build GRCh38
rs972047387 Chr.19: 18531929-18531929 on Build GRCh38
rs907291386 Chr.19: 18531817-18531817 on Build GRCh38
rs1045016923 Chr.19: 18531956-18531956 on Build GRCh38
rs754004129 Chr.19: 18532145-18532145 on Build GRCh38
rs996731561 Chr.19: 18532071-18532071 on Build GRCh38
rs371306435 Chr.19: 18532126-18532126 on Build GRCh38
rs904731949 Chr.19: 18532064-18532064 on Build GRCh38
rs748759133 Chr.19: 18532129-18532129 on Build GRCh38
rs11541425 Chr.19: 18532144-18532144 on Build GRCh38
rs991596739 Chr.19: 18531830-18531830 on Build GRCh38
rs564274350 Chr.19: 18531732-18531732 on Build GRCh38
rs868300966 Chr.19: 18531825-18531825 on Build GRCh38
rs1010235633 Chr.19: 18532030-18532030 on Build GRCh38
rs1032474725 Chr.19: 18531776-18531776 on Build GRCh38
rs1010371627 Chr.19: 18531748-18531748 on Build GRCh38
rs1009215202 Chr.19: 18532036-18532036 on Build GRCh38
rs933580412 Chr.19: 18531939-18531939 on Build GRCh38
rs973236674 Chr.19: 18531913-18531913 on Build GRCh38
rs1051978670 Chr.19: 18531955-18531955 on Build GRCh38
rs1057434276 Chr.19: 18532008-18532008 on Build GRCh38
rs954427330 Chr.19: 18531849-18531849 on Build GRCh38
rs775030656 Chr.19: 18532128-18532128 on Build GRCh38
rs943527226 Chr.19: 18531988-18531988 on Build GRCh38
rs929788033 Chr.19: 18531798-18531798 on Build GRCh38
rs973184223 Chr.19: 18532141-18532141 on Build GRCh38
rs1803588 Chr.19: 18531910-18531910 on Build GRCh38
rs528507244 Chr.19: 18532013-18532013 on Build GRCh38
rs980988668 Chr.19: 18532157-18532157 on Build GRCh38
rs540175404 Chr.19: 18531912-18531912 on Build GRCh38
rs577977094 Chr.19: 18531771-18531771 on Build GRCh38
rs745694170 Chr.19: 18532121-18532121 on Build GRCh38
rs762295462 Chr.19: 18532147-18532147 on Build GRCh38
rs972119509 Chr.19: 18532155-18532155 on Build GRCh38
rs1021660308 Chr.19: 18532042-18532042 on Build GRCh38
rs987521880 Chr.19: 18531851-18531851 on Build GRCh38

View Full Product Details