Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18922514-18923010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX49
Entrez Gene ID
54555
Gene Name
DEAD-box helicase 49
Gene Aliases
R27090_2
UniGene
Hs.143187
Gene Chromosome Location
Chr.19: 18919675-18928633 on Build GRCh38
Transcripts
NM_019070.4 NR_033677.1 XM_011528083.1 XM_011528084.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4750732 c.640A>G p.S214G Chr.19: 18922608-18922608 on Build GRCh38
4075780 c.358C>T p.P120S Chr.19: 18922647-18922647 on Build GRCh38
4075779 c.679C>T p.P227S Chr.19: 18922647-18922647 on Build GRCh38
4075778 c.328G>A p.E110K Chr.19: 18922617-18922617 on Build GRCh38
4075777 c.649G>A p.E217K Chr.19: 18922617-18922617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369552149 Chr.19: 18922580-18922580 on Build GRCh38
rs775026871 Chr.19: 18922665-18922665 on Build GRCh38
rs373378963 Chr.19: 18922690-18922690 on Build GRCh38
rs753913915 Chr.19: 18922707-18922707 on Build GRCh38
rs746972412 Chr.19: 18922716-18922716 on Build GRCh38
rs906519804 Chr.19: 18922798-18922798 on Build GRCh38
rs757519520 Chr.19: 18922579-18922579 on Build GRCh38
rs776218879 Chr.19: 18922613-18922613 on Build GRCh38
rs759486000 Chr.19: 18922788-18922788 on Build GRCh38
rs763506848 Chr.19: 18922635-18922635 on Build GRCh38
rs955036304 Chr.19: 18922907-18922907 on Build GRCh38
rs770744013 Chr.19: 18922604-18922604 on Build GRCh38
rs773768124 Chr.19: 18922766-18922766 on Build GRCh38
rs375677261 Chr.19: 18922664-18922664 on Build GRCh38
rs748702633 Chr.19: 18922741-18922741 on Build GRCh38
rs763064478 Chr.19: 18922801-18922801 on Build GRCh38
rs751432860 Chr.19: 18922792-18922792 on Build GRCh38
rs906569188 Chr.19: 18922971-18922971 on Build GRCh38
rs922249032 Chr.19: 18922685-18922685 on Build GRCh38
rs372388947 Chr.19: 18922632-18922632 on Build GRCh38
rs368968523 Chr.19: 18922631-18922631 on Build GRCh38
rs1005391383 Chr.19: 18922819-18922819 on Build GRCh38
rs987129701 Chr.19: 18922610-18922610 on Build GRCh38
rs527750702 Chr.19: 18922588-18922588 on Build GRCh38
rs1033758711 Chr.19: 18922802-18922802 on Build GRCh38
rs939333337 Chr.19: 18922977-18922977 on Build GRCh38
rs747592114 Chr.19: 18922614-18922614 on Build GRCh38
rs560995094 Chr.19: 18922953-18922953 on Build GRCh38
rs375319625 Chr.19: 18922711-18922711 on Build GRCh38
rs559409160 Chr.19: 18922623-18922623 on Build GRCh38
rs2106660 Chr.19: 18922578-18922578 on Build GRCh38
rs915376679 Chr.19: 18922951-18922951 on Build GRCh38
rs763376503 Chr.19: 18922576-18922576 on Build GRCh38
rs191583884 Chr.19: 18922758-18922758 on Build GRCh38
rs764143898 Chr.19: 18922831-18922831 on Build GRCh38
rs1044895561 Chr.19: 18922912-18922912 on Build GRCh38
rs201989550 Chr.19: 18922536-18922536 on Build GRCh38
rs753193157 Chr.19: 18922566-18922566 on Build GRCh38
rs770732612 Chr.19: 18922915-18922915 on Build GRCh38
rs973044515 Chr.19: 18922883-18922883 on Build GRCh38
rs777648039 Chr.19: 18922713-18922713 on Build GRCh38
rs989678569 Chr.19: 18922949-18922949 on Build GRCh38
rs764520193 Chr.19: 18922577-18922577 on Build GRCh38
rs539892174 Chr.19: 18922762-18922762 on Build GRCh38
rs200288081 Chr.19: 18922727-18922727 on Build GRCh38
rs938748436 Chr.19: 18922935-18922935 on Build GRCh38
rs374874989 Chr.19: 18922590-18922590 on Build GRCh38
rs547478176 Chr.19: 18922950-18922950 on Build GRCh38
rs201216034 Chr.19: 18922592-18922592 on Build GRCh38
rs770619428 Chr.19: 18922546-18922546 on Build GRCh38
rs756355764 Chr.19: 18922701-18922701 on Build GRCh38
rs116675472 Chr.19: 18922735-18922735 on Build GRCh38
rs201391311 Chr.19: 18922750-18922750 on Build GRCh38
rs1054033786 Chr.19: 18922544-18922544 on Build GRCh38
rs945676654 Chr.19: 18922954-18922954 on Build GRCh38
rs780886706 Chr.19: 18922936-18922936 on Build GRCh38
rs878998369 Chr.19: 18922538-18922538 on Build GRCh38
rs776088839 Chr.19: 18922552-18922552 on Build GRCh38
rs574573260 Chr.19: 18922799-18922799 on Build GRCh38
rs759008462 Chr.19: 18922562-18922562 on Build GRCh38
rs764677718 Chr.19: 18922563-18922563 on Build GRCh38
rs780973429 Chr.19: 18922724-18922724 on Build GRCh38
rs545016717 Chr.19: 18922943-18922943 on Build GRCh38
rs961970438 Chr.19: 18922858-18922858 on Build GRCh38
rs756989374 Chr.19: 18922720-18922720 on Build GRCh38
rs144883180 Chr.19: 18922605-18922605 on Build GRCh38
rs910973913 Chr.19: 18922929-18922929 on Build GRCh38
rs865976439 Chr.19: 18922621-18922621 on Build GRCh38
rs371689798 Chr.19: 18922582-18922582 on Build GRCh38
rs370201072 Chr.19: 18922673-18922673 on Build GRCh38
rs1037569273 Chr.19: 18922657-18922657 on Build GRCh38
rs532245322 Chr.19: 18922873-18922873 on Build GRCh38
rs143568852 Chr.19: 18922738-18922738 on Build GRCh38
rs760929072 Chr.19: 18922774-18922774 on Build GRCh38
rs151055717 Chr.19: 18922814-18922814 on Build GRCh38
rs1029314983 Chr.19: 18922889-18922889 on Build GRCh38
rs749089713 Chr.19: 18922909-18922909 on Build GRCh38
rs767988146 Chr.19: 18922760-18922760 on Build GRCh38
rs780001523 Chr.19: 18922584-18922584 on Build GRCh38
rs564920300 Chr.19: 18922967-18922967 on Build GRCh38
rs756201423 Chr.19: 18922583-18922583 on Build GRCh38
rs200119897 Chr.19: 18922706-18922706 on Build GRCh38
rs931431581 Chr.19: 18922689-18922689 on Build GRCh38
rs758325689 Chr.19: 18922709-18922709 on Build GRCh38
rs35802425 Chr.19: 18922633-18922633 on Build GRCh38
rs148028161 Chr.19: 18922739-18922739 on Build GRCh38
rs757104687 Chr.19: 18922914-18922914 on Build GRCh38
rs554962892 Chr.19: 18922775-18922775 on Build GRCh38
rs796154978 Chr.19: 18922987-18922987 on Build GRCh38
rs183041972 Chr.19: 18922871-18922871 on Build GRCh38
rs541107252 Chr.19: 18922757-18922757 on Build GRCh38

View Full Product Details