Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19634119-19634608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMIP
Entrez Gene ID
51291
Gene Name
GEM interacting protein
Gene Aliases
ARHGAP46
UniGene
Hs.49427
Gene Chromosome Location
Chr.19: 19629473-19643667 on Build GRCh38
Transcripts
NM_001288998.1 NM_001288999.1 NM_016573.3 XM_005259927.2 XM_011528048.2 XM_017026861.1 XM_017026862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1524290 c.1987C>T p.P663S Chr.19: 19634604-19634604 on Build GRCh38
993378 c.2040C>T p.D680D Chr.19: 19634551-19634551 on Build GRCh38
4653085 c.2155C>T p.L719L Chr.19: 19634120-19634120 on Build GRCh38
4646595 c.2090C>T p.A697V Chr.19: 19634185-19634185 on Build GRCh38
3892030 c.2009C>T p.S670L Chr.19: 19634582-19634582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765570909 Chr.19: 19634187-19634187 on Build GRCh38
rs779965610 Chr.19: 19634213-19634213 on Build GRCh38
rs1043568638 Chr.19: 19634320-19634320 on Build GRCh38
rs902574476 Chr.19: 19634351-19634351 on Build GRCh38
rs770516747 Chr.19: 19634530-19634530 on Build GRCh38
rs370772118 Chr.19: 19634476-19634476 on Build GRCh38
rs140486275 Chr.19: 19634179-19634179 on Build GRCh38
rs1026029797 Chr.19: 19634177-19634177 on Build GRCh38
rs767621141 Chr.19: 19634582-19634582 on Build GRCh38
rs560142654 Chr.19: 19634383-19634383 on Build GRCh38
rs1015573004 Chr.19: 19634467-19634467 on Build GRCh38
rs757397233 Chr.19: 19634343-19634349 on Build GRCh38
rs529092664 Chr.19: 19634423-19634423 on Build GRCh38
rs746343912 Chr.19: 19634456-19634457 on Build GRCh38
rs200173575 Chr.19: 19634478-19634478 on Build GRCh38
rs980766672 Chr.19: 19634263-19634263 on Build GRCh38
rs768387979 Chr.19: 19634230-19634230 on Build GRCh38
rs771232561 Chr.19: 19634401-19634407 on Build GRCh38
rs765658084 Chr.19: 19634477-19634477 on Build GRCh38
rs936835378 Chr.19: 19634353-19634353 on Build GRCh38
rs760097464 Chr.19: 19634464-19634464 on Build GRCh38
rs779985636 Chr.19: 19634502-19634502 on Build GRCh38
rs375784717 Chr.19: 19634468-19634468 on Build GRCh38
rs761291887 Chr.19: 19634168-19634168 on Build GRCh38
rs775983419 Chr.19: 19634551-19634551 on Build GRCh38
rs777223785 Chr.19: 19634521-19634521 on Build GRCh38
rs1029316590 Chr.19: 19634147-19634147 on Build GRCh38
rs754966075 Chr.19: 19634586-19634586 on Build GRCh38
rs757223620 Chr.19: 19634500-19634500 on Build GRCh38
rs770404237 Chr.19: 19634539-19634539 on Build GRCh38
rs370911598 Chr.19: 19634580-19634580 on Build GRCh38
rs540530651 Chr.19: 19634287-19634287 on Build GRCh38
rs368641677 Chr.19: 19634532-19634532 on Build GRCh38
rs373648012 Chr.19: 19634531-19634531 on Build GRCh38
rs538575908 Chr.19: 19634264-19634264 on Build GRCh38
rs772564661 Chr.19: 19634138-19634138 on Build GRCh38
rs539400941 Chr.19: 19634565-19634565 on Build GRCh38
rs996590270 Chr.19: 19634415-19634415 on Build GRCh38
rs750991171 Chr.19: 19634296-19634296 on Build GRCh38
rs780234716 Chr.19: 19634587-19634588 on Build GRCh38
rs376435223 Chr.19: 19634503-19634503 on Build GRCh38
rs996454529 Chr.19: 19634140-19634140 on Build GRCh38
rs747873153 Chr.19: 19634456-19634456 on Build GRCh38
rs1052750501 Chr.19: 19634302-19634302 on Build GRCh38
rs548752390 Chr.19: 19634425-19634425 on Build GRCh38
rs751732118 Chr.19: 19634497-19634497 on Build GRCh38
rs771569354 Chr.19: 19634465-19634465 on Build GRCh38
rs761908774 Chr.19: 19634581-19634581 on Build GRCh38
rs561873219 Chr.19: 19634455-19634455 on Build GRCh38
rs778728123 Chr.19: 19634512-19634512 on Build GRCh38
rs750477974 Chr.19: 19634585-19634585 on Build GRCh38
rs948153777 Chr.19: 19634247-19634247 on Build GRCh38
rs143664947 Chr.19: 19634479-19634479 on Build GRCh38
rs370255132 Chr.19: 19634509-19634509 on Build GRCh38
rs975071509 Chr.19: 19634547-19634548 on Build GRCh38
rs946534061 Chr.19: 19634309-19634309 on Build GRCh38
rs764385571 Chr.19: 19634572-19634572 on Build GRCh38
rs879217831 Chr.19: 19634566-19634566 on Build GRCh38
rs962499602 Chr.19: 19634199-19634199 on Build GRCh38
rs369818211 Chr.19: 19634190-19634190 on Build GRCh38
rs746542889 Chr.19: 19634527-19634527 on Build GRCh38
rs923264887 Chr.19: 19634280-19634280 on Build GRCh38
rs756138592 Chr.19: 19634209-19634209 on Build GRCh38
rs772835645 Chr.19: 19634462-19634462 on Build GRCh38
rs778564945 Chr.19: 19634238-19634238 on Build GRCh38
rs892399514 Chr.19: 19634304-19634304 on Build GRCh38
rs751571346 Chr.19: 19634205-19634205 on Build GRCh38
rs188195937 Chr.19: 19634195-19634195 on Build GRCh38
rs373482686 Chr.19: 19634487-19634487 on Build GRCh38
rs1026502780 Chr.19: 19634475-19634475 on Build GRCh38
rs771824142 Chr.19: 19634520-19634520 on Build GRCh38
rs1029422870 Chr.19: 19634435-19634435 on Build GRCh38
rs771600609 Chr.19: 19634459-19634459 on Build GRCh38
rs530933438 Chr.19: 19634466-19634466 on Build GRCh38
rs1027866410 Chr.19: 19634535-19634535 on Build GRCh38
rs777952443 Chr.19: 19634200-19634200 on Build GRCh38

View Full Product Details