Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19634933-19635431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMIP
Entrez Gene ID
51291
Gene Name
GEM interacting protein
Gene Aliases
ARHGAP46
UniGene
Hs.49427
Gene Chromosome Location
Chr.19: 19629473-19643667 on Build GRCh38
Transcripts
NM_001288998.1 NM_001288999.1 NM_016573.3 XM_005259927.2 XM_011528048.2 XM_017026861.1 XM_017026862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3970855 c.1725A>C p.I575I Chr.19: 19635049-19635049 on Build GRCh38
4000593 c.1560+10G>C p.? Chr.19: 19635405-19635405 on Build GRCh38
5789620 c.1548G>T p.T516T Chr.19: 19635427-19635427 on Build GRCh38
6195681 c.1624C>T p.L542F Chr.19: 19635150-19635150 on Build GRCh38
3959762 c.1641C>A p.P547P Chr.19: 19635133-19635133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750699499 Chr.19: 19635084-19635084 on Build GRCh38
rs546665082 Chr.19: 19635091-19635091 on Build GRCh38
rs571239033 Chr.19: 19635302-19635302 on Build GRCh38
rs749439675 Chr.19: 19635094-19635094 on Build GRCh38
rs750870350 Chr.19: 19635232-19635232 on Build GRCh38
rs900228246 Chr.19: 19635083-19635083 on Build GRCh38
rs113619686 Chr.19: 19635320-19635320 on Build GRCh38
rs372547969 Chr.19: 19635153-19635153 on Build GRCh38
rs967999793 Chr.19: 19635244-19635244 on Build GRCh38
rs564904493 Chr.19: 19635242-19635242 on Build GRCh38
rs776160578 Chr.19: 19635204-19635217 on Build GRCh38
rs767951734 Chr.19: 19635069-19635069 on Build GRCh38
rs770724061 Chr.19: 19635140-19635140 on Build GRCh38
rs1009615135 Chr.19: 19635217-19635217 on Build GRCh38
rs780297717 Chr.19: 19634983-19634983 on Build GRCh38
rs34970769 Chr.19: 19635092-19635092 on Build GRCh38
rs768702480 Chr.19: 19635397-19635397 on Build GRCh38
rs762495802 Chr.19: 19635187-19635187 on Build GRCh38
rs2304129 Chr.19: 19635405-19635405 on Build GRCh38
rs1021009561 Chr.19: 19635219-19635219 on Build GRCh38
rs779072696 Chr.19: 19635265-19635265 on Build GRCh38
rs754083289 Chr.19: 19635254-19635254 on Build GRCh38
rs987038805 Chr.19: 19635328-19635328 on Build GRCh38
rs375796180 Chr.19: 19635371-19635371 on Build GRCh38
rs969479120 Chr.19: 19635300-19635300 on Build GRCh38
rs377203717 Chr.19: 19634977-19634977 on Build GRCh38
rs930258928 Chr.19: 19635391-19635391 on Build GRCh38
rs775107487 Chr.19: 19635169-19635169 on Build GRCh38
rs780984967 Chr.19: 19635392-19635392 on Build GRCh38
rs184488122 Chr.19: 19635291-19635291 on Build GRCh38
rs868056029 Chr.19: 19635041-19635041 on Build GRCh38
rs767848220 Chr.19: 19635221-19635221 on Build GRCh38
rs896559833 Chr.19: 19635192-19635192 on Build GRCh38
rs1037193670 Chr.19: 19635012-19635012 on Build GRCh38
rs749316704 Chr.19: 19634985-19634985 on Build GRCh38
rs930228584 Chr.19: 19634955-19634955 on Build GRCh38
rs551054738 Chr.19: 19634958-19634958 on Build GRCh38
rs954823244 Chr.19: 19635127-19635127 on Build GRCh38
rs757599218 Chr.19: 19635068-19635068 on Build GRCh38
rs2304128 Chr.19: 19635342-19635342 on Build GRCh38
rs1030883828 Chr.19: 19635229-19635229 on Build GRCh38
rs764811565 Chr.19: 19635037-19635037 on Build GRCh38
rs898131579 Chr.19: 19635016-19635016 on Build GRCh38
rs759340943 Chr.19: 19635378-19635378 on Build GRCh38
rs762442923 Chr.19: 19635055-19635055 on Build GRCh38
rs746830131 Chr.19: 19635014-19635014 on Build GRCh38
rs770728746 Chr.19: 19635030-19635030 on Build GRCh38
rs886101803 Chr.19: 19634975-19634975 on Build GRCh38
rs755087967 Chr.19: 19635100-19635100 on Build GRCh38
rs200920765 Chr.19: 19635000-19635000 on Build GRCh38
rs193153282 Chr.19: 19634965-19634965 on Build GRCh38
rs776249830 Chr.19: 19635031-19635031 on Build GRCh38
rs150339340 Chr.19: 19635102-19635102 on Build GRCh38
rs916669375 Chr.19: 19635289-19635289 on Build GRCh38
rs781314898 Chr.19: 19634970-19634970 on Build GRCh38
rs538009723 Chr.19: 19635271-19635271 on Build GRCh38
rs569483813 Chr.19: 19635191-19635191 on Build GRCh38
rs762399402 Chr.19: 19635220-19635220 on Build GRCh38
rs757221498 Chr.19: 19635388-19635388 on Build GRCh38
rs752080986 Chr.19: 19635067-19635067 on Build GRCh38
rs761110843 Chr.19: 19635241-19635241 on Build GRCh38
rs527487519 Chr.19: 19635260-19635260 on Build GRCh38
rs774984532 Chr.19: 19635042-19635042 on Build GRCh38
rs972479322 Chr.19: 19635353-19635353 on Build GRCh38
rs562554598 Chr.19: 19635271-19635272 on Build GRCh38
rs113298491 Chr.19: 19634956-19634956 on Build GRCh38
rs533401748 Chr.19: 19634982-19634982 on Build GRCh38
rs773189943 Chr.19: 19635009-19635009 on Build GRCh38
rs759254567 Chr.19: 19635033-19635033 on Build GRCh38
rs370305251 Chr.19: 19635123-19635123 on Build GRCh38
rs367726683 Chr.19: 19635155-19635155 on Build GRCh38
rs751933399 Chr.19: 19634964-19634964 on Build GRCh38
rs549666252 Chr.19: 19635108-19635108 on Build GRCh38
rs939859435 Chr.19: 19635351-19635351 on Build GRCh38
rs781052979 Chr.19: 19635141-19635141 on Build GRCh38
rs773690648 Chr.19: 19635204-19635204 on Build GRCh38
rs142924319 Chr.19: 19635151-19635151 on Build GRCh38
rs745781390 Chr.19: 19635058-19635058 on Build GRCh38
rs573265876 Chr.19: 19635404-19635404 on Build GRCh38

View Full Product Details