Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19635297-19635796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMIP
Entrez Gene ID
51291
Gene Name
GEM interacting protein
Gene Aliases
ARHGAP46
UniGene
Hs.49427
Gene Chromosome Location
Chr.19: 19629473-19643667 on Build GRCh38
Transcripts
NM_001288998.1 NM_001288999.1 NM_016573.3 XM_005259927.2 XM_011528048.2 XM_017026861.1 XM_017026862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4000593 c.1560+10G>C p.? Chr.19: 19635405-19635405 on Build GRCh38
3531127 c.1442C>T p.P481L Chr.19: 19635533-19635533 on Build GRCh38
6219670 c.1337C>T p.T446M Chr.19: 19635712-19635712 on Build GRCh38
5789620 c.1548G>T p.T516T Chr.19: 19635427-19635427 on Build GRCh38
1391858 c.1328-2A>G p.? Chr.19: 19635723-19635723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761162347 Chr.19: 19635433-19635433 on Build GRCh38
rs371337326 Chr.19: 19635529-19635529 on Build GRCh38
rs752888327 Chr.19: 19635450-19635450 on Build GRCh38
rs564640311 Chr.19: 19635584-19635584 on Build GRCh38
rs555991456 Chr.19: 19635442-19635442 on Build GRCh38
rs761227217 Chr.19: 19635427-19635427 on Build GRCh38
rs765086067 Chr.19: 19635413-19635413 on Build GRCh38
rs747541980 Chr.19: 19635506-19635506 on Build GRCh38
rs766302473 Chr.19: 19635543-19635543 on Build GRCh38
rs758393068 Chr.19: 19635451-19635451 on Build GRCh38
rs113619686 Chr.19: 19635320-19635320 on Build GRCh38
rs931743991 Chr.19: 19635468-19635468 on Build GRCh38
rs756002441 Chr.19: 19635467-19635467 on Build GRCh38
rs751172520 Chr.19: 19635688-19635688 on Build GRCh38
rs775079808 Chr.19: 19635575-19635575 on Build GRCh38
rs759814406 Chr.19: 19635511-19635511 on Build GRCh38
rs758128475 Chr.19: 19635679-19635679 on Build GRCh38
rs375072311 Chr.19: 19635678-19635678 on Build GRCh38
rs766881890 Chr.19: 19635434-19635434 on Build GRCh38
rs765249347 Chr.19: 19635470-19635470 on Build GRCh38
rs753694296 Chr.19: 19635553-19635553 on Build GRCh38
rs202225255 Chr.19: 19635432-19635432 on Build GRCh38
rs746452486 Chr.19: 19635583-19635583 on Build GRCh38
rs749020673 Chr.19: 19635474-19635474 on Build GRCh38
rs768140288 Chr.19: 19635414-19635414 on Build GRCh38
rs772696325 Chr.19: 19635428-19635428 on Build GRCh38
rs768702480 Chr.19: 19635397-19635397 on Build GRCh38
rs560494801 Chr.19: 19635707-19635707 on Build GRCh38
rs146115077 Chr.19: 19635456-19635456 on Build GRCh38
rs147161315 Chr.19: 19635638-19635638 on Build GRCh38
rs2304129 Chr.19: 19635405-19635405 on Build GRCh38
rs765309752 Chr.19: 19635445-19635445 on Build GRCh38
rs201725101 Chr.19: 19635558-19635558 on Build GRCh38
rs987038805 Chr.19: 19635328-19635328 on Build GRCh38
rs375796180 Chr.19: 19635371-19635371 on Build GRCh38
rs200565808 Chr.19: 19635505-19635505 on Build GRCh38
rs761924954 Chr.19: 19635754-19635754 on Build GRCh38
rs930258928 Chr.19: 19635391-19635391 on Build GRCh38
rs754729901 Chr.19: 19635477-19635477 on Build GRCh38
rs780984967 Chr.19: 19635392-19635392 on Build GRCh38
rs756053760 Chr.19: 19635539-19635539 on Build GRCh38
rs138031462 Chr.19: 19635429-19635429 on Build GRCh38
rs138615732 Chr.19: 19635485-19635485 on Build GRCh38
rs754206094 Chr.19: 19635441-19635441 on Build GRCh38
rs748965805 Chr.19: 19635412-19635412 on Build GRCh38
rs768928335 Chr.19: 19635737-19635737 on Build GRCh38
rs747790075 Chr.19: 19635568-19635568 on Build GRCh38
rs2304128 Chr.19: 19635342-19635342 on Build GRCh38
rs147211671 Chr.19: 19635712-19635712 on Build GRCh38
rs778420161 Chr.19: 19635478-19635478 on Build GRCh38
rs367839582 Chr.19: 19635528-19635528 on Build GRCh38
rs765156944 Chr.19: 19635767-19635767 on Build GRCh38
rs376823436 Chr.19: 19635504-19635504 on Build GRCh38
rs758020885 Chr.19: 19635579-19635579 on Build GRCh38
rs773295556 Chr.19: 19635759-19635759 on Build GRCh38
rs771507788 Chr.19: 19635507-19635507 on Build GRCh38
rs759340943 Chr.19: 19635378-19635378 on Build GRCh38
rs890297939 Chr.19: 19635522-19635522 on Build GRCh38
rs762828487 Chr.19: 19635772-19635772 on Build GRCh38
rs202117195 Chr.19: 19635724-19635724 on Build GRCh38
rs775550340 Chr.19: 19635516-19635516 on Build GRCh38
rs761872001 Chr.19: 19635642-19635642 on Build GRCh38
rs138995642 Chr.19: 19635653-19635653 on Build GRCh38
rs149540867 Chr.19: 19635690-19635690 on Build GRCh38
rs765676198 Chr.19: 19635459-19635459 on Build GRCh38
rs777563458 Chr.19: 19635686-19635686 on Build GRCh38
rs369496932 Chr.19: 19635740-19635740 on Build GRCh38
rs746291562 Chr.19: 19635609-19635609 on Build GRCh38
rs150730686 Chr.19: 19635652-19635652 on Build GRCh38
rs961211023 Chr.19: 19635711-19635711 on Build GRCh38
rs749662765 Chr.19: 19635732-19635732 on Build GRCh38
rs780292004 Chr.19: 19635719-19635719 on Build GRCh38
rs369795775 Chr.19: 19635604-19635604 on Build GRCh38
rs765551565 Chr.19: 19635515-19635515 on Build GRCh38
rs763101975 Chr.19: 19635631-19635631 on Build GRCh38
rs770020220 Chr.19: 19635614-19635614 on Build GRCh38
rs757221498 Chr.19: 19635388-19635388 on Build GRCh38
rs759406226 Chr.19: 19635766-19635766 on Build GRCh38
rs572864861 Chr.19: 19635752-19635752 on Build GRCh38
rs972479322 Chr.19: 19635353-19635353 on Build GRCh38
rs764239391 Chr.19: 19635636-19635636 on Build GRCh38
rs777140420 Chr.19: 19635508-19635508 on Build GRCh38
rs1019964302 Chr.19: 19635685-19635685 on Build GRCh38
rs777439183 Chr.19: 19635582-19635582 on Build GRCh38
rs773835328 Chr.19: 19635430-19635430 on Build GRCh38
rs143679981 Chr.19: 19635518-19635518 on Build GRCh38
rs781773905 Chr.19: 19635703-19635703 on Build GRCh38
rs141198269 Chr.19: 19635648-19635648 on Build GRCh38
rs939859435 Chr.19: 19635351-19635351 on Build GRCh38
rs200090124 Chr.19: 19635457-19635457 on Build GRCh38
rs762820683 Chr.19: 19635678-19635679 on Build GRCh38
rs752501491 Chr.19: 19635771-19635771 on Build GRCh38
rs573265876 Chr.19: 19635404-19635404 on Build GRCh38
rs775881211 Chr.19: 19635624-19635624 on Build GRCh38
rs376815324 Chr.19: 19635559-19635559 on Build GRCh38
rs773920237 Chr.19: 19635418-19635418 on Build GRCh38

View Full Product Details