Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 29715037-29715553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf12
Entrez Gene ID
83636
Gene Name
chromosome 19 open reading frame 12
Gene Aliases
NBIA3, NBIA4, SPG43
UniGene
Hs.529094
Gene Chromosome Location
Chr.19: 29698886-29715789 on Build GRCh38
Transcripts
NM_001031726.3 NM_001256046.1 NM_001256047.1 NM_001282929.1 NM_001282930.1 NM_001282931.1 NM_031448.4

SNP Detail

SNP ID Chromosome Location
rs915700306 Chr.19: 29715404-29715404 on Build GRCh38
rs957655162 Chr.19: 29715251-29715251 on Build GRCh38
rs762850161 Chr.19: 29715471-29715471 on Build GRCh38
rs919857361 Chr.19: 29715203-29715203 on Build GRCh38
rs547652287 Chr.19: 29715285-29715285 on Build GRCh38
rs141427453 Chr.19: 29715499-29715499 on Build GRCh38
rs991636828 Chr.19: 29715388-29715388 on Build GRCh38
rs767536966 Chr.19: 29715256-29715256 on Build GRCh38
rs1010932952 Chr.19: 29715254-29715254 on Build GRCh38
rs970238141 Chr.19: 29715110-29715110 on Build GRCh38
rs981036773 Chr.19: 29715234-29715234 on Build GRCh38
rs775327096 Chr.19: 29715464-29715464 on Build GRCh38
rs759339712 Chr.19: 29715206-29715206 on Build GRCh38
rs986793430 Chr.19: 29715286-29715286 on Build GRCh38
rs770712695 Chr.19: 29715390-29715390 on Build GRCh38
rs1046802410 Chr.19: 29715274-29715274 on Build GRCh38
rs760676412 Chr.19: 29715500-29715500 on Build GRCh38
rs956739128 Chr.19: 29715375-29715375 on Build GRCh38
rs919725902 Chr.19: 29715315-29715315 on Build GRCh38
rs770684342 Chr.19: 29715191-29715191 on Build GRCh38
rs777195384 Chr.19: 29715366-29715366 on Build GRCh38
rs1020515170 Chr.19: 29715279-29715279 on Build GRCh38
rs769044626 Chr.19: 29715364-29715365 on Build GRCh38
rs1013688206 Chr.19: 29715417-29715417 on Build GRCh38
rs929859806 Chr.19: 29715205-29715205 on Build GRCh38
rs1033232140 Chr.19: 29715131-29715131 on Build GRCh38
rs973921359 Chr.19: 29715314-29715314 on Build GRCh38
rs780935305 Chr.19: 29715406-29715406 on Build GRCh38
rs766597352 Chr.19: 29715502-29715502 on Build GRCh38
rs957816284 Chr.19: 29715141-29715141 on Build GRCh38
rs951323300 Chr.19: 29715516-29715516 on Build GRCh38
rs904916822 Chr.19: 29715238-29715238 on Build GRCh38
rs929814469 Chr.19: 29715317-29715317 on Build GRCh38
rs779320924 Chr.19: 29715356-29715356 on Build GRCh38
rs771537351 Chr.19: 29715107-29715107 on Build GRCh38
rs768593539 Chr.19: 29715488-29715488 on Build GRCh38
rs982600203 Chr.19: 29715525-29715525 on Build GRCh38
rs1031083784 Chr.19: 29715373-29715373 on Build GRCh38
rs986908017 Chr.19: 29715182-29715182 on Build GRCh38
rs980230817 Chr.19: 29715111-29715111 on Build GRCh38
rs750497006 Chr.19: 29715259-29715259 on Build GRCh38
rs749553606 Chr.19: 29715147-29715196 on Build GRCh38
rs955575324 Chr.19: 29715463-29715463 on Build GRCh38
rs1043392177 Chr.19: 29715365-29715365 on Build GRCh38
rs964086845 Chr.19: 29715304-29715304 on Build GRCh38
rs1013261098 Chr.19: 29715080-29715080 on Build GRCh38
rs908327495 Chr.19: 29715284-29715284 on Build GRCh38
rs755273072 Chr.19: 29715327-29715327 on Build GRCh38
rs908718489 Chr.19: 29715504-29715504 on Build GRCh38
rs575339528 Chr.19: 29715214-29715214 on Build GRCh38
rs949279088 Chr.19: 29715532-29715532 on Build GRCh38
rs955386974 Chr.19: 29715169-29715169 on Build GRCh38
rs769781525 Chr.19: 29715442-29715442 on Build GRCh38
rs1008234933 Chr.19: 29715482-29715482 on Build GRCh38
rs779321620 Chr.19: 29715103-29715103 on Build GRCh38
rs915824434 Chr.19: 29715290-29715291 on Build GRCh38
rs1047447055 Chr.19: 29715328-29715328 on Build GRCh38
rs540496743 Chr.19: 29715447-29715447 on Build GRCh38
rs753208833 Chr.19: 29715359-29715359 on Build GRCh38
rs1010403290 Chr.19: 29715377-29715377 on Build GRCh38
rs992119127 Chr.19: 29715228-29715228 on Build GRCh38
rs981032547 Chr.19: 29715466-29715466 on Build GRCh38
rs574772189 Chr.19: 29715369-29715369 on Build GRCh38
rs999130832 Chr.19: 29715371-29715371 on Build GRCh38
rs1040655848 Chr.19: 29715329-29715329 on Build GRCh38
rs558329031 Chr.19: 29715152-29715152 on Build GRCh38
rs1000539367 Chr.19: 29715368-29715368 on Build GRCh38
rs537644349 Chr.19: 29715249-29715249 on Build GRCh38
rs746678834 Chr.19: 29715180-29715180 on Build GRCh38
rs754202490 Chr.19: 29715305-29715305 on Build GRCh38
rs775350003 Chr.19: 29715233-29715233 on Build GRCh38
rs959747047 Chr.19: 29715382-29715382 on Build GRCh38
rs942546668 Chr.19: 29715193-29715193 on Build GRCh38
rs917930187 Chr.19: 29715230-29715230 on Build GRCh38
rs758905470 Chr.19: 29715363-29715363 on Build GRCh38
rs956685193 Chr.19: 29715219-29715219 on Build GRCh38
rs766552104 Chr.19: 29715299-29715299 on Build GRCh38
rs896255307 Chr.19: 29715057-29715057 on Build GRCh38
rs560470404 Chr.19: 29715496-29715496 on Build GRCh38
rs968976789 Chr.19: 29715167-29715167 on Build GRCh38
rs776588908 Chr.19: 29715197-29715197 on Build GRCh38
rs949063954 Chr.19: 29715360-29715360 on Build GRCh38
rs747705244 Chr.19: 29715105-29715105 on Build GRCh38
rs983316342 Chr.19: 29715503-29715503 on Build GRCh38
rs996696226 Chr.19: 29715364-29715364 on Build GRCh38
rs931502347 Chr.19: 29715252-29715252 on Build GRCh38
rs745503269 Chr.19: 29715435-29715435 on Build GRCh38
rs1000942011 Chr.19: 29715199-29715199 on Build GRCh38
rs917827556 Chr.19: 29715529-29715529 on Build GRCh38
rs1024954035 Chr.19: 29715452-29715452 on Build GRCh38
rs769866646 Chr.19: 29715218-29715218 on Build GRCh38
rs964139898 Chr.19: 29715494-29715494 on Build GRCh38
rs969399826 Chr.19: 29715465-29715465 on Build GRCh38
rs896170503 Chr.19: 29715227-29715227 on Build GRCh38

View Full Product Details