Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39830364-39830871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK1B
Entrez Gene ID
9149
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
Gene Aliases
AOMS3, MIRK
UniGene
Hs.130988
Gene Chromosome Location
Chr.19: 39825347-39834233 on Build GRCh38
Transcripts
NM_004714.2 NM_006483.2 NM_006484.2 XM_005259398.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7348857 c.92G>A p.R31Q Chr.19: 39830755-39830755 on Build GRCh38
81119 c.156C>T p.L52L Chr.19: 39830691-39830691 on Build GRCh38
310773 c.132G>A p.P44P Chr.19: 39830715-39830715 on Build GRCh38
4078052 c.301G>A p.V101M Chr.19: 39830446-39830446 on Build GRCh38
5823222 c.132G>T p.P44P Chr.19: 39830715-39830715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774906276 Chr.19: 39830805-39830805 on Build GRCh38
rs753510519 Chr.19: 39830629-39830629 on Build GRCh38
rs139697056 Chr.19: 39830517-39830517 on Build GRCh38
rs770222474 Chr.19: 39830753-39830753 on Build GRCh38
rs779889840 Chr.19: 39830747-39830747 on Build GRCh38
rs368230328 Chr.19: 39830807-39830807 on Build GRCh38
rs149054894 Chr.19: 39830682-39830682 on Build GRCh38
rs768807922 Chr.19: 39830602-39830602 on Build GRCh38
rs768717702 Chr.19: 39830668-39830668 on Build GRCh38
rs762706044 Chr.19: 39830832-39830832 on Build GRCh38
rs765600886 Chr.19: 39830611-39830611 on Build GRCh38
rs774335838 Chr.19: 39830525-39830525 on Build GRCh38
rs749102527 Chr.19: 39830510-39830510 on Build GRCh38
rs762086878 Chr.19: 39830610-39830610 on Build GRCh38
rs371524346 Chr.19: 39830447-39830447 on Build GRCh38
rs781197009 Chr.19: 39830766-39830766 on Build GRCh38
rs771927684 Chr.19: 39830588-39830588 on Build GRCh38
rs370702932 Chr.19: 39830768-39830768 on Build GRCh38
rs769928000 Chr.19: 39830677-39830677 on Build GRCh38
rs71356877 Chr.19: 39830815-39830815 on Build GRCh38
rs199768512 Chr.19: 39830767-39830767 on Build GRCh38
rs746098809 Chr.19: 39830489-39830491 on Build GRCh38
rs757118343 Chr.19: 39830576-39830576 on Build GRCh38
rs780629324 Chr.19: 39830483-39830483 on Build GRCh38
rs747094681 Chr.19: 39830597-39830597 on Build GRCh38
rs764858361 Chr.19: 39830817-39830817 on Build GRCh38
rs759459524 Chr.19: 39830526-39830526 on Build GRCh38
rs375205659 Chr.19: 39830484-39830484 on Build GRCh38
rs761241244 Chr.19: 39830816-39830816 on Build GRCh38
rs779866977 Chr.19: 39830660-39830660 on Build GRCh38
rs763465546 Chr.19: 39830615-39830615 on Build GRCh38
rs892096052 Chr.19: 39830474-39830474 on Build GRCh38
rs775793066 Chr.19: 39830568-39830571 on Build GRCh38
rs749011430 Chr.19: 39830566-39830566 on Build GRCh38
rs763058312 Chr.19: 39830567-39830568 on Build GRCh38
rs772161282 Chr.19: 39830531-39830531 on Build GRCh38
rs868166543 Chr.19: 39830477-39830477 on Build GRCh38
rs778722982 Chr.19: 39830653-39830653 on Build GRCh38
rs202191647 Chr.19: 39830542-39830542 on Build GRCh38
rs780909848 Chr.19: 39830387-39830387 on Build GRCh38
rs764800406 Chr.19: 39830735-39830735 on Build GRCh38
rs765576879 Chr.19: 39830541-39830541 on Build GRCh38
rs753422313 Chr.19: 39830731-39830731 on Build GRCh38
rs754222688 Chr.19: 39830448-39830448 on Build GRCh38
rs750039090 Chr.19: 39830818-39830818 on Build GRCh38
rs538648311 Chr.19: 39830414-39830414 on Build GRCh38
rs776728304 Chr.19: 39830672-39830672 on Build GRCh38
rs890210647 Chr.19: 39830795-39830795 on Build GRCh38
rs769967128 Chr.19: 39830498-39830500 on Build GRCh38
rs34587974 Chr.19: 39830764-39830764 on Build GRCh38
rs149848740 Chr.19: 39830456-39830456 on Build GRCh38
rs373240762 Chr.19: 39830741-39830741 on Build GRCh38
rs766827263 Chr.19: 39830710-39830710 on Build GRCh38
rs755676515 Chr.19: 39830625-39830625 on Build GRCh38
rs755764449 Chr.19: 39830557-39830557 on Build GRCh38
rs1031501785 Chr.19: 39830481-39830481 on Build GRCh38
rs763677106 Chr.19: 39830716-39830716 on Build GRCh38
rs778614271 Chr.19: 39830579-39830579 on Build GRCh38
rs201029953 Chr.19: 39830435-39830435 on Build GRCh38
rs747810492 Chr.19: 39830402-39830402 on Build GRCh38
rs767029305 Chr.19: 39830552-39830552 on Build GRCh38
rs374944306 Chr.19: 39830464-39830464 on Build GRCh38
rs777584233 Chr.19: 39830562-39830562 on Build GRCh38
rs764303085 Chr.19: 39830612-39830614 on Build GRCh38
rs142076547 Chr.19: 39830715-39830715 on Build GRCh38
rs775598716 Chr.19: 39830589-39830589 on Build GRCh38
rs144370928 Chr.19: 39830538-39830538 on Build GRCh38
rs777848605 Chr.19: 39830771-39830771 on Build GRCh38
rs776215861 Chr.19: 39830812-39830812 on Build GRCh38
rs754494908 Chr.19: 39830471-39830471 on Build GRCh38
rs1009184227 Chr.19: 39830782-39830782 on Build GRCh38
rs777674210 Chr.19: 39830493-39830493 on Build GRCh38
rs949540060 Chr.19: 39830609-39830609 on Build GRCh38
rs751375246 Chr.19: 39830835-39830835 on Build GRCh38
rs780672797 Chr.19: 39830714-39830714 on Build GRCh38
rs749594362 Chr.19: 39830772-39830772 on Build GRCh38
rs760400549 Chr.19: 39830709-39830709 on Build GRCh38
rs587777380 Chr.19: 39830478-39830478 on Build GRCh38
rs766136186 Chr.19: 39830834-39830834 on Build GRCh38
rs745682157 Chr.19: 39830655-39830655 on Build GRCh38
rs200104062 Chr.19: 39830539-39830539 on Build GRCh38
rs543090482 Chr.19: 39830555-39830555 on Build GRCh38
rs761488320 Chr.19: 39830734-39830734 on Build GRCh38
rs763244310 Chr.19: 39830691-39830691 on Build GRCh38
rs367643250 Chr.19: 39830443-39830443 on Build GRCh38
rs1047427536 Chr.19: 39830725-39830725 on Build GRCh38
rs561806068 Chr.19: 39830617-39830617 on Build GRCh38
rs201768057 Chr.19: 39830407-39830407 on Build GRCh38
rs776820071 Chr.19: 39830607-39830607 on Build GRCh38
rs369809566 Chr.19: 39830537-39830537 on Build GRCh38
rs146381130 Chr.19: 39830465-39830465 on Build GRCh38
rs763767264 Chr.19: 39830628-39830628 on Build GRCh38
rs756909087 Chr.19: 39830650-39830650 on Build GRCh38
rs775483019 Chr.19: 39830535-39830535 on Build GRCh38
rs779162841 Chr.19: 39830794-39830794 on Build GRCh38
rs1008052704 Chr.19: 39830755-39830755 on Build GRCh38
rs1000920589 Chr.19: 39830851-39830851 on Build GRCh38
rs769617031 Chr.19: 39830489-39830489 on Build GRCh38
rs146458379 Chr.19: 39830553-39830553 on Build GRCh38
rs201770775 Chr.19: 39830431-39830431 on Build GRCh38
rs748344036 Chr.19: 39830676-39830676 on Build GRCh38
rs55687541 Chr.19: 39830442-39830442 on Build GRCh38
rs918137811 Chr.19: 39830572-39830572 on Build GRCh38
rs760848598 Chr.19: 39830446-39830446 on Build GRCh38
rs774998571 Chr.19: 39830711-39830711 on Build GRCh38
rs1051493794 Chr.19: 39830789-39830789 on Build GRCh38
rs770939435 Chr.19: 39830511-39830511 on Build GRCh38
rs577567514 Chr.19: 39830846-39830846 on Build GRCh38
rs949236193 Chr.19: 39830724-39830724 on Build GRCh38
rs762958782 Chr.19: 39830421-39830421 on Build GRCh38
rs773496510 Chr.19: 39830613-39830613 on Build GRCh38
rs772738846 Chr.19: 39830811-39830811 on Build GRCh38
rs576584969 Chr.19: 39830657-39830657 on Build GRCh38
rs373850179 Chr.19: 39830545-39830545 on Build GRCh38
rs377044422 Chr.19: 39830624-39830624 on Build GRCh38
rs199710767 Chr.19: 39830756-39830756 on Build GRCh38
rs375929795 Chr.19: 39830434-39830434 on Build GRCh38
rs745796756 Chr.19: 39830586-39830586 on Build GRCh38

View Full Product Details