Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40328322-40328818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf47
Entrez Gene ID
126526
Gene Name
chromosome 19 open reading frame 47
Gene Aliases
-
UniGene
Hs.631557
Gene Chromosome Location
Chr.19: 40292743-40348527 on Build GRCh38
Transcripts
NM_001256440.1 NM_001256441.1 NR_046202.1 XM_005258520.2 XM_011526460.2 XM_017026291.1 XM_017026292.1 XM_017026293.1 XM_017026294.1 XM_017026295.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5741584 c.415G>A p.A139T Chr.19: 40328548-40328548 on Build GRCh38
3960072 c.474G>T p.R158S Chr.19: 40328489-40328489 on Build GRCh38
5748064 c.470C>A p.P157H Chr.19: 40328493-40328493 on Build GRCh38
1393790 c.439C>T p.L147L Chr.19: 40328524-40328524 on Build GRCh38
69908 c.253C>T p.P85S Chr.19: 40328509-40328509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769116992 Chr.19: 40328388-40328388 on Build GRCh38
rs68001945 Chr.19: 40328386-40328386 on Build GRCh38
rs930003977 Chr.19: 40328444-40328444 on Build GRCh38
rs1051214532 Chr.19: 40328780-40328780 on Build GRCh38
rs781275707 Chr.19: 40328553-40328553 on Build GRCh38
rs896768277 Chr.19: 40328613-40328613 on Build GRCh38
rs780350868 Chr.19: 40328520-40328520 on Build GRCh38
rs778716614 Chr.19: 40328465-40328465 on Build GRCh38
rs775117745 Chr.19: 40328493-40328493 on Build GRCh38
rs765167742 Chr.19: 40328591-40328591 on Build GRCh38
rs1052696242 Chr.19: 40328724-40328724 on Build GRCh38
rs368893832 Chr.19: 40328561-40328561 on Build GRCh38
rs753599001 Chr.19: 40328497-40328497 on Build GRCh38
rs117383339 Chr.19: 40328578-40328578 on Build GRCh38
rs186319756 Chr.19: 40328660-40328660 on Build GRCh38
rs918597820 Chr.19: 40328434-40328434 on Build GRCh38
rs765122607 Chr.19: 40328483-40328483 on Build GRCh38
rs1018071246 Chr.19: 40328618-40328618 on Build GRCh38
rs962348822 Chr.19: 40328700-40328700 on Build GRCh38
rs780401575 Chr.19: 40328608-40328608 on Build GRCh38
rs974012562 Chr.19: 40328624-40328624 on Build GRCh38
rs1018527223 Chr.19: 40328766-40328766 on Build GRCh38
rs756364856 Chr.19: 40328605-40328605 on Build GRCh38
rs9973304 Chr.19: 40328487-40328487 on Build GRCh38
rs753703104 Chr.19: 40328459-40328459 on Build GRCh38
rs768311426 Chr.19: 40328580-40328580 on Build GRCh38
rs765169309 Chr.19: 40328508-40328508 on Build GRCh38
rs200907567 Chr.19: 40328476-40328476 on Build GRCh38
rs755172310 Chr.19: 40328548-40328548 on Build GRCh38
rs181273110 Chr.19: 40328392-40328392 on Build GRCh38
rs773854808 Chr.19: 40328414-40328414 on Build GRCh38
rs529231922 Chr.19: 40328666-40328666 on Build GRCh38
rs1026015530 Chr.19: 40328637-40328637 on Build GRCh38
rs143732838 Chr.19: 40328375-40328375 on Build GRCh38
rs905235173 Chr.19: 40328661-40328661 on Build GRCh38
rs918022134 Chr.19: 40328447-40328447 on Build GRCh38
rs979584489 Chr.19: 40328786-40328786 on Build GRCh38
rs550526282 Chr.19: 40328714-40328714 on Build GRCh38
rs772350005 Chr.19: 40328370-40328370 on Build GRCh38
rs1010352203 Chr.19: 40328566-40328566 on Build GRCh38
rs917982548 Chr.19: 40328681-40328681 on Build GRCh38
rs1016678928 Chr.19: 40328440-40328440 on Build GRCh38
rs34077058 Chr.19: 40328524-40328524 on Build GRCh38
rs962560455 Chr.19: 40328622-40328622 on Build GRCh38
rs764561143 Chr.19: 40328382-40328384 on Build GRCh38
rs142469218 Chr.19: 40328488-40328488 on Build GRCh38
rs948132423 Chr.19: 40328696-40328696 on Build GRCh38
rs759051347 Chr.19: 40328415-40328415 on Build GRCh38
rs945715931 Chr.19: 40328760-40328760 on Build GRCh38
rs768896670 Chr.19: 40328706-40328706 on Build GRCh38
rs969889550 Chr.19: 40328412-40328412 on Build GRCh38
rs763189574 Chr.19: 40328381-40328381 on Build GRCh38
rs375894468 Chr.19: 40328596-40328596 on Build GRCh38
rs761651467 Chr.19: 40328585-40328585 on Build GRCh38
rs750438538 Chr.19: 40328510-40328510 on Build GRCh38
rs749563573 Chr.19: 40328655-40328655 on Build GRCh38
rs377203741 Chr.19: 40328701-40328701 on Build GRCh38
rs778221261 Chr.19: 40328574-40328574 on Build GRCh38
rs951399214 Chr.19: 40328638-40328638 on Build GRCh38
rs749858681 Chr.19: 40328577-40328577 on Build GRCh38
rs372214531 Chr.19: 40328445-40328445 on Build GRCh38
rs759024663 Chr.19: 40328492-40328492 on Build GRCh38
rs745955296 Chr.19: 40328369-40328369 on Build GRCh38
rs1013732704 Chr.19: 40328628-40328628 on Build GRCh38
rs955307375 Chr.19: 40328726-40328726 on Build GRCh38
rs1043359762 Chr.19: 40328573-40328573 on Build GRCh38
rs556483719 Chr.19: 40328346-40328346 on Build GRCh38
rs755871542 Chr.19: 40328458-40328458 on Build GRCh38
rs200776998 Chr.19: 40328494-40328494 on Build GRCh38
rs745899352 Chr.19: 40328472-40328472 on Build GRCh38
rs772717818 Chr.19: 40328430-40328431 on Build GRCh38
rs751821059 Chr.19: 40328541-40328541 on Build GRCh38
rs765795709 Chr.19: 40328413-40328413 on Build GRCh38
rs752394590 Chr.19: 40328457-40328457 on Build GRCh38
rs146550099 Chr.19: 40328484-40328484 on Build GRCh38
rs375538388 Chr.19: 40328499-40328499 on Build GRCh38
rs779390490 Chr.19: 40328725-40328725 on Build GRCh38
rs748505221 Chr.19: 40328557-40328557 on Build GRCh38
rs760433414 Chr.19: 40328453-40328454 on Build GRCh38
rs776976388 Chr.19: 40328391-40328391 on Build GRCh38
rs892741833 Chr.19: 40328755-40328755 on Build GRCh38
rs1006723357 Chr.19: 40328695-40328695 on Build GRCh38
rs747278159 Chr.19: 40328478-40328478 on Build GRCh38
rs936864918 Chr.19: 40328715-40328715 on Build GRCh38

View Full Product Details