Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40807404-40807914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGLN2
Entrez Gene ID
112398
Gene Name
egl-9 family hypoxia inducible factor 2
Gene Aliases
EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
UniGene
Hs.515417
Gene Chromosome Location
Chr.19: 40799143-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Gene Symbol
RAB4B-EGLN2
Entrez Gene ID
100529264
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
Gene Aliases
RERT-lncRNA
UniGene
Hs.631539
Gene Chromosome Location
Chr.19: 40778219-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3226292 c.1138C>T p.R380W Chr.19: 40807521-40807521 on Build GRCh38
1393898 c.1199T>C p.V400A Chr.19: 40807839-40807839 on Build GRCh38
1205058 c.1208C>A p.P403Q Chr.19: 40807848-40807848 on Build GRCh38
996836 c.1214C>T p.T405M Chr.19: 40807854-40807854 on Build GRCh38
5933806 c.1101-4C>T p.? Chr.19: 40807480-40807480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867216902 Chr.19: 40807831-40807831 on Build GRCh38
rs533007215 Chr.19: 40807466-40807466 on Build GRCh38
rs759650902 Chr.19: 40807796-40807796 on Build GRCh38
rs369625239 Chr.19: 40807785-40807785 on Build GRCh38
rs756663605 Chr.19: 40807468-40807468 on Build GRCh38
rs753171757 Chr.19: 40807618-40807618 on Build GRCh38
rs757656607 Chr.19: 40807796-40807797 on Build GRCh38
rs751712367 Chr.19: 40807497-40807497 on Build GRCh38
rs748628682 Chr.19: 40807439-40807439 on Build GRCh38
rs768203932 Chr.19: 40807686-40807686 on Build GRCh38
rs374000050 Chr.19: 40807882-40807882 on Build GRCh38
rs747824587 Chr.19: 40807867-40807867 on Build GRCh38
rs771677442 Chr.19: 40807545-40807545 on Build GRCh38
rs557408858 Chr.19: 40807615-40807615 on Build GRCh38
rs573438011 Chr.19: 40807701-40807701 on Build GRCh38
rs540090034 Chr.19: 40807662-40807662 on Build GRCh38
rs769400397 Chr.19: 40807873-40807873 on Build GRCh38
rs775256963 Chr.19: 40807546-40807546 on Build GRCh38
rs920211641 Chr.19: 40807824-40807824 on Build GRCh38
rs148552273 Chr.19: 40807534-40807534 on Build GRCh38
rs776292763 Chr.19: 40807580-40807580 on Build GRCh38
rs377024627 Chr.19: 40807793-40807793 on Build GRCh38
rs758539500 Chr.19: 40807662-40807662 on Build GRCh38
rs768313432 Chr.19: 40807567-40807567 on Build GRCh38
rs756595762 Chr.19: 40807809-40807809 on Build GRCh38
rs375891916 Chr.19: 40807571-40807571 on Build GRCh38
rs749027019 Chr.19: 40807881-40807881 on Build GRCh38
rs764146764 Chr.19: 40807768-40807770 on Build GRCh38
rs541177885 Chr.19: 40807692-40807692 on Build GRCh38
rs761767834 Chr.19: 40807480-40807480 on Build GRCh38
rs772998542 Chr.19: 40807874-40807874 on Build GRCh38
rs566290717 Chr.19: 40807475-40807475 on Build GRCh38
rs199656256 Chr.19: 40807597-40807597 on Build GRCh38
rs769507515 Chr.19: 40807673-40807673 on Build GRCh38
rs759060721 Chr.19: 40807464-40807464 on Build GRCh38
rs35362523 Chr.19: 40807487-40807487 on Build GRCh38
rs767700388 Chr.19: 40807800-40807800 on Build GRCh38
rs780859020 Chr.19: 40807861-40807861 on Build GRCh38
rs576003805 Chr.19: 40807648-40807648 on Build GRCh38
rs779415979 Chr.19: 40807848-40807848 on Build GRCh38
rs746460179 Chr.19: 40807670-40807670 on Build GRCh38
rs756542887 Chr.19: 40807622-40807622 on Build GRCh38
rs750351983 Chr.19: 40807591-40807591 on Build GRCh38
rs535824878 Chr.19: 40807610-40807610 on Build GRCh38
rs758455498 Chr.19: 40807490-40807490 on Build GRCh38
rs141698095 Chr.19: 40807854-40807854 on Build GRCh38
rs372632840 Chr.19: 40807525-40807525 on Build GRCh38
rs202046460 Chr.19: 40807828-40807828 on Build GRCh38
rs778464176 Chr.19: 40807647-40807647 on Build GRCh38
rs112039467 Chr.19: 40807552-40807552 on Build GRCh38
rs769543429 Chr.19: 40807784-40807784 on Build GRCh38
rs751674826 Chr.19: 40807782-40807785 on Build GRCh38
rs912538777 Chr.19: 40807730-40807730 on Build GRCh38
rs777247518 Chr.19: 40807437-40807437 on Build GRCh38
rs760266730 Chr.19: 40807549-40807549 on Build GRCh38
rs748000680 Chr.19: 40807780-40807780 on Build GRCh38
rs551165982 Chr.19: 40807467-40807467 on Build GRCh38
rs755163007 Chr.19: 40807614-40807614 on Build GRCh38
rs376964412 Chr.19: 40807763-40807763 on Build GRCh38
rs755182350 Chr.19: 40807499-40807499 on Build GRCh38
rs913307549 Chr.19: 40807687-40807687 on Build GRCh38
rs974384367 Chr.19: 40807689-40807689 on Build GRCh38
rs765090515 Chr.19: 40807486-40807486 on Build GRCh38
rs748480774 Chr.19: 40807529-40807529 on Build GRCh38
rs765524046 Chr.19: 40807567-40807570 on Build GRCh38
rs753016066 Chr.19: 40807589-40807589 on Build GRCh38
rs779459694 Chr.19: 40807663-40807663 on Build GRCh38
rs763073865 Chr.19: 40807593-40807593 on Build GRCh38
rs781739072 Chr.19: 40807802-40807802 on Build GRCh38
rs745438271 Chr.19: 40807541-40807541 on Build GRCh38
rs754503213 Chr.19: 40807855-40807855 on Build GRCh38
rs34636581 Chr.19: 40807532-40807532 on Build GRCh38
rs367678368 Chr.19: 40807583-40807583 on Build GRCh38
rs774310360 Chr.19: 40807794-40807794 on Build GRCh38
rs377069095 Chr.19: 40807795-40807795 on Build GRCh38
rs772947190 Chr.19: 40807789-40807789 on Build GRCh38
rs541202116 Chr.19: 40807586-40807586 on Build GRCh38
rs372528500 Chr.19: 40807598-40807598 on Build GRCh38
rs780386346 Chr.19: 40807494-40807494 on Build GRCh38
rs370322331 Chr.19: 40807844-40807844 on Build GRCh38
rs375372070 Chr.19: 40807750-40807750 on Build GRCh38
rs374186657 Chr.19: 40807461-40807461 on Build GRCh38
rs764574945 Chr.19: 40807825-40807825 on Build GRCh38

View Full Product Details