Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40808121-40808615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2T1P
Entrez Gene ID
171523
Gene Name
cytochrome P450 family 2 subfamily T member 1, pseudogene
Gene Aliases
CYP2T2P
UniGene
Hs.656713
Gene Chromosome Location
Chr.19: 40808495-40811392 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Gene Symbol
EGLN2
Entrez Gene ID
112398
Gene Name
egl-9 family hypoxia inducible factor 2
Gene Aliases
EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
UniGene
Hs.515417
Gene Chromosome Location
Chr.19: 40799143-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

Gene Symbol
RAB4B-EGLN2
Entrez Gene ID
100529264
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
Gene Aliases
RERT-lncRNA
UniGene
Hs.631539
Gene Chromosome Location
Chr.19: 40778219-40808441 on Build GRCh38
Transcripts
NM_053046.3 NM_080732.3 NR_037791.1

SNP Detail

SNP ID Chromosome Location
rs906864411 Chr.19: 40808163-40808163 on Build GRCh38
rs370534429 Chr.19: 40808226-40808226 on Build GRCh38
rs995672910 Chr.19: 40808222-40808222 on Build GRCh38
rs546812287 Chr.19: 40808145-40808145 on Build GRCh38
rs34788614 Chr.19: 40808281-40808281 on Build GRCh38
rs1041975014 Chr.19: 40808161-40808161 on Build GRCh38
rs908685566 Chr.19: 40808232-40808232 on Build GRCh38
rs914741130 Chr.19: 40808492-40808492 on Build GRCh38
rs573512848 Chr.19: 40808328-40808328 on Build GRCh38
rs1019574391 Chr.19: 40808304-40808304 on Build GRCh38
rs964507361 Chr.19: 40808583-40808583 on Build GRCh38
rs117370786 Chr.19: 40808288-40808288 on Build GRCh38
rs938955472 Chr.19: 40808558-40808558 on Build GRCh38
rs1020221545 Chr.19: 40808473-40808473 on Build GRCh38
rs967553811 Chr.19: 40808506-40808506 on Build GRCh38
rs777357583 Chr.19: 40808577-40808577 on Build GRCh38
rs977567027 Chr.19: 40808514-40808514 on Build GRCh38
rs540150244 Chr.19: 40808286-40808286 on Build GRCh38
rs1019690029 Chr.19: 40808576-40808576 on Build GRCh38
rs568530112 Chr.19: 40808194-40808194 on Build GRCh38
rs550954361 Chr.19: 40808238-40808238 on Build GRCh38
rs759633008 Chr.19: 40808585-40808585 on Build GRCh38
rs965305079 Chr.19: 40808327-40808327 on Build GRCh38
rs775695710 Chr.19: 40808257-40808257 on Build GRCh38
rs770467475 Chr.19: 40808185-40808185 on Build GRCh38
rs761239810 Chr.19: 40808189-40808189 on Build GRCh38
rs766906437 Chr.19: 40808267-40808270 on Build GRCh38
rs902982322 Chr.19: 40808511-40808511 on Build GRCh38
rs1029414835 Chr.19: 40808343-40808343 on Build GRCh38
rs1026813327 Chr.19: 40808274-40808274 on Build GRCh38
rs191342080 Chr.19: 40808578-40808578 on Build GRCh38
rs186950462 Chr.19: 40808422-40808422 on Build GRCh38
rs1011471368 Chr.19: 40808472-40808472 on Build GRCh38
rs761827320 Chr.19: 40808205-40808205 on Build GRCh38
rs34474563 Chr.19: 40808305-40808305 on Build GRCh38
rs555219470 Chr.19: 40808556-40808556 on Build GRCh38
rs946298805 Chr.19: 40808160-40808160 on Build GRCh38
rs990664374 Chr.19: 40808477-40808477 on Build GRCh38
rs142583688 Chr.19: 40808557-40808557 on Build GRCh38
rs1055893151 Chr.19: 40808379-40808379 on Build GRCh38
rs776807547 Chr.19: 40808563-40808563 on Build GRCh38
rs983010834 Chr.19: 40808221-40808221 on Build GRCh38

View Full Product Details