Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40850119-40850624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A6
Entrez Gene ID
1548
Gene Name
cytochrome P450 family 2 subfamily A member 6
Gene Aliases
CPA6, CYP2A, CYP2A3, CYPIIA6, P450C2A, P450PB
UniGene
Hs.250615
Gene Chromosome Location
Chr.19: 40843538-40850447 on Build GRCh38
Transcripts
NM_000762.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74312 c.18G>A p.M6I Chr.19: 40850409-40850409 on Build GRCh38
996851 c.15G>A p.G5G Chr.19: 40850412-40850412 on Build GRCh38
4078386 c.80G>C p.R27T Chr.19: 40850347-40850347 on Build GRCh38
4562702 c.93G>A p.G31G Chr.19: 40850334-40850334 on Build GRCh38
4560591 c.84G>A p.K28K Chr.19: 40850343-40850343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs570229222 Chr.19: 40850175-40850175 on Build GRCh38
rs760760778 Chr.19: 40850410-40850410 on Build GRCh38
rs778163533 Chr.19: 40850202-40850202 on Build GRCh38
rs761123162 Chr.19: 40850203-40850203 on Build GRCh38
rs779528823 Chr.19: 40850523-40850523 on Build GRCh38
rs370605094 Chr.19: 40850377-40850377 on Build GRCh38
rs553074588 Chr.19: 40850229-40850229 on Build GRCh38
rs761726982 Chr.19: 40850393-40850393 on Build GRCh38
rs747479415 Chr.19: 40850300-40850300 on Build GRCh38
rs530706018 Chr.19: 40850418-40850418 on Build GRCh38
rs781710360 Chr.19: 40850434-40850434 on Build GRCh38
rs183167151 Chr.19: 40850537-40850537 on Build GRCh38
rs752873366 Chr.19: 40850254-40850254 on Build GRCh38
rs771845020 Chr.19: 40850454-40850454 on Build GRCh38
rs370019771 Chr.19: 40850340-40850340 on Build GRCh38
rs749480640 Chr.19: 40850206-40850206 on Build GRCh38
rs8192720 Chr.19: 40850405-40850405 on Build GRCh38
rs528755675 Chr.19: 40850518-40850518 on Build GRCh38
rs756293343 Chr.19: 40850256-40850256 on Build GRCh38
rs780557694 Chr.19: 40850530-40850530 on Build GRCh38
rs191965231 Chr.19: 40850150-40850150 on Build GRCh38
rs760521860 Chr.19: 40850456-40850456 on Build GRCh38
rs149703281 Chr.19: 40850408-40850408 on Build GRCh38
rs749484240 Chr.19: 40850266-40850266 on Build GRCh38
rs151097223 Chr.19: 40850385-40850385 on Build GRCh38
rs762603502 Chr.19: 40850231-40850231 on Build GRCh38
rs767510919 Chr.19: 40850395-40850395 on Build GRCh38
rs777933432 Chr.19: 40850257-40850257 on Build GRCh38
rs751357687 Chr.19: 40850240-40850240 on Build GRCh38
rs28399434 Chr.19: 40850414-40850414 on Build GRCh38
rs750666218 Chr.19: 40850423-40850423 on Build GRCh38
rs767380497 Chr.19: 40850338-40850338 on Build GRCh38
rs141128348 Chr.19: 40850144-40850144 on Build GRCh38
rs781482841 Chr.19: 40850191-40850191 on Build GRCh38
rs755519888 Chr.19: 40850432-40850432 on Build GRCh38
rs144279119 Chr.19: 40850296-40850296 on Build GRCh38
rs774076511 Chr.19: 40850403-40850403 on Build GRCh38
rs377713545 Chr.19: 40850323-40850323 on Build GRCh38
rs1137115 Chr.19: 40850376-40850376 on Build GRCh38
rs1023971687 Chr.19: 40850507-40850507 on Build GRCh38
rs766052140 Chr.19: 40850318-40850318 on Build GRCh38
rs28399433 Chr.19: 40850474-40850474 on Build GRCh38
rs112069025 Chr.19: 40850594-40850594 on Build GRCh38
rs535366981 Chr.19: 40850227-40850227 on Build GRCh38
rs575050301 Chr.19: 40850299-40850299 on Build GRCh38
rs201793905 Chr.19: 40850453-40850453 on Build GRCh38
rs775409921 Chr.19: 40850409-40850409 on Build GRCh38
rs563981730 Chr.19: 40850348-40850348 on Build GRCh38
rs761592914 Chr.19: 40850478-40850478 on Build GRCh38
rs762573512 Chr.19: 40850309-40850309 on Build GRCh38
rs770479084 Chr.19: 40850443-40850443 on Build GRCh38
rs547290288 Chr.19: 40850520-40850520 on Build GRCh38
rs746024795 Chr.19: 40850367-40850367 on Build GRCh38
rs112784824 Chr.19: 40850322-40850322 on Build GRCh38
rs775462957 Chr.19: 40850455-40850455 on Build GRCh38
rs764682418 Chr.19: 40850228-40850228 on Build GRCh38
rs139764932 Chr.19: 40850358-40850358 on Build GRCh38
rs8192723 Chr.19: 40850149-40850149 on Build GRCh38
rs745452690 Chr.19: 40850532-40850532 on Build GRCh38
rs369561609 Chr.19: 40850233-40850233 on Build GRCh38
rs770181229 Chr.19: 40850201-40850201 on Build GRCh38
rs757376520 Chr.19: 40850420-40850420 on Build GRCh38
rs376218645 Chr.19: 40850352-40850352 on Build GRCh38
rs558145012 Chr.19: 40850320-40850320 on Build GRCh38
rs989014391 Chr.19: 40850174-40850174 on Build GRCh38
rs140179029 Chr.19: 40850217-40850217 on Build GRCh38
rs372992538 Chr.19: 40850345-40850345 on Build GRCh38
rs980668046 Chr.19: 40850156-40850156 on Build GRCh38
rs756056996 Chr.19: 40850347-40850347 on Build GRCh38
rs933628804 Chr.19: 40850211-40850211 on Build GRCh38
rs8192721 Chr.19: 40850283-40850283 on Build GRCh38
rs765187344 Chr.19: 40850422-40850422 on Build GRCh38
rs781244650 Chr.19: 40850243-40850243 on Build GRCh38
rs139344949 Chr.19: 40850292-40850292 on Build GRCh38
rs753944846 Chr.19: 40850354-40850354 on Build GRCh38
rs546724333 Chr.19: 40850364-40850364 on Build GRCh38
rs373933186 Chr.19: 40850500-40850500 on Build GRCh38
rs28399436 Chr.19: 40850190-40850190 on Build GRCh38
rs768283281 Chr.19: 40850468-40850468 on Build GRCh38
rs772654177 Chr.19: 40850308-40850308 on Build GRCh38
rs748234903 Chr.19: 40850199-40850199 on Build GRCh38
rs778218518 Chr.19: 40850437-40850437 on Build GRCh38
rs769302279 Chr.19: 40850305-40850305 on Build GRCh38
rs972908489 Chr.19: 40850569-40850569 on Build GRCh38
rs572785621 Chr.19: 40850369-40850369 on Build GRCh38
rs776154526 Chr.19: 40850219-40850219 on Build GRCh38
rs28399435 Chr.19: 40850341-40850341 on Build GRCh38
rs1001522596 Chr.19: 40850521-40850521 on Build GRCh38
rs374253823 Chr.19: 40850278-40850278 on Build GRCh38
rs8192722 Chr.19: 40850218-40850218 on Build GRCh38
rs758561850 Chr.19: 40850424-40850424 on Build GRCh38
rs780456650 Chr.19: 40850427-40850427 on Build GRCh38
rs373993802 Chr.19: 40850389-40850389 on Build GRCh38
rs72549432 Chr.19: 40850411-40850411 on Build GRCh38
rs761287104 Chr.19: 40850220-40850220 on Build GRCh38
rs941512214 Chr.19: 40850181-40850181 on Build GRCh38
rs910090342 Chr.19: 40850178-40850178 on Build GRCh38
rs369357668 Chr.19: 40850212-40850212 on Build GRCh38

View Full Product Details