Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41089915-41090418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A13
Entrez Gene ID
1553
Gene Name
cytochrome P450 family 2 subfamily A member 13
Gene Aliases
CPAD, CYP2A, CYPIIA13
UniGene
Hs.567252
Gene Chromosome Location
Chr.19: 41088451-41096195 on Build GRCh38
Transcripts
NM_000766.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3823156 c.493+1G>T p.? Chr.19: 41090197-41090197 on Build GRCh38
6639144 c.427C>T p.R143C Chr.19: 41090130-41090130 on Build GRCh38
3823155 c.367C>T p.R123C Chr.19: 41090070-41090070 on Build GRCh38
996911 c.345C>T p.G115G Chr.19: 41090048-41090048 on Build GRCh38
3823154 c.350C>T p.A117V Chr.19: 41090053-41090053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759128544 Chr.19: 41090142-41090142 on Build GRCh38
rs771339064 Chr.19: 41090377-41090377 on Build GRCh38
rs536561804 Chr.19: 41090184-41090184 on Build GRCh38
rs201711732 Chr.19: 41090185-41090185 on Build GRCh38
rs376978363 Chr.19: 41090091-41090091 on Build GRCh38
rs751583562 Chr.19: 41090242-41090242 on Build GRCh38
rs761427976 Chr.19: 41090387-41090387 on Build GRCh38
rs753627766 Chr.19: 41090161-41090161 on Build GRCh38
rs769755765 Chr.19: 41090029-41090029 on Build GRCh38
rs113406157 Chr.19: 41090000-41090000 on Build GRCh38
rs371158573 Chr.19: 41090390-41090390 on Build GRCh38
rs747584567 Chr.19: 41090193-41090194 on Build GRCh38
rs1004718486 Chr.19: 41090298-41090298 on Build GRCh38
rs748082919 Chr.19: 41090028-41090028 on Build GRCh38
rs773878128 Chr.19: 41090202-41090202 on Build GRCh38
rs750404962 Chr.19: 41090232-41090232 on Build GRCh38
rs547131472 Chr.19: 41089984-41089984 on Build GRCh38
rs765943632 Chr.19: 41090048-41090048 on Build GRCh38
rs67927428 Chr.19: 41090365-41090365 on Build GRCh38
rs758007882 Chr.19: 41090047-41090047 on Build GRCh38
rs202084076 Chr.19: 41089971-41089971 on Build GRCh38
rs763010987 Chr.19: 41090399-41090399 on Build GRCh38
rs758346061 Chr.19: 41090311-41090311 on Build GRCh38
rs1014813059 Chr.19: 41090301-41090301 on Build GRCh38
rs965879310 Chr.19: 41090310-41090310 on Build GRCh38
rs760443855 Chr.19: 41090219-41090219 on Build GRCh38
rs200738334 Chr.19: 41090175-41090175 on Build GRCh38
rs746517204 Chr.19: 41090378-41090378 on Build GRCh38
rs778665701 Chr.19: 41090191-41090192 on Build GRCh38
rs761622314 Chr.19: 41090227-41090227 on Build GRCh38
rs372587042 Chr.19: 41090111-41090111 on Build GRCh38
rs199521387 Chr.19: 41090110-41090110 on Build GRCh38
rs545287361 Chr.19: 41090008-41090008 on Build GRCh38
rs756623104 Chr.19: 41090009-41090009 on Build GRCh38
rs768857745 Chr.19: 41090098-41090098 on Build GRCh38
rs749719085 Chr.19: 41090209-41090209 on Build GRCh38
rs375894456 Chr.19: 41090398-41090398 on Build GRCh38
rs201720562 Chr.19: 41090139-41090139 on Build GRCh38
rs775911719 Chr.19: 41090037-41090037 on Build GRCh38
rs769802930 Chr.19: 41089996-41089996 on Build GRCh38
rs780349763 Chr.19: 41090132-41090132 on Build GRCh38
rs867835424 Chr.19: 41090186-41090186 on Build GRCh38
rs757175323 Chr.19: 41090120-41090120 on Build GRCh38
rs187611451 Chr.19: 41089991-41089991 on Build GRCh38
rs745954355 Chr.19: 41090125-41090125 on Build GRCh38
rs754847632 Chr.19: 41090026-41090026 on Build GRCh38
rs554686517 Chr.19: 41089964-41089964 on Build GRCh38
rs757765881 Chr.19: 41090007-41090007 on Build GRCh38
rs1645690 Chr.19: 41090228-41090228 on Build GRCh38
rs915204201 Chr.19: 41090351-41090351 on Build GRCh38
rs758241453 Chr.19: 41090234-41090234 on Build GRCh38
rs776300476 Chr.19: 41090385-41090385 on Build GRCh38
rs770939986 Chr.19: 41090031-41090031 on Build GRCh38
rs755958303 Chr.19: 41090113-41090113 on Build GRCh38
rs200698076 Chr.19: 41090015-41090015 on Build GRCh38
rs1049983500 Chr.19: 41090008-41090008 on Build GRCh38
rs149323050 Chr.19: 41089988-41089988 on Build GRCh38
rs755015057 Chr.19: 41090247-41090247 on Build GRCh38
rs112337232 Chr.19: 41090177-41090177 on Build GRCh38
rs200559239 Chr.19: 41090212-41090212 on Build GRCh38
rs759905830 Chr.19: 41090004-41090004 on Build GRCh38
rs138225940 Chr.19: 41090089-41090089 on Build GRCh38
rs751293256 Chr.19: 41090049-41090049 on Build GRCh38
rs199643958 Chr.19: 41090133-41090133 on Build GRCh38
rs779869857 Chr.19: 41090021-41090021 on Build GRCh38
rs940102144 Chr.19: 41089987-41089987 on Build GRCh38
rs930590711 Chr.19: 41089957-41089957 on Build GRCh38
rs766032618 Chr.19: 41090107-41090107 on Build GRCh38
rs770224275 Chr.19: 41090201-41090201 on Build GRCh38
rs747969619 Chr.19: 41090054-41090054 on Build GRCh38
rs146067729 Chr.19: 41090145-41090145 on Build GRCh38
rs754582979 Chr.19: 41090110-41090110 on Build GRCh38
rs572363151 Chr.19: 41090320-41090320 on Build GRCh38
rs781100904 Chr.19: 41090027-41090027 on Build GRCh38
rs771991041 Chr.19: 41090131-41090131 on Build GRCh38
rs112900999 Chr.19: 41090272-41090272 on Build GRCh38
rs558508405 Chr.19: 41090195-41090195 on Build GRCh38
rs548359879 Chr.19: 41089940-41089940 on Build GRCh38
rs57082577 Chr.19: 41090105-41090106 on Build GRCh38
rs1035199013 Chr.19: 41090059-41090059 on Build GRCh38
rs201516283 Chr.19: 41090019-41090019 on Build GRCh38
rs779032831 Chr.19: 41090124-41090124 on Build GRCh38
rs372684091 Chr.19: 41089952-41089952 on Build GRCh38
rs954270239 Chr.19: 41090321-41090321 on Build GRCh38
rs756577449 Chr.19: 41090366-41090366 on Build GRCh38
rs1027269258 Chr.19: 41090278-41090278 on Build GRCh38
rs75703079 Chr.19: 41090053-41090053 on Build GRCh38
rs983228735 Chr.19: 41089961-41089961 on Build GRCh38
rs959041488 Chr.19: 41090065-41090065 on Build GRCh38
rs141733817 Chr.19: 41090064-41090064 on Build GRCh38
rs758540832 Chr.19: 41090183-41090183 on Build GRCh38
rs778251538 Chr.19: 41090367-41090367 on Build GRCh38
rs768745189 Chr.19: 41090191-41090191 on Build GRCh38
rs754246141 Chr.19: 41090044-41090044 on Build GRCh38
rs199984436 Chr.19: 41090108-41090108 on Build GRCh38
rs372079170 Chr.19: 41089999-41089999 on Build GRCh38
rs762374305 Chr.19: 41090101-41090101 on Build GRCh38
rs372525815 Chr.19: 41090237-41090237 on Build GRCh38
rs774588226 Chr.19: 41090033-41090033 on Build GRCh38
rs746607856 Chr.19: 41090025-41090025 on Build GRCh38
rs566841796 Chr.19: 41090100-41090101 on Build GRCh38
rs532238603 Chr.19: 41090166-41090166 on Build GRCh38
rs751713325 Chr.19: 41090001-41090001 on Build GRCh38
rs571383026 Chr.19: 41090144-41090144 on Build GRCh38
rs773666440 Chr.19: 41089994-41089994 on Build GRCh38
rs150550130 Chr.19: 41090081-41090081 on Build GRCh38
rs753821369 Chr.19: 41090117-41090117 on Build GRCh38
rs767829290 Chr.19: 41090007-41090007 on Build GRCh38
rs762466120 Chr.19: 41090134-41090134 on Build GRCh38
rs776210715 Chr.19: 41090221-41090221 on Build GRCh38
rs764669755 Chr.19: 41090043-41090043 on Build GRCh38
rs761118346 Chr.19: 41090097-41090097 on Build GRCh38
rs781731358 Chr.19: 41090192-41090192 on Build GRCh38
rs767920103 Chr.19: 41090036-41090036 on Build GRCh38
rs560496474 Chr.19: 41090030-41090030 on Build GRCh38
rs764008365 Chr.19: 41090154-41090154 on Build GRCh38
rs749130656 Chr.19: 41090066-41090066 on Build GRCh38
rs894867889 Chr.19: 41090256-41090256 on Build GRCh38
rs760222098 Chr.19: 41090148-41090148 on Build GRCh38
rs781476160 Chr.19: 41090017-41090018 on Build GRCh38
rs749864512 Chr.19: 41090372-41090372 on Build GRCh38
rs777222816 Chr.19: 41090099-41090099 on Build GRCh38
rs170054 Chr.19: 41089951-41089951 on Build GRCh38
rs748526923 Chr.19: 41090194-41090194 on Build GRCh38
rs761155616 Chr.19: 41090038-41090038 on Build GRCh38
rs777400164 Chr.19: 41090114-41090114 on Build GRCh38
rs554034652 Chr.19: 41090277-41090277 on Build GRCh38
rs112733608 Chr.19: 41090331-41090331 on Build GRCh38
rs764714733 Chr.19: 41090011-41090011 on Build GRCh38
rs951722999 Chr.19: 41090288-41090288 on Build GRCh38

View Full Product Details