Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41193115-41193618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2S1
Entrez Gene ID
29785
Gene Name
cytochrome P450 family 2 subfamily S member 1
Gene Aliases
CYPIIS1
UniGene
Hs.98370
Gene Chromosome Location
Chr.19: 41193207-41207539 on Build GRCh38
Transcripts
NM_030622.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6839918 c.164C>T p.S55L Chr.19: 41193428-41193428 on Build GRCh38
1393933 c.171C>T p.L57L Chr.19: 41193435-41193435 on Build GRCh38
3823163 c.59C>T p.T20M Chr.19: 41193323-41193323 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs528991454 Chr.19: 41193225-41193225 on Build GRCh38
rs761191600 Chr.19: 41193357-41193357 on Build GRCh38
rs1014363746 Chr.19: 41193366-41193366 on Build GRCh38
rs77508431 Chr.19: 41193414-41193414 on Build GRCh38
rs764485658 Chr.19: 41193551-41193551 on Build GRCh38
rs942273064 Chr.19: 41193338-41193338 on Build GRCh38
rs181317441 Chr.19: 41193396-41193396 on Build GRCh38
rs577645029 Chr.19: 41193570-41193570 on Build GRCh38
rs942415144 Chr.19: 41193527-41193527 on Build GRCh38
rs531882193 Chr.19: 41193570-41193572 on Build GRCh38
rs889662079 Chr.19: 41193143-41193143 on Build GRCh38
rs557730144 Chr.19: 41193543-41193543 on Build GRCh38
rs575040372 Chr.19: 41193491-41193491 on Build GRCh38
rs781443736 Chr.19: 41193262-41193262 on Build GRCh38
rs749447120 Chr.19: 41193261-41193261 on Build GRCh38
rs202041018 Chr.19: 41193277-41193277 on Build GRCh38
rs773929588 Chr.19: 41193276-41193276 on Build GRCh38
rs539457724 Chr.19: 41193362-41193362 on Build GRCh38
rs910020561 Chr.19: 41193272-41193272 on Build GRCh38
rs962846957 Chr.19: 41193470-41193470 on Build GRCh38
rs909491571 Chr.19: 41193330-41193330 on Build GRCh38
rs369980187 Chr.19: 41193278-41193278 on Build GRCh38
rs1031795435 Chr.19: 41193185-41193185 on Build GRCh38
rs750706590 Chr.19: 41193365-41193365 on Build GRCh38
rs747522945 Chr.19: 41193254-41193254 on Build GRCh38
rs758485767 Chr.19: 41193373-41193373 on Build GRCh38
rs781642659 Chr.19: 41193416-41193416 on Build GRCh38
rs970688523 Chr.19: 41193256-41193256 on Build GRCh38
rs3840911 Chr.19: 41193243-41193245 on Build GRCh38
rs768600512 Chr.19: 41193499-41193499 on Build GRCh38
rs1052236872 Chr.19: 41193587-41193587 on Build GRCh38
rs546793639 Chr.19: 41193179-41193179 on Build GRCh38
rs956023683 Chr.19: 41193198-41193198 on Build GRCh38
rs12460196 Chr.19: 41193539-41193539 on Build GRCh38
rs766411039 Chr.19: 41193387-41193387 on Build GRCh38
rs1030842948 Chr.19: 41193350-41193350 on Build GRCh38
rs528554522 Chr.19: 41193153-41193153 on Build GRCh38
rs923274570 Chr.19: 41193295-41193295 on Build GRCh38
rs957048655 Chr.19: 41193286-41193286 on Build GRCh38
rs373729517 Chr.19: 41193263-41193263 on Build GRCh38
rs748526945 Chr.19: 41193468-41193468 on Build GRCh38
rs1050684318 Chr.19: 41193293-41193294 on Build GRCh38
rs772201496 Chr.19: 41193243-41193261 on Build GRCh38
rs138605512 Chr.19: 41193240-41193242 on Build GRCh38
rs568806591 Chr.19: 41193358-41193358 on Build GRCh38
rs553614240 Chr.19: 41193480-41193480 on Build GRCh38
rs760565459 Chr.19: 41193322-41193322 on Build GRCh38
rs561510333 Chr.19: 41193245-41193245 on Build GRCh38
rs1030842768 Chr.19: 41193500-41193500 on Build GRCh38
rs748901437 Chr.19: 41193273-41193273 on Build GRCh38
rs976154000 Chr.19: 41193287-41193287 on Build GRCh38
rs903677745 Chr.19: 41193328-41193328 on Build GRCh38
rs929716286 Chr.19: 41193436-41193436 on Build GRCh38
rs758543117 Chr.19: 41193228-41193228 on Build GRCh38
rs1012583037 Chr.19: 41193236-41193236 on Build GRCh38
rs529159973 Chr.19: 41193275-41193275 on Build GRCh38
rs1056180742 Chr.19: 41193445-41193445 on Build GRCh38
rs892871310 Chr.19: 41193356-41193356 on Build GRCh38
rs903708533 Chr.19: 41193172-41193172 on Build GRCh38
rs201574261 Chr.19: 41193238-41193239 on Build GRCh38
rs889471733 Chr.19: 41193311-41193311 on Build GRCh38
rs189383179 Chr.19: 41193545-41193545 on Build GRCh38
rs869115069 Chr.19: 41193241-41193243 on Build GRCh38
rs111844943 Chr.19: 41193479-41193479 on Build GRCh38
rs35166159 Chr.19: 41193534-41193534 on Build GRCh38
rs763906465 Chr.19: 41193329-41193329 on Build GRCh38
rs776430271 Chr.19: 41193337-41193337 on Build GRCh38
rs773280588 Chr.19: 41193298-41193299 on Build GRCh38
rs897990238 Chr.19: 41193435-41193435 on Build GRCh38
rs1022519608 Chr.19: 41193158-41193158 on Build GRCh38
rs373031805 Chr.19: 41193319-41193319 on Build GRCh38
rs992789596 Chr.19: 41193223-41193223 on Build GRCh38
rs970104453 Chr.19: 41193410-41193410 on Build GRCh38
rs903807818 Chr.19: 41193494-41193494 on Build GRCh38
rs755339656 Chr.19: 41193258-41193258 on Build GRCh38
rs972835539 Chr.19: 41193471-41193471 on Build GRCh38
rs184450806 Chr.19: 41193516-41193516 on Build GRCh38

View Full Product Details