Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41584247-41584748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM21
Entrez Gene ID
90273
Gene Name
carcinoembryonic antigen related cell adhesion molecule 21
Gene Aliases
CEACAM3, R29124_1
UniGene
Hs.655885
Gene Chromosome Location
Chr.19: 41549518-41586844 on Build GRCh38
Transcripts
NM_001098506.3 NM_001288773.2 NM_001290113.1 NM_033543.5 XM_005278396.4 XM_005278397.3 XM_011527450.1 XM_011527452.1 XM_017027430.1 XM_017027431.1 XM_017027432.1 XM_017027433.1 XM_017027434.1 XM_017027435.1 XM_017027436.1 XM_017027437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3534422 c.749T>C p.L250P Chr.19: 41584395-41584395 on Build GRCh38
4653142 c.718G>T p.G240C Chr.19: 41584364-41584364 on Build GRCh38
4557517 c.731G>A p.G244E Chr.19: 41584377-41584377 on Build GRCh38
1393981 c.701-1G>A p.? Chr.19: 41584346-41584346 on Build GRCh38
3227248 c.784C>T p.R262* Chr.19: 41584430-41584430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115885052 Chr.19: 41584323-41584323 on Build GRCh38
rs782306284 Chr.19: 41584431-41584431 on Build GRCh38
rs184081521 Chr.19: 41584528-41584528 on Build GRCh38
rs535087962 Chr.19: 41584350-41584350 on Build GRCh38
rs939286103 Chr.19: 41584367-41584367 on Build GRCh38
rs782818774 Chr.19: 41584468-41584468 on Build GRCh38
rs531215335 Chr.19: 41584723-41584723 on Build GRCh38
rs372128642 Chr.19: 41584283-41584283 on Build GRCh38
rs781848124 Chr.19: 41584504-41584504 on Build GRCh38
rs782046893 Chr.19: 41584346-41584346 on Build GRCh38
rs373871010 Chr.19: 41584481-41584481 on Build GRCh38
rs782215060 Chr.19: 41584302-41584302 on Build GRCh38
rs782726389 Chr.19: 41584363-41584363 on Build GRCh38
rs984778150 Chr.19: 41584273-41584273 on Build GRCh38
rs782029303 Chr.19: 41584344-41584344 on Build GRCh38
rs952267914 Chr.19: 41584537-41584537 on Build GRCh38
rs781833242 Chr.19: 41584482-41584482 on Build GRCh38
rs782119771 Chr.19: 41584326-41584326 on Build GRCh38
rs768819009 Chr.19: 41584462-41584462 on Build GRCh38
rs782376796 Chr.19: 41584542-41584542 on Build GRCh38
rs782046463 Chr.19: 41584360-41584361 on Build GRCh38
rs80270538 Chr.19: 41584687-41584687 on Build GRCh38
rs545812661 Chr.19: 41584546-41584546 on Build GRCh38
rs782753932 Chr.19: 41584358-41584358 on Build GRCh38
rs782792210 Chr.19: 41584356-41584357 on Build GRCh38
rs782362815 Chr.19: 41584458-41584458 on Build GRCh38
rs782289171 Chr.19: 41584720-41584720 on Build GRCh38
rs530489352 Chr.19: 41584618-41584618 on Build GRCh38
rs782613223 Chr.19: 41584461-41584461 on Build GRCh38
rs782631269 Chr.19: 41584394-41584394 on Build GRCh38
rs916535363 Chr.19: 41584682-41584682 on Build GRCh38
rs781942727 Chr.19: 41584297-41584297 on Build GRCh38
rs782422095 Chr.19: 41584716-41584716 on Build GRCh38
rs557907751 Chr.19: 41584549-41584549 on Build GRCh38
rs781820785 Chr.19: 41584457-41584458 on Build GRCh38
rs572969116 Chr.19: 41584560-41584560 on Build GRCh38
rs981520853 Chr.19: 41584614-41584614 on Build GRCh38
rs782088097 Chr.19: 41584441-41584441 on Build GRCh38
rs782498595 Chr.19: 41584486-41584486 on Build GRCh38
rs782571725 Chr.19: 41584376-41584376 on Build GRCh38
rs990604067 Chr.19: 41584660-41584660 on Build GRCh38
rs373575632 Chr.19: 41584375-41584375 on Build GRCh38
rs782173632 Chr.19: 41584277-41584277 on Build GRCh38
rs111323306 Chr.19: 41584548-41584548 on Build GRCh38
rs200587219 Chr.19: 41584379-41584379 on Build GRCh38
rs782065863 Chr.19: 41584476-41584476 on Build GRCh38
rs781889103 Chr.19: 41584363-41584363 on Build GRCh38
rs782634413 Chr.19: 41584488-41584488 on Build GRCh38
rs375544850 Chr.19: 41584650-41584650 on Build GRCh38
rs568421412 Chr.19: 41584430-41584430 on Build GRCh38
rs782401788 Chr.19: 41584411-41584411 on Build GRCh38
rs782477896 Chr.19: 41584385-41584385 on Build GRCh38
rs202112556 Chr.19: 41584408-41584408 on Build GRCh38
rs370515100 Chr.19: 41584467-41584467 on Build GRCh38
rs191889923 Chr.19: 41584494-41584494 on Build GRCh38
rs782360498 Chr.19: 41584304-41584304 on Build GRCh38
rs570982859 Chr.19: 41584327-41584327 on Build GRCh38
rs782495486 Chr.19: 41584352-41584352 on Build GRCh38
rs996325773 Chr.19: 41584519-41584519 on Build GRCh38
rs772294544 Chr.19: 41584633-41584633 on Build GRCh38
rs557436798 Chr.19: 41584495-41584495 on Build GRCh38
rs927786089 Chr.19: 41584359-41584359 on Build GRCh38
rs782128831 Chr.19: 41584304-41584304 on Build GRCh38
rs901986310 Chr.19: 41584507-41584507 on Build GRCh38
rs782650253 Chr.19: 41584417-41584417 on Build GRCh38
rs543799265 Chr.19: 41584666-41584666 on Build GRCh38
rs940785293 Chr.19: 41584287-41584287 on Build GRCh38
rs114252198 Chr.19: 41584727-41584727 on Build GRCh38
rs539895104 Chr.19: 41584647-41584647 on Build GRCh38
rs946594555 Chr.19: 41584693-41584693 on Build GRCh38
rs539900760 Chr.19: 41584570-41584570 on Build GRCh38

View Full Product Details