Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41621564-41622066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM4
Entrez Gene ID
1089
Gene Name
carcinoembryonic antigen related cell adhesion molecule 4
Gene Aliases
CGM7, CGM7_HUMAN, NCA
UniGene
Hs.12
Gene Chromosome Location
Chr.19: 41616986-41627087 on Build GRCh38
Transcripts
NM_001817.3 XM_006722977.3 XM_011526343.1 XM_011526345.2 XM_011526348.2 XM_011526349.2 XM_011526350.2 XM_011526352.1 XM_011526354.1 XM_011526357.2 XM_017026199.1 XM_017026200.1 XM_017026201.1 XM_017026202.1 XM_017026203.1 XM_017026204.1 XM_017026205.1 XM_017026206.1 XM_017026207.1 XM_017026208.1 XM_017026209.1 XM_017026210.1 XM_017026211.1 XM_017026212.1 XM_017026213.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
997061 c.457G>A p.V153I Chr.19: 41621736-41621736 on Build GRCh38
997062 c.456C>T p.A152A Chr.19: 41621737-41621737 on Build GRCh38
997060 c.517T>G p.F173V Chr.19: 41621676-41621676 on Build GRCh38
6229437 c.459C>T p.V153V Chr.19: 41621734-41621734 on Build GRCh38
4406048 c.531C>T p.S177S Chr.19: 41621662-41621662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782629761 Chr.19: 41621625-41621625 on Build GRCh38
rs902175662 Chr.19: 41621662-41621662 on Build GRCh38
rs779481582 Chr.19: 41621962-41621962 on Build GRCh38
rs8111279 Chr.19: 41621829-41621829 on Build GRCh38
rs782457919 Chr.19: 41621690-41621690 on Build GRCh38
rs781909625 Chr.19: 41621689-41621689 on Build GRCh38
rs557614281 Chr.19: 41621753-41621753 on Build GRCh38
rs369353624 Chr.19: 41621615-41621615 on Build GRCh38
rs782480356 Chr.19: 41621708-41621708 on Build GRCh38
rs782417152 Chr.19: 41621729-41621729 on Build GRCh38
rs148761752 Chr.19: 41621734-41621734 on Build GRCh38
rs782687928 Chr.19: 41621663-41621663 on Build GRCh38
rs979332522 Chr.19: 41621833-41621833 on Build GRCh38
rs146702199 Chr.19: 41621731-41621731 on Build GRCh38
rs562602221 Chr.19: 41621906-41621906 on Build GRCh38
rs377465277 Chr.19: 41622017-41622017 on Build GRCh38
rs782159542 Chr.19: 41621659-41621659 on Build GRCh38
rs782617338 Chr.19: 41621637-41621637 on Build GRCh38
rs782582724 Chr.19: 41621697-41621697 on Build GRCh38
rs1006364987 Chr.19: 41621931-41621931 on Build GRCh38
rs781855029 Chr.19: 41621789-41621789 on Build GRCh38
rs183900488 Chr.19: 41621963-41621963 on Build GRCh38
rs782259603 Chr.19: 41621638-41621638 on Build GRCh38
rs2019253 Chr.19: 41621605-41621605 on Build GRCh38
rs782574576 Chr.19: 41621620-41621620 on Build GRCh38
rs781818092 Chr.19: 41621622-41621622 on Build GRCh38
rs901626563 Chr.19: 41621891-41621891 on Build GRCh38
rs1015202007 Chr.19: 41621970-41621970 on Build GRCh38
rs367565204 Chr.19: 41621646-41621646 on Build GRCh38
rs73931735 Chr.19: 41621930-41621930 on Build GRCh38
rs935002469 Chr.19: 41621858-41621858 on Build GRCh38
rs782228228 Chr.19: 41621753-41621754 on Build GRCh38
rs782470943 Chr.19: 41621768-41621768 on Build GRCh38
rs782210982 Chr.19: 41621706-41621706 on Build GRCh38
rs782481867 Chr.19: 41621624-41621624 on Build GRCh38
rs146123396 Chr.19: 41621724-41621724 on Build GRCh38
rs191524049 Chr.19: 41621645-41621645 on Build GRCh38
rs555540003 Chr.19: 41621825-41621825 on Build GRCh38
rs118113337 Chr.19: 41621846-41621846 on Build GRCh38
rs781803136 Chr.19: 41621675-41621675 on Build GRCh38
rs782727091 Chr.19: 41621661-41621661 on Build GRCh38
rs782105663 Chr.19: 41621742-41621742 on Build GRCh38
rs1039980364 Chr.19: 41621886-41621886 on Build GRCh38
rs199597961 Chr.19: 41621793-41621793 on Build GRCh38
rs898046554 Chr.19: 41622018-41622018 on Build GRCh38
rs1035786924 Chr.19: 41621771-41621771 on Build GRCh38
rs201492288 Chr.19: 41621733-41621733 on Build GRCh38
rs568416449 Chr.19: 41621618-41621618 on Build GRCh38
rs781960425 Chr.19: 41621737-41621737 on Build GRCh38
rs368076616 Chr.19: 41621736-41621736 on Build GRCh38
rs782008619 Chr.19: 41621758-41621759 on Build GRCh38
rs79017845 Chr.19: 41621617-41621617 on Build GRCh38
rs375597956 Chr.19: 41621730-41621730 on Build GRCh38
rs782384332 Chr.19: 41621756-41621757 on Build GRCh38
rs929501 Chr.19: 41621919-41621919 on Build GRCh38
rs546839119 Chr.19: 41622035-41622035 on Build GRCh38
rs781815366 Chr.19: 41621761-41621761 on Build GRCh38
rs199931005 Chr.19: 41621735-41621735 on Build GRCh38
rs981417106 Chr.19: 41621769-41621769 on Build GRCh38
rs34542832 Chr.19: 41621738-41621739 on Build GRCh38
rs782639080 Chr.19: 41621714-41621714 on Build GRCh38
rs782407753 Chr.19: 41621641-41621641 on Build GRCh38
rs140151631 Chr.19: 41621760-41621760 on Build GRCh38
rs782172302 Chr.19: 41621755-41621755 on Build GRCh38
rs547195118 Chr.19: 41621608-41621608 on Build GRCh38
rs990709297 Chr.19: 41621788-41621788 on Build GRCh38
rs528785504 Chr.19: 41621985-41621985 on Build GRCh38
rs373663683 Chr.19: 41621642-41621642 on Build GRCh38
rs2019254 Chr.19: 41621600-41621600 on Build GRCh38
rs782079711 Chr.19: 41621682-41621682 on Build GRCh38
rs782773078 Chr.19: 41621752-41621752 on Build GRCh38
rs200832407 Chr.19: 41621778-41621778 on Build GRCh38
rs371323836 Chr.19: 41621655-41621655 on Build GRCh38
rs782221247 Chr.19: 41622026-41622026 on Build GRCh38
rs782754820 Chr.19: 41621687-41621687 on Build GRCh38
rs782671637 Chr.19: 41621804-41621804 on Build GRCh38
rs1006460135 Chr.19: 41621740-41621740 on Build GRCh38
rs1040120130 Chr.19: 41621698-41621698 on Build GRCh38
rs557074639 Chr.19: 41621688-41621688 on Build GRCh38

View Full Product Details