Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41761171-41761679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM6
Entrez Gene ID
4680
Gene Name
carcinoembryonic antigen related cell adhesion molecule 6
Gene Aliases
CD66c, CEAL, NCA
UniGene
Hs.466814
Gene Chromosome Location
Chr.19: 41755421-41772211 on Build GRCh38
Transcripts
NM_002483.6 XM_011526990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
712741 c.563G>C p.S188T Chr.19: 41761387-41761387 on Build GRCh38
7077413 c.508G>A p.E170K Chr.19: 41761332-41761332 on Build GRCh38
6035761 c.593T>G p.M198R Chr.19: 41761417-41761417 on Build GRCh38
997115 c.620G>T p.R207M Chr.19: 41761444-41761444 on Build GRCh38
7080996 c.654G>C p.Q218H Chr.19: 41761478-41761478 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772076356 Chr.19: 41761502-41761502 on Build GRCh38
rs559254485 Chr.19: 41761376-41761376 on Build GRCh38
rs782232827 Chr.19: 41761531-41761532 on Build GRCh38
rs139700050 Chr.19: 41761311-41761311 on Build GRCh38
rs782389028 Chr.19: 41761210-41761210 on Build GRCh38
rs548133416 Chr.19: 41761597-41761597 on Build GRCh38
rs782614417 Chr.19: 41761431-41761431 on Build GRCh38
rs552158385 Chr.19: 41761496-41761496 on Build GRCh38
rs62117368 Chr.19: 41761547-41761547 on Build GRCh38
rs782527923 Chr.19: 41761262-41761262 on Build GRCh38
rs371477019 Chr.19: 41761215-41761215 on Build GRCh38
rs782690797 Chr.19: 41761280-41761280 on Build GRCh38
rs369021656 Chr.19: 41761437-41761437 on Build GRCh38
rs781833457 Chr.19: 41761307-41761307 on Build GRCh38
rs782339128 Chr.19: 41761487-41761487 on Build GRCh38
rs782639699 Chr.19: 41761474-41761474 on Build GRCh38
rs1050104541 Chr.19: 41761605-41761605 on Build GRCh38
rs576685461 Chr.19: 41761298-41761298 on Build GRCh38
rs782038014 Chr.19: 41761292-41761292 on Build GRCh38
rs372425901 Chr.19: 41761238-41761238 on Build GRCh38
rs782483457 Chr.19: 41761249-41761249 on Build GRCh38
rs781930019 Chr.19: 41761488-41761488 on Build GRCh38
rs187864306 Chr.19: 41761497-41761497 on Build GRCh38
rs979580658 Chr.19: 41761448-41761448 on Build GRCh38
rs781985233 Chr.19: 41761212-41761212 on Build GRCh38
rs200497564 Chr.19: 41761412-41761412 on Build GRCh38
rs1005354555 Chr.19: 41761615-41761615 on Build GRCh38
rs781793742 Chr.19: 41761527-41761527 on Build GRCh38
rs576498692 Chr.19: 41761265-41761265 on Build GRCh38
rs199611051 Chr.19: 41761413-41761413 on Build GRCh38
rs367563451 Chr.19: 41761207-41761207 on Build GRCh38
rs781869151 Chr.19: 41761331-41761331 on Build GRCh38
rs372688720 Chr.19: 41761556-41761556 on Build GRCh38
rs782563294 Chr.19: 41761342-41761342 on Build GRCh38
rs530670530 Chr.19: 41761420-41761420 on Build GRCh38
rs781903587 Chr.19: 41761256-41761256 on Build GRCh38
rs782395163 Chr.19: 41761294-41761294 on Build GRCh38
rs782244185 Chr.19: 41761269-41761269 on Build GRCh38
rs1005427634 Chr.19: 41761329-41761329 on Build GRCh38
rs376023129 Chr.19: 41761232-41761232 on Build GRCh38
rs781858359 Chr.19: 41761566-41761566 on Build GRCh38
rs78338897 Chr.19: 41761645-41761645 on Build GRCh38
rs782483251 Chr.19: 41761577-41761577 on Build GRCh38
rs781897263 Chr.19: 41761348-41761348 on Build GRCh38
rs782702223 Chr.19: 41761378-41761378 on Build GRCh38
rs889710658 Chr.19: 41761609-41761609 on Build GRCh38
rs782511112 Chr.19: 41761334-41761334 on Build GRCh38
rs536472575 Chr.19: 41761234-41761234 on Build GRCh38
rs781784609 Chr.19: 41761408-41761408 on Build GRCh38
rs782730368 Chr.19: 41761510-41761510 on Build GRCh38
rs782531554 Chr.19: 41761557-41761557 on Build GRCh38
rs782709435 Chr.19: 41761407-41761407 on Build GRCh38
rs782047725 Chr.19: 41761491-41761491 on Build GRCh38
rs781845947 Chr.19: 41761248-41761248 on Build GRCh38
rs782517580 Chr.19: 41761424-41761424 on Build GRCh38
rs537042421 Chr.19: 41761289-41761289 on Build GRCh38
rs782682266 Chr.19: 41761204-41761204 on Build GRCh38
rs782552580 Chr.19: 41761542-41761542 on Build GRCh38
rs369556770 Chr.19: 41761354-41761354 on Build GRCh38
rs782465758 Chr.19: 41761201-41761201 on Build GRCh38
rs143709500 Chr.19: 41761449-41761449 on Build GRCh38
rs782742243 Chr.19: 41761335-41761335 on Build GRCh38
rs782416001 Chr.19: 41761492-41761492 on Build GRCh38
rs782484195 Chr.19: 41761273-41761273 on Build GRCh38
rs782139826 Chr.19: 41761239-41761239 on Build GRCh38
rs781826090 Chr.19: 41761436-41761436 on Build GRCh38
rs782332402 Chr.19: 41761226-41761226 on Build GRCh38
rs782048041 Chr.19: 41761314-41761314 on Build GRCh38
rs200014119 Chr.19: 41761328-41761328 on Build GRCh38
rs782451361 Chr.19: 41761336-41761336 on Build GRCh38
rs201269236 Chr.19: 41761495-41761495 on Build GRCh38
rs369397467 Chr.19: 41761236-41761236 on Build GRCh38
rs782220668 Chr.19: 41761486-41761486 on Build GRCh38
rs563388006 Chr.19: 41761417-41761417 on Build GRCh38
rs782085834 Chr.19: 41761509-41761509 on Build GRCh38
rs373281695 Chr.19: 41761359-41761359 on Build GRCh38
rs782127498 Chr.19: 41761379-41761379 on Build GRCh38
rs143159805 Chr.19: 41761293-41761293 on Build GRCh38
rs782769519 Chr.19: 41761243-41761243 on Build GRCh38
rs147402597 Chr.19: 41761299-41761299 on Build GRCh38
rs782296053 Chr.19: 41761465-41761465 on Build GRCh38
rs572052717 Chr.19: 41761601-41761601 on Build GRCh38
rs150279040 Chr.19: 41761250-41761250 on Build GRCh38
rs201070424 Chr.19: 41761353-41761353 on Build GRCh38
rs376476934 Chr.19: 41761403-41761403 on Build GRCh38
rs781933343 Chr.19: 41761382-41761382 on Build GRCh38
rs782594731 Chr.19: 41761288-41761288 on Build GRCh38
rs781844141 Chr.19: 41761501-41761501 on Build GRCh38
rs782389697 Chr.19: 41761473-41761473 on Build GRCh38
rs782366279 Chr.19: 41761381-41761381 on Build GRCh38
rs782201805 Chr.19: 41761373-41761373 on Build GRCh38
rs1040486616 Chr.19: 41761524-41761524 on Build GRCh38
rs144169216 Chr.19: 41761498-41761498 on Build GRCh38
rs191699288 Chr.19: 41761633-41761633 on Build GRCh38
rs373480488 Chr.19: 41761399-41761399 on Build GRCh38
rs782607578 Chr.19: 41761369-41761369 on Build GRCh38

View Full Product Details