Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41847335-41847827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTC2
Entrez Gene ID
63946
Gene Name
DMRT like family C2
Gene Aliases
-
UniGene
Hs.350507
Gene Chromosome Location
Chr.19: 41844743-41852330 on Build GRCh38
Transcripts
NM_001040283.2 XM_011527214.1 XM_011527215.1 XM_011527216.2 XM_017027123.1 XM_017027124.1 XM_017027125.1 XM_017027126.1 XM_017027127.1 XM_017027128.1 XM_017027129.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4855042 c.198C>T p.C66C Chr.19: 41847626-41847626 on Build GRCh38
6557941 c.274G>T p.E92* Chr.19: 41847785-41847785 on Build GRCh38
4828862 c.31C>T p.H11Y Chr.19: 41847459-41847459 on Build GRCh38
5566496 c.175C>T p.R59C Chr.19: 41847603-41847603 on Build GRCh38
6539188 c.162C>G p.L54L Chr.19: 41847590-41847590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782473934 Chr.19: 41847450-41847450 on Build GRCh38
rs571848350 Chr.19: 41847449-41847449 on Build GRCh38
rs201970115 Chr.19: 41847511-41847511 on Build GRCh38
rs782769934 Chr.19: 41847448-41847448 on Build GRCh38
rs782622667 Chr.19: 41847460-41847460 on Build GRCh38
rs533708576 Chr.19: 41847789-41847789 on Build GRCh38
rs1000189887 Chr.19: 41847796-41847796 on Build GRCh38
rs371245351 Chr.19: 41847388-41847388 on Build GRCh38
rs150751049 Chr.19: 41847565-41847565 on Build GRCh38
rs782737068 Chr.19: 41847571-41847571 on Build GRCh38
rs782716976 Chr.19: 41847582-41847582 on Build GRCh38
rs782449430 Chr.19: 41847647-41847647 on Build GRCh38
rs539184228 Chr.19: 41847478-41847478 on Build GRCh38
rs782711278 Chr.19: 41847771-41847771 on Build GRCh38
rs781834567 Chr.19: 41847763-41847763 on Build GRCh38
rs781868878 Chr.19: 41847604-41847604 on Build GRCh38
rs782056763 Chr.19: 41847581-41847581 on Build GRCh38
rs782804371 Chr.19: 41847739-41847739 on Build GRCh38
rs543455872 Chr.19: 41847655-41847655 on Build GRCh38
rs987224037 Chr.19: 41847368-41847368 on Build GRCh38
rs201763027 Chr.19: 41847554-41847554 on Build GRCh38
rs782794659 Chr.19: 41847697-41847698 on Build GRCh38
rs782213752 Chr.19: 41847529-41847529 on Build GRCh38
rs781937831 Chr.19: 41847732-41847732 on Build GRCh38
rs781830059 Chr.19: 41847774-41847774 on Build GRCh38
rs782504508 Chr.19: 41847503-41847503 on Build GRCh38
rs751438055 Chr.19: 41847727-41847727 on Build GRCh38
rs782133613 Chr.19: 41847686-41847686 on Build GRCh38
rs375134520 Chr.19: 41847627-41847627 on Build GRCh38
rs373577927 Chr.19: 41847638-41847638 on Build GRCh38
rs781984329 Chr.19: 41847741-41847741 on Build GRCh38
rs782394832 Chr.19: 41847559-41847559 on Build GRCh38
rs554112253 Chr.19: 41847482-41847482 on Build GRCh38
rs782388845 Chr.19: 41847387-41847387 on Build GRCh38
rs569349371 Chr.19: 41847389-41847389 on Build GRCh38
rs781879998 Chr.19: 41847772-41847772 on Build GRCh38
rs782399912 Chr.19: 41847687-41847687 on Build GRCh38
rs782408451 Chr.19: 41847806-41847806 on Build GRCh38
rs782484476 Chr.19: 41847784-41847784 on Build GRCh38
rs782562491 Chr.19: 41847598-41847598 on Build GRCh38
rs781909314 Chr.19: 41847710-41847710 on Build GRCh38
rs369030903 Chr.19: 41847626-41847626 on Build GRCh38
rs929537731 Chr.19: 41847548-41847548 on Build GRCh38
rs781914367 Chr.19: 41847537-41847537 on Build GRCh38
rs782126126 Chr.19: 41847748-41847748 on Build GRCh38
rs145938783 Chr.19: 41847428-41847428 on Build GRCh38
rs782674944 Chr.19: 41847496-41847496 on Build GRCh38
rs782547299 Chr.19: 41847773-41847773 on Build GRCh38
rs782357738 Chr.19: 41847532-41847532 on Build GRCh38
rs782313602 Chr.19: 41847712-41847712 on Build GRCh38
rs200294003 Chr.19: 41847790-41847790 on Build GRCh38
rs782141711 Chr.19: 41847566-41847566 on Build GRCh38
rs199625418 Chr.19: 41847603-41847603 on Build GRCh38
rs782698986 Chr.19: 41847403-41847403 on Build GRCh38
rs781971874 Chr.19: 41847578-41847578 on Build GRCh38
rs200605160 Chr.19: 41847420-41847420 on Build GRCh38
rs187163908 Chr.19: 41847722-41847722 on Build GRCh38
rs920271297 Chr.19: 41847533-41847533 on Build GRCh38
rs782740687 Chr.19: 41847749-41847749 on Build GRCh38
rs782248520 Chr.19: 41847662-41847662 on Build GRCh38
rs139845709 Chr.19: 41847584-41847584 on Build GRCh38
rs782434394 Chr.19: 41847769-41847769 on Build GRCh38
rs781989007 Chr.19: 41847483-41847483 on Build GRCh38
rs554987329 Chr.19: 41847621-41847621 on Build GRCh38
rs905985897 Chr.19: 41847706-41847706 on Build GRCh38
rs782151810 Chr.19: 41847733-41847733 on Build GRCh38

View Full Product Details