Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41851703-41852243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTC2
Entrez Gene ID
63946
Gene Name
DMRT like family C2
Gene Aliases
-
UniGene
Hs.350507
Gene Chromosome Location
Chr.19: 41844743-41852330 on Build GRCh38
Transcripts
NM_001040283.2 XM_011527214.1 XM_011527215.1 XM_017027123.1 XM_017027124.1 XM_017027125.1 XM_017027126.1 XM_017027127.1 XM_017027128.1 XM_017027129.1

SNP Detail

SNP ID Chromosome Location
rs551623821 Chr.19: 41851889-41851889 on Build GRCh38
rs533842344 Chr.19: 41851818-41851819 on Build GRCh38
rs561837589 Chr.19: 41851747-41851747 on Build GRCh38
rs968618829 Chr.19: 41852030-41852030 on Build GRCh38
rs551685272 Chr.19: 41851822-41851823 on Build GRCh38
rs782782962 Chr.19: 41851736-41851736 on Build GRCh38
rs190902622 Chr.19: 41851741-41851741 on Build GRCh38
rs772421846 Chr.19: 41852023-41852023 on Build GRCh38
rs566773043 Chr.19: 41851932-41851932 on Build GRCh38
rs537974852 Chr.19: 41852062-41852062 on Build GRCh38
rs953408182 Chr.19: 41852116-41852116 on Build GRCh38
rs782123177 Chr.19: 41851801-41851801 on Build GRCh38
rs77891616 Chr.19: 41851846-41851846 on Build GRCh38
rs781820676 Chr.19: 41852080-41852080 on Build GRCh38
rs370552271 Chr.19: 41851913-41851913 on Build GRCh38
rs562934494 Chr.19: 41851812-41851812 on Build GRCh38
rs956750648 Chr.19: 41852072-41852072 on Build GRCh38
rs782155737 Chr.19: 41851735-41851735 on Build GRCh38
rs976156670 Chr.19: 41852092-41852092 on Build GRCh38
rs896164868 Chr.19: 41852005-41852005 on Build GRCh38
rs782344057 Chr.19: 41851828-41851828 on Build GRCh38
rs1012488961 Chr.19: 41852006-41852006 on Build GRCh38
rs985765546 Chr.19: 41852207-41852207 on Build GRCh38
rs1056117428 Chr.19: 41852000-41852000 on Build GRCh38
rs933887577 Chr.19: 41851793-41851793 on Build GRCh38
rs147791847 Chr.19: 41851816-41851823 on Build GRCh38
rs183975552 Chr.19: 41851798-41851798 on Build GRCh38
rs147929586 Chr.19: 41852054-41852054 on Build GRCh38
rs112716915 Chr.19: 41851938-41851938 on Build GRCh38
rs574880301 Chr.19: 41851817-41851818 on Build GRCh38
rs533932430 Chr.19: 41852020-41852020 on Build GRCh38
rs911182579 Chr.19: 41852217-41852217 on Build GRCh38
rs782008570 Chr.19: 41851733-41851733 on Build GRCh38
rs976022828 Chr.19: 41851753-41851753 on Build GRCh38
rs924088450 Chr.19: 41851731-41851731 on Build GRCh38
rs782318596 Chr.19: 41851732-41851732 on Build GRCh38
rs3826704 Chr.19: 41852033-41852033 on Build GRCh38
rs529394250 Chr.19: 41851773-41851773 on Build GRCh38
rs371124095 Chr.19: 41851818-41851818 on Build GRCh38
rs782764514 Chr.19: 41852058-41852058 on Build GRCh38
rs782716183 Chr.19: 41852004-41852004 on Build GRCh38
rs1030260553 Chr.19: 41852113-41852113 on Build GRCh38
rs111402662 Chr.19: 41852094-41852094 on Build GRCh38
rs549999165 Chr.19: 41851868-41851868 on Build GRCh38

View Full Product Details