Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41966441-41966969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A3
Entrez Gene ID
478
Gene Name
ATPase Na+/K+ transporting subunit alpha 3
Gene Aliases
AHC2, ATP1A1, CAPOS, DYT12, RDP
UniGene
Hs.515427
Gene Chromosome Location
Chr.19: 41966582-41994276 on Build GRCh38
Transcripts
NM_001256213.1 NM_001256214.1 NM_152296.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4541752 c.3024G>A p.E1008E Chr.19: 41966955-41966955 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370102288 Chr.19: 41966520-41966520 on Build GRCh38
rs782304908 Chr.19: 41966893-41966893 on Build GRCh38
rs781980860 Chr.19: 41966639-41966639 on Build GRCh38
rs782285482 Chr.19: 41966944-41966944 on Build GRCh38
rs573172380 Chr.19: 41966788-41966788 on Build GRCh38
rs782668761 Chr.19: 41966852-41966852 on Build GRCh38
rs915232441 Chr.19: 41966650-41966650 on Build GRCh38
rs546998241 Chr.19: 41966514-41966514 on Build GRCh38
rs571857150 Chr.19: 41966622-41966622 on Build GRCh38
rs568616491 Chr.19: 41966534-41966534 on Build GRCh38
rs782594272 Chr.19: 41966923-41966923 on Build GRCh38
rs970844234 Chr.19: 41966600-41966600 on Build GRCh38
rs1026634929 Chr.19: 41966542-41966542 on Build GRCh38
rs34627136 Chr.19: 41966912-41966913 on Build GRCh38
rs1048572454 Chr.19: 41966841-41966841 on Build GRCh38
rs1016217926 Chr.19: 41966590-41966590 on Build GRCh38
rs781872675 Chr.19: 41966551-41966551 on Build GRCh38
rs782239785 Chr.19: 41966810-41966810 on Build GRCh38
rs546192339 Chr.19: 41966638-41966638 on Build GRCh38
rs782573407 Chr.19: 41966804-41966805 on Build GRCh38
rs782135411 Chr.19: 41966896-41966896 on Build GRCh38
rs1055731723 Chr.19: 41966493-41966493 on Build GRCh38
rs1015811125 Chr.19: 41966868-41966868 on Build GRCh38
rs782205141 Chr.19: 41966564-41966564 on Build GRCh38
rs781965317 Chr.19: 41966895-41966895 on Build GRCh38
rs539551681 Chr.19: 41966655-41966655 on Build GRCh38
rs370433301 Chr.19: 41966885-41966885 on Build GRCh38
rs886054472 Chr.19: 41966657-41966657 on Build GRCh38
rs180885057 Chr.19: 41966831-41966831 on Build GRCh38
rs772022012 Chr.19: 41966678-41966678 on Build GRCh38
rs199854166 Chr.19: 41966701-41966701 on Build GRCh38
rs565195548 Chr.19: 41966690-41966690 on Build GRCh38
rs782677695 Chr.19: 41966739-41966739 on Build GRCh38
rs947305379 Chr.19: 41966502-41966502 on Build GRCh38
rs372647884 Chr.19: 41966783-41966783 on Build GRCh38
rs553335568 Chr.19: 41966596-41966596 on Build GRCh38
rs782747281 Chr.19: 41966687-41966687 on Build GRCh38
rs782512492 Chr.19: 41966929-41966929 on Build GRCh38
rs1004462525 Chr.19: 41966579-41966579 on Build GRCh38
rs776731215 Chr.19: 41966884-41966884 on Build GRCh38
rs186061886 Chr.19: 41966865-41966865 on Build GRCh38
rs781878302 Chr.19: 41966924-41966924 on Build GRCh38
rs903008222 Chr.19: 41966512-41966512 on Build GRCh38
rs782433771 Chr.19: 41966677-41966677 on Build GRCh38
rs781981545 Chr.19: 41966633-41966633 on Build GRCh38
rs368592213 Chr.19: 41966478-41966478 on Build GRCh38
rs374234511 Chr.19: 41966934-41966934 on Build GRCh38
rs996446142 Chr.19: 41966541-41966541 on Build GRCh38
rs782076542 Chr.19: 41966635-41966635 on Build GRCh38
rs1041970570 Chr.19: 41966508-41966508 on Build GRCh38
rs1007934902 Chr.19: 41966513-41966513 on Build GRCh38
rs782765609 Chr.19: 41966897-41966897 on Build GRCh38
rs397515382 Chr.19: 41966938-41966939 on Build GRCh38
rs782140294 Chr.19: 41966904-41966904 on Build GRCh38
rs375738720 Chr.19: 41966510-41966510 on Build GRCh38
rs899617373 Chr.19: 41966709-41966709 on Build GRCh38
rs899522430 Chr.19: 41966535-41966535 on Build GRCh38
rs535688266 Chr.19: 41966550-41966550 on Build GRCh38
rs782491617 Chr.19: 41966922-41966922 on Build GRCh38
rs587778772 Chr.19: 41966738-41966739 on Build GRCh38
rs782566365 Chr.19: 41966638-41966639 on Build GRCh38
rs937959112 Chr.19: 41966483-41966483 on Build GRCh38
rs782063577 Chr.19: 41966891-41966891 on Build GRCh38
rs368003751 Chr.19: 41966918-41966918 on Build GRCh38
rs149277536 Chr.19: 41966641-41966641 on Build GRCh38
rs535287905 Chr.19: 41966501-41966501 on Build GRCh38
rs782804940 Chr.19: 41966653-41966653 on Build GRCh38
rs781891453 Chr.19: 41966781-41966781 on Build GRCh38

View Full Product Details