Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43506288-43506832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETHE1
Entrez Gene ID
23474
Gene Name
ETHE1, persulfide dioxygenase
Gene Aliases
HSCO, YF13H12
UniGene
Hs.7486
Gene Chromosome Location
Chr.19: 43506719-43527256 on Build GRCh38
Transcripts
NM_001320867.1 NM_001320868.1 NM_001320869.1 NM_014297.4 XM_005258687.3

SNP Detail

SNP ID Chromosome Location
rs1028944335 Chr.19: 43506367-43506367 on Build GRCh38
rs769741765 Chr.19: 43506722-43506722 on Build GRCh38
rs547341456 Chr.19: 43506712-43506712 on Build GRCh38
rs916748051 Chr.19: 43506733-43506733 on Build GRCh38
rs554983564 Chr.19: 43506318-43506318 on Build GRCh38
rs1037064912 Chr.19: 43506505-43506505 on Build GRCh38
rs563865883 Chr.19: 43506631-43506631 on Build GRCh38
rs991018000 Chr.19: 43506524-43506524 on Build GRCh38
rs996070381 Chr.19: 43506364-43506364 on Build GRCh38
rs928114067 Chr.19: 43506469-43506469 on Build GRCh38
rs1048565396 Chr.19: 43506693-43506693 on Build GRCh38
rs201842186 Chr.19: 43506795-43506795 on Build GRCh38
rs906457057 Chr.19: 43506506-43506506 on Build GRCh38
rs544042359 Chr.19: 43506448-43506448 on Build GRCh38
rs939716292 Chr.19: 43506787-43506787 on Build GRCh38
rs939546688 Chr.19: 43506474-43506474 on Build GRCh38
rs763755684 Chr.19: 43506546-43506546 on Build GRCh38
rs909634666 Chr.19: 43506707-43506707 on Build GRCh38
rs182986403 Chr.19: 43506308-43506308 on Build GRCh38
rs187615470 Chr.19: 43506319-43506319 on Build GRCh38
rs958852589 Chr.19: 43506313-43506313 on Build GRCh38
rs888143031 Chr.19: 43506459-43506459 on Build GRCh38
rs1011500488 Chr.19: 43506670-43506670 on Build GRCh38
rs966756194 Chr.19: 43506686-43506686 on Build GRCh38
rs921426514 Chr.19: 43506405-43506405 on Build GRCh38
rs775680830 Chr.19: 43506776-43506776 on Build GRCh38
rs751373825 Chr.19: 43506671-43506671 on Build GRCh38
rs527305410 Chr.19: 43506692-43506692 on Build GRCh38
rs752936698 Chr.19: 43506354-43506354 on Build GRCh38
rs1041227303 Chr.19: 43506393-43506393 on Build GRCh38
rs968748827 Chr.19: 43506629-43506629 on Build GRCh38
rs916862717 Chr.19: 43506570-43506570 on Build GRCh38
rs932429207 Chr.19: 43506430-43506430 on Build GRCh38
rs777509773 Chr.19: 43506802-43506802 on Build GRCh38
rs977408670 Chr.19: 43506661-43506661 on Build GRCh38
rs930854445 Chr.19: 43506758-43506758 on Build GRCh38
rs967165934 Chr.19: 43506455-43506455 on Build GRCh38
rs960851778 Chr.19: 43506723-43506723 on Build GRCh38
rs924909187 Chr.19: 43506357-43506357 on Build GRCh38
rs1049163601 Chr.19: 43506585-43506585 on Build GRCh38
rs1003895562 Chr.19: 43506461-43506461 on Build GRCh38
rs954020656 Chr.19: 43506373-43506373 on Build GRCh38
rs765481129 Chr.19: 43506324-43506324 on Build GRCh38
rs371417357 Chr.19: 43506647-43506647 on Build GRCh38
rs59584199 Chr.19: 43506516-43506516 on Build GRCh38
rs780341837 Chr.19: 43506801-43506802 on Build GRCh38
rs545104246 Chr.19: 43506677-43506677 on Build GRCh38
rs991528564 Chr.19: 43506316-43506316 on Build GRCh38
rs564931279 Chr.19: 43506689-43506689 on Build GRCh38
rs1003862298 Chr.19: 43506512-43506512 on Build GRCh38
rs1036732356 Chr.19: 43506793-43506793 on Build GRCh38
rs989926984 Chr.19: 43506394-43506394 on Build GRCh38
rs1012980752 Chr.19: 43506541-43506541 on Build GRCh38
rs1028913069 Chr.19: 43506665-43506665 on Build GRCh38
rs930321611 Chr.19: 43506576-43506576 on Build GRCh38
rs959178023 Chr.19: 43506533-43506533 on Build GRCh38
rs557741336 Chr.19: 43506726-43506727 on Build GRCh38
rs977453321 Chr.19: 43506630-43506630 on Build GRCh38
rs915248691 Chr.19: 43506414-43506414 on Build GRCh38
rs763284936 Chr.19: 43506539-43506539 on Build GRCh38
rs547312075 Chr.19: 43506590-43506591 on Build GRCh38
rs944249546 Chr.19: 43506372-43506372 on Build GRCh38
rs893019769 Chr.19: 43506666-43506666 on Build GRCh38
rs1021676203 Chr.19: 43506547-43506547 on Build GRCh38
rs759612379 Chr.19: 43506638-43506638 on Build GRCh38
rs940406229 Chr.19: 43506598-43506598 on Build GRCh38
rs886113769 Chr.19: 43506591-43506591 on Build GRCh38
rs529990989 Chr.19: 43506800-43506800 on Build GRCh38
rs947375668 Chr.19: 43506338-43506338 on Build GRCh38
rs921466143 Chr.19: 43506663-43506663 on Build GRCh38

View Full Product Details