Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44708109-44708619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM16
Entrez Gene ID
388551
Gene Name
carcinoembryonic antigen related cell adhesion molecule 16
Gene Aliases
CEAL2, DFNA4B
UniGene
Hs.456381
Gene Chromosome Location
Chr.19: 44699151-44710714 on Build GRCh38
Transcripts
NM_001039213.3 XM_017026795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6225437 c.1422G>T p.Q474H Chr.19: 44708147-44708147 on Build GRCh38
1394492 c.1453C>A p.Q485K Chr.19: 44708178-44708178 on Build GRCh38
3535733 c.1462C>T p.P488S Chr.19: 44708187-44708187 on Build GRCh38
397020 c.1459G>A p.A487T Chr.19: 44708184-44708184 on Build GRCh38
4888601 c.1386C>A p.D462E Chr.19: 44708111-44708111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759768646 Chr.19: 44708199-44708199 on Build GRCh38
rs908301068 Chr.19: 44708172-44708172 on Build GRCh38
rs1022038251 Chr.19: 44708517-44708517 on Build GRCh38
rs948887638 Chr.19: 44708239-44708239 on Build GRCh38
rs1023249239 Chr.19: 44708592-44708592 on Build GRCh38
rs561516925 Chr.19: 44708444-44708444 on Build GRCh38
rs759951345 Chr.19: 44708409-44708409 on Build GRCh38
rs973815150 Chr.19: 44708186-44708186 on Build GRCh38
rs62120569 Chr.19: 44708574-44708574 on Build GRCh38
rs902512546 Chr.19: 44708257-44708257 on Build GRCh38
rs747041563 Chr.19: 44708181-44708189 on Build GRCh38
rs577088481 Chr.19: 44708307-44708307 on Build GRCh38
rs900683485 Chr.19: 44708289-44708289 on Build GRCh38
rs572555812 Chr.19: 44708357-44708357 on Build GRCh38
rs767060571 Chr.19: 44708283-44708283 on Build GRCh38
rs937846445 Chr.19: 44708210-44708210 on Build GRCh38
rs753279949 Chr.19: 44708567-44708567 on Build GRCh38
rs545556026 Chr.19: 44708322-44708322 on Build GRCh38
rs1055463913 Chr.19: 44708211-44708211 on Build GRCh38
rs182111313 Chr.19: 44708213-44708213 on Build GRCh38
rs11880816 Chr.19: 44708240-44708240 on Build GRCh38
rs552756707 Chr.19: 44708336-44708336 on Build GRCh38
rs901181602 Chr.19: 44708335-44708335 on Build GRCh38
rs61744497 Chr.19: 44708135-44708135 on Build GRCh38
rs1049652198 Chr.19: 44708391-44708391 on Build GRCh38
rs748518854 Chr.19: 44708145-44708145 on Build GRCh38
rs542168859 Chr.19: 44708333-44708333 on Build GRCh38
rs374368349 Chr.19: 44708337-44708337 on Build GRCh38
rs370134847 Chr.19: 44708264-44708264 on Build GRCh38
rs866353392 Chr.19: 44708583-44708583 on Build GRCh38
rs766512544 Chr.19: 44708188-44708188 on Build GRCh38
rs377372333 Chr.19: 44708141-44708141 on Build GRCh38
rs753704465 Chr.19: 44708207-44708207 on Build GRCh38
rs199597608 Chr.19: 44708169-44708169 on Build GRCh38
rs774230396 Chr.19: 44708195-44708195 on Build GRCh38
rs939786744 Chr.19: 44708185-44708185 on Build GRCh38
rs771457333 Chr.19: 44708215-44708216 on Build GRCh38
rs765896356 Chr.19: 44708410-44708410 on Build GRCh38
rs996336356 Chr.19: 44708300-44708300 on Build GRCh38
rs374925634 Chr.19: 44708206-44708206 on Build GRCh38
rs902302585 Chr.19: 44708519-44708519 on Build GRCh38
rs866661251 Chr.19: 44708515-44708515 on Build GRCh38
rs984102367 Chr.19: 44708484-44708484 on Build GRCh38
rs763317239 Chr.19: 44708187-44708187 on Build GRCh38
rs150420308 Chr.19: 44708585-44708585 on Build GRCh38
rs777876987 Chr.19: 44708134-44708135 on Build GRCh38
rs555476199 Chr.19: 44708204-44708204 on Build GRCh38
rs1015306729 Chr.19: 44708509-44708509 on Build GRCh38
rs999286917 Chr.19: 44708573-44708573 on Build GRCh38
rs916054646 Chr.19: 44708224-44708224 on Build GRCh38
rs1032378629 Chr.19: 44708269-44708269 on Build GRCh38
rs541570508 Chr.19: 44708369-44708369 on Build GRCh38
rs746661190 Chr.19: 44708136-44708136 on Build GRCh38
rs896377955 Chr.19: 44708212-44708212 on Build GRCh38

View Full Product Details