Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44987094-44987592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLPTM1
Entrez Gene ID
1209
Gene Name
CLPTM1, transmembrane protein
Gene Aliases
-
UniGene
Hs.444441
Gene Chromosome Location
Chr.19: 44954585-44993346 on Build GRCh38
Transcripts
NM_001282175.1 NM_001282176.1 NM_001294.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5973005 c.811G>A p.V271M Chr.19: 44987196-44987196 on Build GRCh38
998107 c.984C>T p.N328N Chr.19: 44987369-44987369 on Build GRCh38
5680077 c.894delG p.A299fs*47 Chr.19: 44987279-44987279 on Build GRCh38
6630615 c.940C>T p.R314C Chr.19: 44987325-44987325 on Build GRCh38
4672896 c.810C>T p.A270A Chr.19: 44987195-44987195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372588500 Chr.19: 44987296-44987296 on Build GRCh38
rs939017878 Chr.19: 44987327-44987327 on Build GRCh38
rs761964588 Chr.19: 44987141-44987141 on Build GRCh38
rs781566274 Chr.19: 44987497-44987497 on Build GRCh38
rs752574567 Chr.19: 44987148-44987148 on Build GRCh38
rs760587810 Chr.19: 44987375-44987375 on Build GRCh38
rs576754517 Chr.19: 44987282-44987282 on Build GRCh38
rs1004104177 Chr.19: 44987480-44987480 on Build GRCh38
rs573473703 Chr.19: 44987360-44987360 on Build GRCh38
rs376974203 Chr.19: 44987144-44987144 on Build GRCh38
rs747269591 Chr.19: 44987318-44987318 on Build GRCh38
rs537352065 Chr.19: 44987506-44987506 on Build GRCh38
rs780552418 Chr.19: 44987316-44987316 on Build GRCh38
rs769057466 Chr.19: 44987326-44987326 on Build GRCh38
rs376613031 Chr.19: 44987298-44987298 on Build GRCh38
rs142500092 Chr.19: 44987189-44987189 on Build GRCh38
rs371580610 Chr.19: 44987390-44987390 on Build GRCh38
rs755484031 Chr.19: 44987409-44987409 on Build GRCh38
rs201718309 Chr.19: 44987454-44987454 on Build GRCh38
rs745412208 Chr.19: 44987303-44987303 on Build GRCh38
rs773471647 Chr.19: 44987280-44987280 on Build GRCh38
rs990612086 Chr.19: 44987528-44987528 on Build GRCh38
rs768579180 Chr.19: 44987564-44987564 on Build GRCh38
rs777498031 Chr.19: 44987169-44987169 on Build GRCh38
rs779011527 Chr.19: 44987536-44987536 on Build GRCh38
rs541902572 Chr.19: 44987477-44987477 on Build GRCh38
rs763142444 Chr.19: 44987363-44987363 on Build GRCh38
rs375387209 Chr.19: 44987391-44987391 on Build GRCh38
rs770087065 Chr.19: 44987431-44987431 on Build GRCh38
rs150554721 Chr.19: 44987444-44987444 on Build GRCh38
rs753515972 Chr.19: 44987297-44987297 on Build GRCh38
rs776437723 Chr.19: 44987446-44987446 on Build GRCh38
rs775565145 Chr.19: 44987372-44987372 on Build GRCh38
rs933676304 Chr.19: 44987457-44987457 on Build GRCh38
rs762003789 Chr.19: 44987260-44987260 on Build GRCh38
rs781106900 Chr.19: 44987147-44987147 on Build GRCh38
rs756893559 Chr.19: 44987379-44987379 on Build GRCh38
rs199925354 Chr.19: 44987271-44987271 on Build GRCh38
rs745954999 Chr.19: 44987562-44987562 on Build GRCh38
rs776913389 Chr.19: 44987351-44987351 on Build GRCh38
rs754893409 Chr.19: 44987403-44987405 on Build GRCh38
rs1031698858 Chr.19: 44987501-44987501 on Build GRCh38
rs768702439 Chr.19: 44987226-44987226 on Build GRCh38
rs757907129 Chr.19: 44987389-44987389 on Build GRCh38
rs543748772 Chr.19: 44987432-44987432 on Build GRCh38
rs752486866 Chr.19: 44987291-44987291 on Build GRCh38
rs866054495 Chr.19: 44987493-44987493 on Build GRCh38
rs762648202 Chr.19: 44987466-44987466 on Build GRCh38
rs899497370 Chr.19: 44987193-44987193 on Build GRCh38
rs748376529 Chr.19: 44987352-44987352 on Build GRCh38
rs553445762 Chr.19: 44987210-44987210 on Build GRCh38
rs749877186 Chr.19: 44987388-44987388 on Build GRCh38
rs774622430 Chr.19: 44987441-44987441 on Build GRCh38
rs866202546 Chr.19: 44987453-44987453 on Build GRCh38
rs150943861 Chr.19: 44987220-44987220 on Build GRCh38
rs374073636 Chr.19: 44987145-44987145 on Build GRCh38
rs755945704 Chr.19: 44987295-44987295 on Build GRCh38
rs531152315 Chr.19: 44987129-44987129 on Build GRCh38
rs371697771 Chr.19: 44987124-44987124 on Build GRCh38
rs751536373 Chr.19: 44987143-44987143 on Build GRCh38
rs748487644 Chr.19: 44987423-44987423 on Build GRCh38
rs1017540543 Chr.19: 44987152-44987152 on Build GRCh38
rs750071785 Chr.19: 44987456-44987456 on Build GRCh38
rs530379882 Chr.19: 44987518-44987518 on Build GRCh38
rs756546670 Chr.19: 44987505-44987505 on Build GRCh38
rs771121772 Chr.19: 44987436-44987436 on Build GRCh38
rs755678028 Chr.19: 44987165-44987165 on Build GRCh38
rs3786505 Chr.19: 44987312-44987312 on Build GRCh38
rs561359383 Chr.19: 44987504-44987504 on Build GRCh38
rs767587118 Chr.19: 44987286-44987286 on Build GRCh38
rs112936176 Chr.19: 44987123-44987123 on Build GRCh38
rs998109234 Chr.19: 44987201-44987201 on Build GRCh38
rs528222489 Chr.19: 44987469-44987469 on Build GRCh38
rs1021721487 Chr.19: 44987549-44987549 on Build GRCh38
rs10422273 Chr.19: 44987435-44987435 on Build GRCh38
rs766159541 Chr.19: 44987142-44987142 on Build GRCh38
rs781585955 Chr.19: 44987418-44987418 on Build GRCh38
rs559477546 Chr.19: 44987455-44987455 on Build GRCh38
rs776912395 Chr.19: 44987253-44987253 on Build GRCh38
rs113421963 Chr.19: 44987178-44987178 on Build GRCh38
rs7257610 Chr.19: 44987170-44987170 on Build GRCh38
rs375159260 Chr.19: 44987202-44987202 on Build GRCh38
rs767499149 Chr.19: 44987368-44987368 on Build GRCh38
rs752168697 Chr.19: 44987499-44987499 on Build GRCh38
rs204468 Chr.19: 44987378-44987378 on Build GRCh38
rs771320019 Chr.19: 44987535-44987535 on Build GRCh38
rs773334767 Chr.19: 44987491-44987492 on Build GRCh38
rs373820261 Chr.19: 44987195-44987195 on Build GRCh38
rs769944481 Chr.19: 44987353-44987353 on Build GRCh38
rs370922483 Chr.19: 44987172-44987172 on Build GRCh38
rs143527825 Chr.19: 44987385-44987385 on Build GRCh38
rs751068046 Chr.19: 44987470-44987470 on Build GRCh38
rs768508773 Chr.19: 44987445-44987445 on Build GRCh38
rs763928897 Chr.19: 44987377-44987377 on Build GRCh38
rs960720845 Chr.19: 44987568-44987568 on Build GRCh38

View Full Product Details