Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45178603-45179110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S3
Entrez Gene ID
388552
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
BLOS3, HPS8, RP
UniGene
Hs.103902
Gene Chromosome Location
Chr.19: 45178745-45202715 on Build GRCh38
Transcripts
NM_212550.4

SNP Detail

SNP ID Chromosome Location
rs116105443 Chr.19: 45178835-45178835 on Build GRCh38
rs565339223 Chr.19: 45178769-45178769 on Build GRCh38
rs1034441358 Chr.19: 45179002-45179002 on Build GRCh38
rs894555347 Chr.19: 45179019-45179019 on Build GRCh38
rs995944949 Chr.19: 45178778-45178778 on Build GRCh38
rs949087707 Chr.19: 45178691-45178691 on Build GRCh38
rs747466522 Chr.19: 45178639-45178639 on Build GRCh38
rs918367347 Chr.19: 45178674-45178674 on Build GRCh38
rs1014463193 Chr.19: 45179035-45179035 on Build GRCh38
rs556491399 Chr.19: 45178712-45178712 on Build GRCh38
rs752562425 Chr.19: 45178707-45178707 on Build GRCh38
rs1052208149 Chr.19: 45178735-45178735 on Build GRCh38
rs1046777995 Chr.19: 45178711-45178711 on Build GRCh38
rs1048148686 Chr.19: 45178682-45178682 on Build GRCh38
rs765084301 Chr.19: 45178626-45178626 on Build GRCh38
rs983162563 Chr.19: 45178850-45178850 on Build GRCh38
rs551606732 Chr.19: 45178857-45178857 on Build GRCh38
rs907519636 Chr.19: 45178858-45178858 on Build GRCh38
rs951692029 Chr.19: 45179057-45179057 on Build GRCh38
rs114784352 Chr.19: 45178793-45178793 on Build GRCh38
rs576806648 Chr.19: 45178766-45178766 on Build GRCh38
rs1041603814 Chr.19: 45178985-45178985 on Build GRCh38
rs573571955 Chr.19: 45178685-45178685 on Build GRCh38
rs889185838 Chr.19: 45179012-45179012 on Build GRCh38
rs1008894702 Chr.19: 45178760-45178760 on Build GRCh38
rs59215448 Chr.19: 45178770-45178770 on Build GRCh38
rs138914315 Chr.19: 45178999-45178999 on Build GRCh38
rs549913562 Chr.19: 45178937-45178937 on Build GRCh38
rs547211242 Chr.19: 45178992-45178992 on Build GRCh38
rs755507173 Chr.19: 45179043-45179043 on Build GRCh38
rs973147347 Chr.19: 45178870-45178870 on Build GRCh38
rs983879015 Chr.19: 45178785-45178785 on Build GRCh38
rs935263388 Chr.19: 45178807-45178807 on Build GRCh38
rs543407501 Chr.19: 45178799-45178799 on Build GRCh38
rs1010965402 Chr.19: 45178739-45178739 on Build GRCh38
rs571377637 Chr.19: 45178878-45178878 on Build GRCh38
rs562480723 Chr.19: 45178800-45178800 on Build GRCh38
rs563933221 Chr.19: 45178981-45178981 on Build GRCh38
rs904540992 Chr.19: 45178994-45178994 on Build GRCh38
rs1052027635 Chr.19: 45178897-45178897 on Build GRCh38
rs536301059 Chr.19: 45179028-45179028 on Build GRCh38
rs893614191 Chr.19: 45178737-45178737 on Build GRCh38
rs8104024 Chr.19: 45178768-45178768 on Build GRCh38
rs536238393 Chr.19: 45178706-45178706 on Build GRCh38
rs367571876 Chr.19: 45178958-45178958 on Build GRCh38
rs906448663 Chr.19: 45178720-45178720 on Build GRCh38
rs780764556 Chr.19: 45178955-45178955 on Build GRCh38
rs916323775 Chr.19: 45178789-45178789 on Build GRCh38
rs953513879 Chr.19: 45178747-45178747 on Build GRCh38
rs556926381 Chr.19: 45178750-45178750 on Build GRCh38
rs556204314 Chr.19: 45179065-45179065 on Build GRCh38
rs373669358 Chr.19: 45178809-45178809 on Build GRCh38
rs1024593979 Chr.19: 45178827-45178827 on Build GRCh38
rs1018958131 Chr.19: 45179048-45179048 on Build GRCh38
rs931061867 Chr.19: 45178679-45178679 on Build GRCh38
rs1024875710 Chr.19: 45179041-45179041 on Build GRCh38
rs369144610 Chr.19: 45178780-45178780 on Build GRCh38
rs917450666 Chr.19: 45178686-45178686 on Build GRCh38
rs993097512 Chr.19: 45178655-45178655 on Build GRCh38
rs527449999 Chr.19: 45178951-45178951 on Build GRCh38
rs947712607 Chr.19: 45178709-45178709 on Build GRCh38
rs545634026 Chr.19: 45178816-45178816 on Build GRCh38
rs116448245 Chr.19: 45178784-45178784 on Build GRCh38
rs1034410181 Chr.19: 45178802-45178802 on Build GRCh38
rs370457632 Chr.19: 45178730-45178730 on Build GRCh38
rs142628440 Chr.19: 45178649-45178650 on Build GRCh38
rs916233747 Chr.19: 45178684-45178684 on Build GRCh38
rs1029041889 Chr.19: 45178746-45178746 on Build GRCh38
rs931825299 Chr.19: 45178895-45178895 on Build GRCh38

View Full Product Details