Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45180116-45180610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S3
Entrez Gene ID
388552
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
BLOS3, HPS8, RP
UniGene
Hs.103902
Gene Chromosome Location
Chr.19: 45178745-45202715 on Build GRCh38
Transcripts
NM_212550.4

SNP Detail

SNP ID Chromosome Location
rs368024066 Chr.19: 45180418-45180418 on Build GRCh38
rs1002746754 Chr.19: 45180485-45180485 on Build GRCh38
rs1038756617 Chr.19: 45180259-45180259 on Build GRCh38
rs1019731510 Chr.19: 45180527-45180527 on Build GRCh38
rs1026929560 Chr.19: 45180310-45180310 on Build GRCh38
rs537234718 Chr.19: 45180271-45180271 on Build GRCh38
rs924230500 Chr.19: 45180377-45180377 on Build GRCh38
rs34965337 Chr.19: 45180235-45180236 on Build GRCh38
rs927543670 Chr.19: 45180307-45180307 on Build GRCh38
rs970536894 Chr.19: 45180149-45180149 on Build GRCh38
rs889927229 Chr.19: 45180220-45180220 on Build GRCh38
rs115683085 Chr.19: 45180370-45180370 on Build GRCh38
rs371023414 Chr.19: 45180218-45180218 on Build GRCh38
rs938658189 Chr.19: 45180165-45180165 on Build GRCh38
rs76467674 Chr.19: 45180145-45180145 on Build GRCh38
rs1016517016 Chr.19: 45180352-45180352 on Build GRCh38
rs928647238 Chr.19: 45180155-45180155 on Build GRCh38
rs563411988 Chr.19: 45180517-45180517 on Build GRCh38
rs978996897 Chr.19: 45180524-45180524 on Build GRCh38
rs1016375313 Chr.19: 45180146-45180146 on Build GRCh38
rs561150528 Chr.19: 45180488-45180488 on Build GRCh38
rs942852799 Chr.19: 45180536-45180536 on Build GRCh38
rs185088220 Chr.19: 45180392-45180392 on Build GRCh38
rs950565662 Chr.19: 45180140-45180140 on Build GRCh38
rs906938163 Chr.19: 45180426-45180426 on Build GRCh38
rs924094637 Chr.19: 45180546-45180546 on Build GRCh38
rs775607885 Chr.19: 45180410-45180410 on Build GRCh38
rs985580629 Chr.19: 45180330-45180330 on Build GRCh38
rs183067434 Chr.19: 45180542-45180542 on Build GRCh38
rs117001334 Chr.19: 45180294-45180294 on Build GRCh38
rs913469463 Chr.19: 45180191-45180191 on Build GRCh38
rs1002040561 Chr.19: 45180406-45180406 on Build GRCh38
rs1026835663 Chr.19: 45180502-45180502 on Build GRCh38
rs58074751 Chr.19: 45180577-45180577 on Build GRCh38
rs995488894 Chr.19: 45180290-45180290 on Build GRCh38
rs182068302 Chr.19: 45180369-45180369 on Build GRCh38
rs1047698629 Chr.19: 45180315-45180315 on Build GRCh38
rs922996709 Chr.19: 45180557-45180557 on Build GRCh38
rs913520491 Chr.19: 45180500-45180500 on Build GRCh38
rs368390636 Chr.19: 45180207-45180207 on Build GRCh38
rs960055233 Chr.19: 45180449-45180449 on Build GRCh38
rs550362452 Chr.19: 45180348-45180348 on Build GRCh38
rs60498449 Chr.19: 45180259-45180259 on Build GRCh38
rs901521454 Chr.19: 45180292-45180292 on Build GRCh38
rs142584236 Chr.19: 45180331-45180331 on Build GRCh38
rs1045877658 Chr.19: 45180231-45180231 on Build GRCh38
rs543635649 Chr.19: 45180508-45180508 on Build GRCh38
rs762649204 Chr.19: 45180513-45180513 on Build GRCh38
rs541332923 Chr.19: 45180458-45180458 on Build GRCh38
rs961274233 Chr.19: 45180275-45180275 on Build GRCh38
rs1044157395 Chr.19: 45180424-45180424 on Build GRCh38
rs189995520 Chr.19: 45180523-45180523 on Build GRCh38
rs6509200 Chr.19: 45180190-45180190 on Build GRCh38
rs148427902 Chr.19: 45180151-45180151 on Build GRCh38
rs948347510 Chr.19: 45180419-45180419 on Build GRCh38
rs958167071 Chr.19: 45180558-45180558 on Build GRCh38
rs1006090146 Chr.19: 45180341-45180341 on Build GRCh38
rs568334676 Chr.19: 45180225-45180225 on Build GRCh38
rs1003084258 Chr.19: 45180219-45180219 on Build GRCh38
rs189672488 Chr.19: 45180212-45180212 on Build GRCh38
rs747825460 Chr.19: 45180236-45180236 on Build GRCh38
rs574712542 Chr.19: 45180497-45180497 on Build GRCh38
rs1036806146 Chr.19: 45180489-45180489 on Build GRCh38
rs947596880 Chr.19: 45180195-45180195 on Build GRCh38
rs933010449 Chr.19: 45180565-45180565 on Build GRCh38
rs551937969 Chr.19: 45180528-45180528 on Build GRCh38

View Full Product Details