Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45180606-45181105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S3
Entrez Gene ID
388552
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
BLOS3, HPS8, RP
UniGene
Hs.103902
Gene Chromosome Location
Chr.19: 45178745-45202715 on Build GRCh38
Transcripts
NM_212550.4

SNP Detail

SNP ID Chromosome Location
rs141286841 Chr.19: 45181038-45181038 on Build GRCh38
rs116251719 Chr.19: 45180991-45180991 on Build GRCh38
rs1040940662 Chr.19: 45181013-45181013 on Build GRCh38
rs976942512 Chr.19: 45180906-45180906 on Build GRCh38
rs938415860 Chr.19: 45180685-45180685 on Build GRCh38
rs1037596802 Chr.19: 45180760-45180760 on Build GRCh38
rs979819412 Chr.19: 45180873-45180873 on Build GRCh38
rs1010243849 Chr.19: 45180841-45180841 on Build GRCh38
rs1035777652 Chr.19: 45180797-45180797 on Build GRCh38
rs911463692 Chr.19: 45180875-45180875 on Build GRCh38
rs574772380 Chr.19: 45181067-45181067 on Build GRCh38
rs1047492712 Chr.19: 45180969-45180969 on Build GRCh38
rs1010294703 Chr.19: 45181027-45181027 on Build GRCh38
rs964583045 Chr.19: 45180898-45180898 on Build GRCh38
rs150959722 Chr.19: 45180767-45180767 on Build GRCh38
rs59591789 Chr.19: 45180692-45180692 on Build GRCh38
rs889734463 Chr.19: 45181009-45181009 on Build GRCh38
rs1006932406 Chr.19: 45180794-45180794 on Build GRCh38
rs895613848 Chr.19: 45181017-45181017 on Build GRCh38
rs796862928 Chr.19: 45180718-45180718 on Build GRCh38
rs1020245476 Chr.19: 45180850-45180850 on Build GRCh38
rs923047748 Chr.19: 45180908-45180908 on Build GRCh38
rs764004487 Chr.19: 45180809-45180809 on Build GRCh38
rs1020016613 Chr.19: 45180807-45180807 on Build GRCh38
rs999626019 Chr.19: 45180820-45180820 on Build GRCh38
rs535827858 Chr.19: 45180996-45180996 on Build GRCh38
rs901288065 Chr.19: 45180814-45180814 on Build GRCh38
rs554947014 Chr.19: 45181006-45181006 on Build GRCh38
rs931110092 Chr.19: 45180723-45180723 on Build GRCh38
rs1031123394 Chr.19: 45181068-45181068 on Build GRCh38
rs1001358783 Chr.19: 45180770-45180770 on Build GRCh38
rs755013546 Chr.19: 45180967-45180967 on Build GRCh38
rs937206994 Chr.19: 45181005-45181005 on Build GRCh38
rs941694172 Chr.19: 45180980-45180980 on Build GRCh38
rs928302654 Chr.19: 45180900-45180900 on Build GRCh38
rs111903160 Chr.19: 45180851-45180851 on Build GRCh38
rs1038047058 Chr.19: 45180989-45180989 on Build GRCh38
rs371885127 Chr.19: 45180817-45180817 on Build GRCh38
rs999490044 Chr.19: 45181056-45181056 on Build GRCh38
rs188598670 Chr.19: 45180830-45180830 on Build GRCh38
rs550753235 Chr.19: 45180870-45180870 on Build GRCh38
rs979436683 Chr.19: 45180858-45180858 on Build GRCh38
rs1048308164 Chr.19: 45180765-45180765 on Build GRCh38
rs1036711834 Chr.19: 45180697-45180697 on Build GRCh38
rs370426154 Chr.19: 45180954-45180954 on Build GRCh38
rs958197958 Chr.19: 45180849-45180849 on Build GRCh38
rs553156600 Chr.19: 45180974-45180974 on Build GRCh38
rs928426500 Chr.19: 45180654-45180654 on Build GRCh38
rs942601336 Chr.19: 45181011-45181011 on Build GRCh38
rs1020296333 Chr.19: 45181049-45181049 on Build GRCh38
rs781377437 Chr.19: 45181037-45181038 on Build GRCh38
rs570917374 Chr.19: 45180888-45180888 on Build GRCh38
rs937562978 Chr.19: 45180749-45180749 on Build GRCh38
rs886069784 Chr.19: 45180669-45180669 on Build GRCh38

View Full Product Details