Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45181134-45181646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S3
Entrez Gene ID
388552
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 3
Gene Aliases
BLOS3, HPS8, RP
UniGene
Hs.103902
Gene Chromosome Location
Chr.19: 45178745-45202715 on Build GRCh38
Transcripts
NM_212550.4

SNP Detail

SNP ID Chromosome Location
rs941732046 Chr.19: 45181202-45181202 on Build GRCh38
rs973086277 Chr.19: 45181236-45181236 on Build GRCh38
rs114644261 Chr.19: 45181549-45181549 on Build GRCh38
rs879713453 Chr.19: 45181266-45181266 on Build GRCh38
rs922534514 Chr.19: 45181245-45181245 on Build GRCh38
rs758633856 Chr.19: 45181403-45181403 on Build GRCh38
rs375016316 Chr.19: 45181182-45181182 on Build GRCh38
rs935357148 Chr.19: 45181246-45181247 on Build GRCh38
rs904602648 Chr.19: 45181359-45181359 on Build GRCh38
rs755664764 Chr.19: 45181213-45181213 on Build GRCh38
rs765731212 Chr.19: 45181279-45181279 on Build GRCh38
rs554604574 Chr.19: 45181311-45181311 on Build GRCh38
rs952446725 Chr.19: 45181175-45181175 on Build GRCh38
rs911061158 Chr.19: 45181203-45181204 on Build GRCh38
rs747159416 Chr.19: 45181425-45181425 on Build GRCh38
rs1056954878 Chr.19: 45181302-45181302 on Build GRCh38
rs992732880 Chr.19: 45181595-45181595 on Build GRCh38
rs781613896 Chr.19: 45181524-45181524 on Build GRCh38
rs571038530 Chr.19: 45181380-45181380 on Build GRCh38
rs778205907 Chr.19: 45181404-45181404 on Build GRCh38
rs79968982 Chr.19: 45181191-45181191 on Build GRCh38
rs1056618303 Chr.19: 45181604-45181604 on Build GRCh38
rs910937341 Chr.19: 45181451-45181451 on Build GRCh38
rs1045564383 Chr.19: 45181570-45181570 on Build GRCh38
rs541422676 Chr.19: 45181221-45181221 on Build GRCh38
rs748256999 Chr.19: 45181179-45181179 on Build GRCh38
rs76151738 Chr.19: 45181605-45181605 on Build GRCh38
rs1029909320 Chr.19: 45181431-45181431 on Build GRCh38
rs964231841 Chr.19: 45181471-45181471 on Build GRCh38
rs868389116 Chr.19: 45181494-45181494 on Build GRCh38
rs905269479 Chr.19: 45181591-45181591 on Build GRCh38
rs972365156 Chr.19: 45181174-45181174 on Build GRCh38
rs745911670 Chr.19: 45181551-45181551 on Build GRCh38
rs11669523 Chr.19: 45181203-45181203 on Build GRCh38
rs1052414452 Chr.19: 45181254-45181254 on Build GRCh38
rs952354586 Chr.19: 45181432-45181432 on Build GRCh38
rs1025252330 Chr.19: 45181416-45181416 on Build GRCh38
rs926918507 Chr.19: 45181247-45181247 on Build GRCh38
rs533346539 Chr.19: 45181486-45181486 on Build GRCh38
rs1035715134 Chr.19: 45181395-45181395 on Build GRCh38
rs758853439 Chr.19: 45181281-45181281 on Build GRCh38
rs895666292 Chr.19: 45181308-45181308 on Build GRCh38
rs982947597 Chr.19: 45181475-45181475 on Build GRCh38
rs935221747 Chr.19: 45181518-45181518 on Build GRCh38
rs757522265 Chr.19: 45181482-45181482 on Build GRCh38
rs1000637685 Chr.19: 45181374-45181374 on Build GRCh38
rs1041522965 Chr.19: 45181616-45181616 on Build GRCh38
rs1041365424 Chr.19: 45181241-45181241 on Build GRCh38
rs1041807518 Chr.19: 45181321-45181321 on Build GRCh38
rs562085964 Chr.19: 45181251-45181251 on Build GRCh38
rs73939649 Chr.19: 45181376-45181376 on Build GRCh38
rs10404558 Chr.19: 45181553-45181553 on Build GRCh38
rs753263213 Chr.19: 45181301-45181301 on Build GRCh38
rs528119623 Chr.19: 45181218-45181218 on Build GRCh38
rs567632512 Chr.19: 45181399-45181399 on Build GRCh38
rs1045130441 Chr.19: 45181347-45181347 on Build GRCh38
rs576936281 Chr.19: 45181158-45181158 on Build GRCh38
rs531120532 Chr.19: 45181350-45181350 on Build GRCh38
rs893922630 Chr.19: 45181284-45181284 on Build GRCh38
rs775553024 Chr.19: 45181597-45181597 on Build GRCh38
rs867174019 Chr.19: 45181516-45181516 on Build GRCh38
rs10425237 Chr.19: 45181358-45181358 on Build GRCh38
rs545902071 Chr.19: 45181176-45181176 on Build GRCh38
rs778241189 Chr.19: 45181396-45181396 on Build GRCh38
rs951551100 Chr.19: 45181460-45181460 on Build GRCh38
rs984298489 Chr.19: 45181438-45181438 on Build GRCh38
rs113072092 Chr.19: 45181318-45181318 on Build GRCh38
rs191704392 Chr.19: 45181567-45181567 on Build GRCh38
rs770064737 Chr.19: 45181587-45181587 on Build GRCh38
rs747345556 Chr.19: 45181560-45181560 on Build GRCh38

View Full Product Details