Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45317926-45318454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKM
Entrez Gene ID
1158
Gene Name
creatine kinase, M-type
Gene Aliases
CKMM, M-CK
UniGene
Hs.334347
Gene Chromosome Location
Chr.19: 45306413-45322977 on Build GRCh38
Transcripts
NM_001824.4 XM_017026240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5571844 c.223G>A p.V75M Chr.19: 45317950-45317950 on Build GRCh38
237203 c.222C>T p.C74C Chr.19: 45317951-45317951 on Build GRCh38
1394582 c.247G>A p.E83K Chr.19: 45317926-45317926 on Build GRCh38
4401894 c.233A>G p.D78G Chr.19: 45317940-45317940 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563111119 Chr.19: 45317951-45317951 on Build GRCh38
rs771533664 Chr.19: 45317950-45317950 on Build GRCh38
rs555903473 Chr.19: 45318029-45318029 on Build GRCh38
rs746241019 Chr.19: 45317952-45317952 on Build GRCh38
rs547036105 Chr.19: 45317988-45317988 on Build GRCh38
rs543159254 Chr.19: 45318226-45318226 on Build GRCh38
rs191229714 Chr.19: 45317990-45317990 on Build GRCh38
rs1026074273 Chr.19: 45318228-45318228 on Build GRCh38
rs995862776 Chr.19: 45318048-45318048 on Build GRCh38
rs946133297 Chr.19: 45318236-45318236 on Build GRCh38
rs1042197727 Chr.19: 45318267-45318267 on Build GRCh38
rs1015052213 Chr.19: 45318141-45318141 on Build GRCh38
rs936516546 Chr.19: 45318337-45318337 on Build GRCh38
rs758727666 Chr.19: 45318001-45318023 on Build GRCh38
rs1803285 Chr.19: 45317959-45317959 on Build GRCh38
rs762415226 Chr.19: 45318413-45318413 on Build GRCh38
rs531821215 Chr.19: 45318342-45318342 on Build GRCh38
rs78043494 Chr.19: 45318424-45318424 on Build GRCh38
rs374682241 Chr.19: 45318179-45318179 on Build GRCh38
rs1027498188 Chr.19: 45318140-45318141 on Build GRCh38
rs952127006 Chr.19: 45318164-45318164 on Build GRCh38
rs778778671 Chr.19: 45317984-45317984 on Build GRCh38
rs1018130991 Chr.19: 45318426-45318426 on Build GRCh38
rs753240233 Chr.19: 45317984-45317985 on Build GRCh38
rs370509495 Chr.19: 45318121-45318121 on Build GRCh38
rs565434890 Chr.19: 45318414-45318414 on Build GRCh38
rs760997348 Chr.19: 45317964-45317964 on Build GRCh38
rs432979 Chr.19: 45318433-45318433 on Build GRCh38
rs958971140 Chr.19: 45318165-45318165 on Build GRCh38
rs374179495 Chr.19: 45318030-45318030 on Build GRCh38
rs574168539 Chr.19: 45318222-45318222 on Build GRCh38
rs533543986 Chr.19: 45317979-45317979 on Build GRCh38
rs560500225 Chr.19: 45318158-45318158 on Build GRCh38
rs563044312 Chr.19: 45318227-45318227 on Build GRCh38
rs182320411 Chr.19: 45317999-45317999 on Build GRCh38
rs779460216 Chr.19: 45317947-45317947 on Build GRCh38
rs867641920 Chr.19: 45318147-45318147 on Build GRCh38
rs966172981 Chr.19: 45318430-45318430 on Build GRCh38
rs774984373 Chr.19: 45318373-45318373 on Build GRCh38
rs558589324 Chr.19: 45318080-45318080 on Build GRCh38
rs571181525 Chr.19: 45317972-45317972 on Build GRCh38
rs769477627 Chr.19: 45318268-45318268 on Build GRCh38
rs752272939 Chr.19: 45317994-45317994 on Build GRCh38
rs753391216 Chr.19: 45318006-45318006 on Build GRCh38
rs904842747 Chr.19: 45318366-45318366 on Build GRCh38
rs914744625 Chr.19: 45318336-45318336 on Build GRCh38
rs992773960 Chr.19: 45318219-45318219 on Build GRCh38
rs554249174 Chr.19: 45318398-45318399 on Build GRCh38
rs990242336 Chr.19: 45318320-45318320 on Build GRCh38
rs946483796 Chr.19: 45318347-45318347 on Build GRCh38
rs376182290 Chr.19: 45318421-45318421 on Build GRCh38
rs757644382 Chr.19: 45318009-45318009 on Build GRCh38
rs11559023 Chr.19: 45317957-45317957 on Build GRCh38
rs371804927 Chr.19: 45318374-45318374 on Build GRCh38
rs927554764 Chr.19: 45318154-45318154 on Build GRCh38
rs897641506 Chr.19: 45318072-45318072 on Build GRCh38
rs1045248418 Chr.19: 45318354-45318354 on Build GRCh38
rs779411481 Chr.19: 45318012-45318012 on Build GRCh38
rs1030028366 Chr.19: 45318056-45318056 on Build GRCh38
rs934926866 Chr.19: 45318296-45318296 on Build GRCh38
rs576014859 Chr.19: 45318413-45318413 on Build GRCh38
rs80301995 Chr.19: 45318073-45318073 on Build GRCh38
rs900139863 Chr.19: 45318034-45318034 on Build GRCh38
rs576164882 Chr.19: 45318065-45318065 on Build GRCh38
rs140148281 Chr.19: 45318084-45318084 on Build GRCh38
rs986661750 Chr.19: 45318214-45318214 on Build GRCh38
rs946162795 Chr.19: 45317986-45317986 on Build GRCh38
rs772046364 Chr.19: 45318153-45318153 on Build GRCh38
rs377725988 Chr.19: 45317960-45317960 on Build GRCh38
rs201511125 Chr.19: 45317993-45317993 on Build GRCh38
rs972794895 Chr.19: 45318182-45318182 on Build GRCh38
rs776827903 Chr.19: 45317973-45317973 on Build GRCh38

View Full Product Details