Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45407614-45408115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD3EAP
Entrez Gene ID
10849
Gene Name
CD3e molecule associated protein
Gene Aliases
ASE-1, ASE1, CAST, PAF49
UniGene
Hs.710495
Gene Chromosome Location
Chr.19: 45406129-45410766 on Build GRCh38
Transcripts
NM_001297590.1 NM_012099.1 XM_017026180.1 XM_017026181.1 NM_001166049.1 NM_001983.3 XM_017026460.1 XM_017026461.1 XM_017026462.1 XM_017026463.1 XM_017026464.1 XM_017026465.1 XM_017026466.1

Gene Symbol
ERCC1
Entrez Gene ID
2067
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Gene Aliases
COFS4, RAD10, UV20
UniGene
Hs.435981
Gene Chromosome Location
Chr.19: 45407333-45478866 on Build GRCh38
Transcripts
NM_001297590.1 NM_012099.1 XM_017026180.1 XM_017026181.1 NM_001166049.1 NM_001983.3 XM_017026460.1 XM_017026461.1 XM_017026462.1 XM_017026463.1 XM_017026464.1 XM_017026465.1 XM_017026466.1

SNP Detail

SNP ID Chromosome Location
rs924484490 Chr.19: 45407677-45407677 on Build GRCh38
rs889903027 Chr.19: 45407978-45407978 on Build GRCh38
rs35708119 Chr.19: 45408060-45408061 on Build GRCh38
rs770141832 Chr.19: 45408082-45408082 on Build GRCh38
rs763402713 Chr.19: 45407752-45407752 on Build GRCh38
rs764315369 Chr.19: 45407758-45407758 on Build GRCh38
rs578208320 Chr.19: 45407865-45407865 on Build GRCh38
rs567269017 Chr.19: 45407736-45407736 on Build GRCh38
rs555484529 Chr.19: 45407782-45407782 on Build GRCh38
rs542570048 Chr.19: 45408002-45408002 on Build GRCh38
rs1047272823 Chr.19: 45407794-45407794 on Build GRCh38
rs547087785 Chr.19: 45408051-45408052 on Build GRCh38
rs547191855 Chr.19: 45408078-45408078 on Build GRCh38
rs75205129 Chr.19: 45408053-45408053 on Build GRCh38
rs540871734 Chr.19: 45407951-45407951 on Build GRCh38
rs1053376869 Chr.19: 45407937-45407937 on Build GRCh38
rs907370976 Chr.19: 45407920-45407920 on Build GRCh38
rs558309295 Chr.19: 45407803-45407803 on Build GRCh38
rs193096276 Chr.19: 45407787-45407787 on Build GRCh38
rs2013521 Chr.19: 45408072-45408072 on Build GRCh38
rs986398382 Chr.19: 45407725-45407725 on Build GRCh38
rs908981385 Chr.19: 45407827-45407827 on Build GRCh38
rs984535155 Chr.19: 45407826-45407826 on Build GRCh38
rs368358045 Chr.19: 45408051-45408051 on Build GRCh38
rs1044995785 Chr.19: 45407881-45407881 on Build GRCh38
rs200025030 Chr.19: 45408060-45408060 on Build GRCh38
rs1003138740 Chr.19: 45407805-45407805 on Build GRCh38
rs948905285 Chr.19: 45407843-45407843 on Build GRCh38
rs187794054 Chr.19: 45408073-45408073 on Build GRCh38
rs934457740 Chr.19: 45407698-45407698 on Build GRCh38
rs373645122 Chr.19: 45408072-45408072 on Build GRCh38
rs965674545 Chr.19: 45407853-45407853 on Build GRCh38
rs567181439 Chr.19: 45408091-45408091 on Build GRCh38
rs1012014599 Chr.19: 45407963-45407963 on Build GRCh38
rs1044405628 Chr.19: 45407797-45407797 on Build GRCh38
rs938879545 Chr.19: 45407927-45407927 on Build GRCh38
rs749522470 Chr.19: 45408092-45408092 on Build GRCh38
rs1007391807 Chr.19: 45407980-45407980 on Build GRCh38
rs553640912 Chr.19: 45407699-45407699 on Build GRCh38
rs531407458 Chr.19: 45408023-45408023 on Build GRCh38
rs771100281 Chr.19: 45408093-45408093 on Build GRCh38
rs143529478 Chr.19: 45407757-45407757 on Build GRCh38
rs996116077 Chr.19: 45407825-45407825 on Build GRCh38
rs117128015 Chr.19: 45407645-45407645 on Build GRCh38
rs183074620 Chr.19: 45407995-45407995 on Build GRCh38
rs961569382 Chr.19: 45407822-45407822 on Build GRCh38
rs202063629 Chr.19: 45408071-45408071 on Build GRCh38
rs747498774 Chr.19: 45408062-45408063 on Build GRCh38
rs965974902 Chr.19: 45407783-45407783 on Build GRCh38
rs767797029 Chr.19: 45407821-45407821 on Build GRCh38
rs1017353870 Chr.19: 45408003-45408003 on Build GRCh38
rs8103749 Chr.19: 45408062-45408062 on Build GRCh38
rs5828233 Chr.19: 45408061-45408061 on Build GRCh38
rs934593217 Chr.19: 45407983-45407983 on Build GRCh38
rs868539528 Chr.19: 45407766-45407766 on Build GRCh38
rs985858065 Chr.19: 45407837-45407837 on Build GRCh38
rs560766196 Chr.19: 45407968-45407968 on Build GRCh38
rs1021698537 Chr.19: 45407969-45407969 on Build GRCh38
rs148038609 Chr.19: 45407799-45407799 on Build GRCh38
rs545753812 Chr.19: 45408009-45408009 on Build GRCh38
rs907220893 Chr.19: 45408032-45408032 on Build GRCh38
rs11314106 Chr.19: 45408062-45408062 on Build GRCh38
rs910239756 Chr.19: 45407846-45407846 on Build GRCh38
rs533771641 Chr.19: 45407678-45407678 on Build GRCh38
rs990425265 Chr.19: 45407991-45407991 on Build GRCh38
rs77052728 Chr.19: 45408064-45408064 on Build GRCh38
rs891776741 Chr.19: 45407960-45407960 on Build GRCh38

View Full Product Details