Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45470493-45471010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERCC1
Entrez Gene ID
2067
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Gene Aliases
COFS4, RAD10, UV20
UniGene
Hs.435981
Gene Chromosome Location
Chr.19: 45407333-45478866 on Build GRCh38
Transcripts
XM_005258636.4 XM_017026464.1 NM_001114171.1 NM_006732.2 XM_005258691.1

Gene Symbol
FOSB
Entrez Gene ID
2354
Gene Name
FosB proto-oncogene, AP-1 transcription factor subunit
Gene Aliases
AP-1, G0S3, GOS3, GOSB
UniGene
Hs.590958
Gene Chromosome Location
Chr.19: 45467995-45475179 on Build GRCh38
Transcripts
XM_005258636.4 XM_017026464.1 NM_001114171.1 NM_006732.2 XM_005258691.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6666021 c.346G>A p.G116R Chr.19: 45470848-45470848 on Build GRCh38
3104196 c.156T>C p.G52G Chr.19: 45470658-45470658 on Build GRCh38
4788445 c.414C>A p.A138A Chr.19: 45470916-45470916 on Build GRCh38
3104201 c.276G>A p.P92P Chr.19: 45470778-45470778 on Build GRCh38
6666019 c.282C>T p.V94V Chr.19: 45470784-45470784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780193148 Chr.19: 45470722-45470722 on Build GRCh38
rs745617183 Chr.19: 45470754-45470754 on Build GRCh38
rs952454220 Chr.19: 45470522-45470522 on Build GRCh38
rs915362601 Chr.19: 45470643-45470643 on Build GRCh38
rs377014678 Chr.19: 45470881-45470881 on Build GRCh38
rs769317042 Chr.19: 45470938-45470938 on Build GRCh38
rs969633804 Chr.19: 45470539-45470539 on Build GRCh38
rs1046677173 Chr.19: 45470889-45470889 on Build GRCh38
rs149541859 Chr.19: 45470538-45470538 on Build GRCh38
rs891300831 Chr.19: 45470670-45470670 on Build GRCh38
rs773872677 Chr.19: 45470582-45470582 on Build GRCh38
rs750762281 Chr.19: 45470716-45470716 on Build GRCh38
rs147923411 Chr.19: 45470661-45470661 on Build GRCh38
rs143780767 Chr.19: 45470848-45470848 on Build GRCh38
rs756161522 Chr.19: 45470825-45470825 on Build GRCh38
rs200272813 Chr.19: 45470713-45470713 on Build GRCh38
rs756210224 Chr.19: 45470896-45470896 on Build GRCh38
rs901080791 Chr.19: 45470874-45470874 on Build GRCh38
rs755048094 Chr.19: 45470726-45470726 on Build GRCh38
rs746820250 Chr.19: 45470851-45470851 on Build GRCh38
rs373545463 Chr.19: 45470863-45470863 on Build GRCh38
rs376676928 Chr.19: 45470984-45470984 on Build GRCh38
rs542156920 Chr.19: 45470963-45470963 on Build GRCh38
rs754284435 Chr.19: 45470612-45470613 on Build GRCh38
rs1004104256 Chr.19: 45470970-45470970 on Build GRCh38
rs776831910 Chr.19: 45470583-45470583 on Build GRCh38
rs139180415 Chr.19: 45470516-45470516 on Build GRCh38
rs759371386 Chr.19: 45470611-45470611 on Build GRCh38
rs747027527 Chr.19: 45470635-45470635 on Build GRCh38
rs557145094 Chr.19: 45470565-45470565 on Build GRCh38
rs751335932 Chr.19: 45470917-45470917 on Build GRCh38
rs940150662 Chr.19: 45470983-45470983 on Build GRCh38
rs1005680812 Chr.19: 45470526-45470526 on Build GRCh38
rs745855821 Chr.19: 45470838-45470838 on Build GRCh38
rs776235889 Chr.19: 45470776-45470776 on Build GRCh38
rs760266207 Chr.19: 45470601-45470601 on Build GRCh38
rs940036099 Chr.19: 45470615-45470615 on Build GRCh38
rs764116136 Chr.19: 45470908-45470908 on Build GRCh38
rs767756271 Chr.19: 45470808-45470808 on Build GRCh38
rs780174960 Chr.19: 45470912-45470912 on Build GRCh38
rs775295136 Chr.19: 45470673-45470673 on Build GRCh38
rs747823029 Chr.19: 45470923-45470923 on Build GRCh38
rs371768649 Chr.19: 45470613-45470613 on Build GRCh38
rs746697548 Chr.19: 45470772-45470772 on Build GRCh38
rs570937964 Chr.19: 45470623-45470623 on Build GRCh38
rs770818762 Chr.19: 45470579-45470579 on Build GRCh38
rs183683887 Chr.19: 45470655-45470655 on Build GRCh38
rs766662878 Chr.19: 45470604-45470607 on Build GRCh38
rs767097253 Chr.19: 45470592-45470592 on Build GRCh38
rs765713415 Chr.19: 45470790-45470790 on Build GRCh38
rs752214866 Chr.19: 45470600-45470600 on Build GRCh38
rs898478668 Chr.19: 45470656-45470656 on Build GRCh38
rs763363699 Chr.19: 45470794-45470794 on Build GRCh38
rs757176467 Chr.19: 45470746-45470746 on Build GRCh38
rs751752030 Chr.19: 45470802-45470802 on Build GRCh38
rs761208292 Chr.19: 45470584-45470587 on Build GRCh38
rs760245782 Chr.19: 45470679-45470679 on Build GRCh38
rs946749683 Chr.19: 45470779-45470779 on Build GRCh38
rs765687773 Chr.19: 45470715-45470715 on Build GRCh38
rs780887411 Chr.19: 45470920-45470920 on Build GRCh38
rs201846388 Chr.19: 45470987-45470987 on Build GRCh38
rs369628703 Chr.19: 45470826-45470826 on Build GRCh38
rs368913532 Chr.19: 45470586-45470586 on Build GRCh38
rs781290122 Chr.19: 45470736-45470736 on Build GRCh38
rs754363313 Chr.19: 45470691-45470691 on Build GRCh38
rs568499235 Chr.19: 45470517-45470517 on Build GRCh38
rs768255804 Chr.19: 45470773-45470773 on Build GRCh38
rs771687459 Chr.19: 45470671-45470671 on Build GRCh38
rs752610193 Chr.19: 45470975-45470975 on Build GRCh38
rs777674537 Chr.19: 45470638-45470638 on Build GRCh38
rs375983768 Chr.19: 45470637-45470637 on Build GRCh38
rs932925064 Chr.19: 45470658-45470658 on Build GRCh38
rs1023774695 Chr.19: 45470906-45470906 on Build GRCh38
rs1023722390 Chr.19: 45470534-45470534 on Build GRCh38
rs755244213 Chr.19: 45470803-45470803 on Build GRCh38
rs749291738 Chr.19: 45470644-45470644 on Build GRCh38
rs758698866 Chr.19: 45470719-45470719 on Build GRCh38
rs761262027 Chr.19: 45470777-45470777 on Build GRCh38
rs905390627 Chr.19: 45470948-45470948 on Build GRCh38
rs754837542 Chr.19: 45470918-45470918 on Build GRCh38
rs758176585 Chr.19: 45470842-45470842 on Build GRCh38
rs759880826 Chr.19: 45470986-45470986 on Build GRCh38
rs763411269 Chr.19: 45470880-45470880 on Build GRCh38
rs911028906 Chr.19: 45470795-45470795 on Build GRCh38
rs758600513 Chr.19: 45470634-45470634 on Build GRCh38
rs775272926 Chr.19: 45470771-45470771 on Build GRCh38
rs778720566 Chr.19: 45470750-45470750 on Build GRCh38
rs573196315 Chr.19: 45470911-45470911 on Build GRCh38
rs141764521 Chr.19: 45470793-45470793 on Build GRCh38
rs201193101 Chr.19: 45470892-45470892 on Build GRCh38
rs942514584 Chr.19: 45470847-45470847 on Build GRCh38
rs748248206 Chr.19: 45470745-45470745 on Build GRCh38
rs768386636 Chr.19: 45470852-45470852 on Build GRCh38
rs763759245 Chr.19: 45470685-45470685 on Build GRCh38
rs370113314 Chr.19: 45470894-45470894 on Build GRCh38
rs201608348 Chr.19: 45470865-45470865 on Build GRCh38
rs199733739 Chr.19: 45470801-45470801 on Build GRCh38
rs981275085 Chr.19: 45470931-45470931 on Build GRCh38
rs771817357 Chr.19: 45470766-45470766 on Build GRCh38
rs747640609 Chr.19: 45470861-45470861 on Build GRCh38
rs774657495 Chr.19: 45470886-45470886 on Build GRCh38
rs377466585 Chr.19: 45470893-45470893 on Build GRCh38
rs759281171 Chr.19: 45470587-45470587 on Build GRCh38
rs776431088 Chr.19: 45470853-45470853 on Build GRCh38
rs750349108 Chr.19: 45470909-45470909 on Build GRCh38
rs773764980 Chr.19: 45470872-45470872 on Build GRCh38
rs757689027 Chr.19: 45470617-45470617 on Build GRCh38
rs982359382 Chr.19: 45470541-45470541 on Build GRCh38

View Full Product Details