Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45614464-45614915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML2
Entrez Gene ID
24139
Gene Name
echinoderm microtubule associated protein like 2
Gene Aliases
ELP70, EMAP-2, EMAP2
UniGene
Hs.24178
Gene Chromosome Location
Chr.19: 45609396-45645601 on Build GRCh38
Transcripts
NM_001193268.1 NM_001193269.1 NM_012155.2 NR_034098.1 XM_006723125.2 XM_006723126.2 XM_011526694.2 XM_011526695.2 XM_011526698.2 XM_011526700.2 XM_011526701.2 XM_017026550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653763 c.2078C>A p.A693D Chr.19: 45614661-45614661 on Build GRCh38
6999154 c.1643G>A p.R548K Chr.19: 45614655-45614655 on Build GRCh38
6653754 c.2065A>G p.T689A Chr.19: 45614674-45614674 on Build GRCh38
5929875 c.2039-3C>T p.? Chr.19: 45614703-45614703 on Build GRCh38
5929880 c.2201-4C>T p.? Chr.19: 45614704-45614704 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755470509 Chr.19: 45614835-45614835 on Build GRCh38
rs1015718584 Chr.19: 45614874-45614874 on Build GRCh38
rs1006750018 Chr.19: 45614891-45614891 on Build GRCh38
rs756884749 Chr.19: 45614704-45614704 on Build GRCh38
rs995288907 Chr.19: 45614892-45614892 on Build GRCh38
rs890931617 Chr.19: 45614862-45614862 on Build GRCh38
rs747549620 Chr.19: 45614574-45614574 on Build GRCh38
rs746170320 Chr.19: 45614709-45614709 on Build GRCh38
rs769328978 Chr.19: 45614577-45614577 on Build GRCh38
rs771363008 Chr.19: 45614698-45614698 on Build GRCh38
rs3816046 Chr.19: 45614869-45614869 on Build GRCh38
rs767033715 Chr.19: 45614729-45614729 on Build GRCh38
rs750362474 Chr.19: 45614666-45614666 on Build GRCh38
rs563968762 Chr.19: 45614770-45614770 on Build GRCh38
rs959603221 Chr.19: 45614664-45614664 on Build GRCh38
rs919631184 Chr.19: 45614662-45614662 on Build GRCh38
rs562299343 Chr.19: 45614556-45614556 on Build GRCh38
rs761025379 Chr.19: 45614731-45614731 on Build GRCh38
rs113810892 Chr.19: 45614582-45614582 on Build GRCh38
rs150352479 Chr.19: 45614873-45614873 on Build GRCh38
rs541955404 Chr.19: 45614547-45614547 on Build GRCh38
rs764307594 Chr.19: 45614649-45614649 on Build GRCh38
rs201142610 Chr.19: 45614619-45614619 on Build GRCh38
rs749846144 Chr.19: 45614583-45614583 on Build GRCh38
rs932557914 Chr.19: 45614832-45614832 on Build GRCh38
rs991097281 Chr.19: 45614678-45614678 on Build GRCh38
rs1019580852 Chr.19: 45614502-45614502 on Build GRCh38
rs774936223 Chr.19: 45614700-45614700 on Build GRCh38
rs749793807 Chr.19: 45614697-45614697 on Build GRCh38
rs749973349 Chr.19: 45614776-45614776 on Build GRCh38
rs545781628 Chr.19: 45614568-45614568 on Build GRCh38
rs35344004 Chr.19: 45614620-45614620 on Build GRCh38
rs376047965 Chr.19: 45614670-45614670 on Build GRCh38
rs780672327 Chr.19: 45614690-45614690 on Build GRCh38
rs571019446 Chr.19: 45614681-45614681 on Build GRCh38
rs775413699 Chr.19: 45614550-45614550 on Build GRCh38
rs376733886 Chr.19: 45614779-45614779 on Build GRCh38
rs372961924 Chr.19: 45614639-45614639 on Build GRCh38
rs760797620 Chr.19: 45614640-45614640 on Build GRCh38
rs530921456 Chr.19: 45614880-45614880 on Build GRCh38
rs772518360 Chr.19: 45614720-45614720 on Build GRCh38
rs145412457 Chr.19: 45614611-45614611 on Build GRCh38
rs777325409 Chr.19: 45614695-45614695 on Build GRCh38
rs767577888 Chr.19: 45614627-45614627 on Build GRCh38
rs764254242 Chr.19: 45614733-45614733 on Build GRCh38
rs936630239 Chr.19: 45614748-45614748 on Build GRCh38
rs191770936 Chr.19: 45614751-45614751 on Build GRCh38
rs924059048 Chr.19: 45614723-45614723 on Build GRCh38
rs775542399 Chr.19: 45614728-45614728 on Build GRCh38
rs201278347 Chr.19: 45614693-45614693 on Build GRCh38
rs771275133 Chr.19: 45614602-45614602 on Build GRCh38
rs762778606 Chr.19: 45614667-45614667 on Build GRCh38
rs549908283 Chr.19: 45614875-45614875 on Build GRCh38
rs376117039 Chr.19: 45614750-45614750 on Build GRCh38
rs140164971 Chr.19: 45614694-45614694 on Build GRCh38

View Full Product Details