Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45730757-45731265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO46
Entrez Gene ID
23403
Gene Name
F-box protein 46
Gene Aliases
20D7-FC4, FBXO34L, Fbx46
UniGene
Hs.128702
Gene Chromosome Location
Chr.19: 45710629-45730893 on Build GRCh38
Transcripts
NM_001080469.1

SNP Detail

SNP ID Chromosome Location
rs966798635 Chr.19: 45731192-45731192 on Build GRCh38
rs1039602753 Chr.19: 45730940-45730940 on Build GRCh38
rs140788200 Chr.19: 45731229-45731229 on Build GRCh38
rs982557579 Chr.19: 45731158-45731158 on Build GRCh38
rs925307415 Chr.19: 45730901-45730901 on Build GRCh38
rs548776658 Chr.19: 45731120-45731120 on Build GRCh38
rs1002449886 Chr.19: 45731073-45731073 on Build GRCh38
rs975632362 Chr.19: 45731183-45731183 on Build GRCh38
rs1024054179 Chr.19: 45731141-45731141 on Build GRCh38
rs192900066 Chr.19: 45730854-45730854 on Build GRCh38
rs933876856 Chr.19: 45730965-45730965 on Build GRCh38
rs1021554992 Chr.19: 45731172-45731172 on Build GRCh38
rs879448781 Chr.19: 45731060-45731060 on Build GRCh38
rs921086773 Chr.19: 45730952-45730952 on Build GRCh38
rs17878252 Chr.19: 45730897-45730897 on Build GRCh38
rs1051266815 Chr.19: 45730968-45730968 on Build GRCh38
rs111608777 Chr.19: 45730874-45730874 on Build GRCh38
rs748468258 Chr.19: 45730932-45730932 on Build GRCh38
rs34964212 Chr.19: 45730837-45730837 on Build GRCh38
rs1013784611 Chr.19: 45731130-45731130 on Build GRCh38
rs549318232 Chr.19: 45730802-45730802 on Build GRCh38
rs1034250580 Chr.19: 45731086-45731086 on Build GRCh38
rs1041975562 Chr.19: 45731011-45731011 on Build GRCh38
rs904365133 Chr.19: 45731012-45731012 on Build GRCh38
rs1033814349 Chr.19: 45730840-45730840 on Build GRCh38
rs951238607 Chr.19: 45731151-45731151 on Build GRCh38
rs945358792 Chr.19: 45730892-45730892 on Build GRCh38
rs925250273 Chr.19: 45731213-45731213 on Build GRCh38
rs958338384 Chr.19: 45731104-45731104 on Build GRCh38
rs1004382347 Chr.19: 45731031-45731031 on Build GRCh38
rs867000678 Chr.19: 45730875-45730875 on Build GRCh38
rs534810145 Chr.19: 45731029-45731029 on Build GRCh38
rs192848430 Chr.19: 45731036-45731036 on Build GRCh38
rs1000033077 Chr.19: 45731034-45731034 on Build GRCh38
rs569432374 Chr.19: 45730833-45730833 on Build GRCh38
rs981038414 Chr.19: 45730811-45730811 on Build GRCh38
rs992514786 Chr.19: 45730801-45730801 on Build GRCh38
rs539581309 Chr.19: 45731009-45731009 on Build GRCh38
rs898172522 Chr.19: 45731072-45731072 on Build GRCh38
rs899810942 Chr.19: 45730906-45730906 on Build GRCh38
rs933991335 Chr.19: 45730915-45730915 on Build GRCh38
rs960848133 Chr.19: 45731111-45731111 on Build GRCh38
rs935883624 Chr.19: 45730925-45730925 on Build GRCh38
rs568316830 Chr.19: 45731218-45731218 on Build GRCh38
rs979930346 Chr.19: 45731208-45731208 on Build GRCh38
rs914013291 Chr.19: 45730876-45730876 on Build GRCh38
rs1051166961 Chr.19: 45731006-45731006 on Build GRCh38
rs759370017 Chr.19: 45731109-45731109 on Build GRCh38
rs909999024 Chr.19: 45731166-45731166 on Build GRCh38
rs554914428 Chr.19: 45731030-45731030 on Build GRCh38
rs1019814787 Chr.19: 45730855-45730855 on Build GRCh38

View Full Product Details