Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45872371-45872870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXA3
Entrez Gene ID
3171
Gene Name
forkhead box A3
Gene Aliases
FKHH3, HNF3G, TCF3G
UniGene
Hs.36137
Gene Chromosome Location
Chr.19: 45864260-45873797 on Build GRCh38
Transcripts
NM_004497.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
248772 c.512T>C p.F171S Chr.19: 45872517-45872517 on Build GRCh38
5800770 c.518A>G p.D173G Chr.19: 45872523-45872523 on Build GRCh38
5741645 c.626G>A p.R209H Chr.19: 45872631-45872631 on Build GRCh38
7204650 c.539G>A p.R180H Chr.19: 45872544-45872544 on Build GRCh38
5707081 c.583C>T p.P195S Chr.19: 45872588-45872588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772799849 Chr.19: 45872833-45872833 on Build GRCh38
rs760139463 Chr.19: 45872844-45872844 on Build GRCh38
rs753914826 Chr.19: 45872403-45872403 on Build GRCh38
rs531001040 Chr.19: 45872701-45872701 on Build GRCh38
rs756819807 Chr.19: 45872577-45872577 on Build GRCh38
rs28666870 Chr.19: 45872785-45872785 on Build GRCh38
rs762659076 Chr.19: 45872834-45872834 on Build GRCh38
rs776353999 Chr.19: 45872737-45872737 on Build GRCh38
rs1004526212 Chr.19: 45872805-45872805 on Build GRCh38
rs770985467 Chr.19: 45872800-45872800 on Build GRCh38
rs747341541 Chr.19: 45872626-45872626 on Build GRCh38
rs780833457 Chr.19: 45872452-45872452 on Build GRCh38
rs764817302 Chr.19: 45872702-45872702 on Build GRCh38
rs535321568 Chr.19: 45872646-45872646 on Build GRCh38
rs973562577 Chr.19: 45872515-45872515 on Build GRCh38
rs373998257 Chr.19: 45872542-45872542 on Build GRCh38
rs768531867 Chr.19: 45872819-45872819 on Build GRCh38
rs777278399 Chr.19: 45872458-45872458 on Build GRCh38
rs769611712 Chr.19: 45872832-45872832 on Build GRCh38
rs556790003 Chr.19: 45872755-45872755 on Build GRCh38
rs758774573 Chr.19: 45872605-45872605 on Build GRCh38
rs879018916 Chr.19: 45872760-45872760 on Build GRCh38
rs746349583 Chr.19: 45872471-45872471 on Build GRCh38
rs766381222 Chr.19: 45872407-45872407 on Build GRCh38
rs200377130 Chr.19: 45872469-45872469 on Build GRCh38
rs865961018 Chr.19: 45872552-45872552 on Build GRCh38
rs750062748 Chr.19: 45872754-45872754 on Build GRCh38
rs759526623 Chr.19: 45872665-45872665 on Build GRCh38
rs746963773 Chr.19: 45872812-45872812 on Build GRCh38
rs748746722 Chr.19: 45872461-45872461 on Build GRCh38
rs141782286 Chr.19: 45872404-45872404 on Build GRCh38
rs926920586 Chr.19: 45872475-45872475 on Build GRCh38
rs12976619 Chr.19: 45872492-45872492 on Build GRCh38
rs1005769337 Chr.19: 45872631-45872631 on Build GRCh38
rs771010717 Chr.19: 45872733-45872733 on Build GRCh38
rs372030164 Chr.19: 45872566-45872566 on Build GRCh38
rs201747924 Chr.19: 45872748-45872748 on Build GRCh38
rs937004765 Chr.19: 45872743-45872743 on Build GRCh38
rs768096522 Chr.19: 45872845-45872845 on Build GRCh38
rs200743469 Chr.19: 45872716-45872716 on Build GRCh38
rs765990333 Chr.19: 45872836-45872836 on Build GRCh38
rs778283292 Chr.19: 45872593-45872593 on Build GRCh38
rs753556194 Chr.19: 45872680-45872680 on Build GRCh38
rs781684132 Chr.19: 45872722-45872722 on Build GRCh38
rs374769245 Chr.19: 45872446-45872446 on Build GRCh38
rs759167859 Chr.19: 45872556-45872556 on Build GRCh38
rs199787706 Chr.19: 45872497-45872497 on Build GRCh38
rs762026809 Chr.19: 45872526-45872526 on Build GRCh38
rs773364143 Chr.19: 45872730-45872730 on Build GRCh38
rs767496597 Chr.19: 45872511-45872511 on Build GRCh38
rs773073897 Chr.19: 45872650-45872652 on Build GRCh38
rs12984947 Chr.19: 45872576-45872576 on Build GRCh38
rs769846056 Chr.19: 45872727-45872727 on Build GRCh38
rs767307934 Chr.19: 45872670-45872670 on Build GRCh38
rs755785237 Chr.19: 45872551-45872551 on Build GRCh38
rs548866684 Chr.19: 45872707-45872707 on Build GRCh38
rs751051964 Chr.19: 45872839-45872839 on Build GRCh38
rs763864928 Chr.19: 45872678-45872678 on Build GRCh38
rs760500600 Chr.19: 45872677-45872677 on Build GRCh38
rs760604416 Chr.19: 45872736-45872736 on Build GRCh38
rs751423514 Chr.19: 45872439-45872439 on Build GRCh38
rs765382731 Chr.19: 45872530-45872530 on Build GRCh38
rs566405768 Chr.19: 45872572-45872572 on Build GRCh38
rs769980450 Chr.19: 45872660-45872660 on Build GRCh38
rs371687599 Chr.19: 45872756-45872756 on Build GRCh38
rs371065526 Chr.19: 45872630-45872630 on Build GRCh38
rs946035893 Chr.19: 45872554-45872554 on Build GRCh38
rs1047679583 Chr.19: 45872767-45872767 on Build GRCh38
rs1018297057 Chr.19: 45872640-45872640 on Build GRCh38
rs769113907 Chr.19: 45872628-45872628 on Build GRCh38
rs778898121 Chr.19: 45872801-45872801 on Build GRCh38
rs749657388 Chr.19: 45872721-45872721 on Build GRCh38
rs141446079 Chr.19: 45872498-45872498 on Build GRCh38
rs552726739 Chr.19: 45872798-45872798 on Build GRCh38
rs773095488 Chr.19: 45872664-45872664 on Build GRCh38
rs1019491339 Chr.19: 45872729-45872729 on Build GRCh38
rs12977291 Chr.19: 45872488-45872488 on Build GRCh38
rs751569343 Chr.19: 45872531-45872531 on Build GRCh38
rs772555492 Chr.19: 45872483-45872483 on Build GRCh38
rs746391666 Chr.19: 45872599-45872599 on Build GRCh38
rs1030143565 Chr.19: 45872696-45872696 on Build GRCh38
rs753059025 Chr.19: 45872769-45872769 on Build GRCh38
rs780494558 Chr.19: 45872619-45872619 on Build GRCh38
rs775650612 Chr.19: 45872484-45872484 on Build GRCh38
rs776446460 Chr.19: 45872826-45872826 on Build GRCh38
rs775453392 Chr.19: 45872735-45872735 on Build GRCh38
rs771457315 Chr.19: 45872468-45872468 on Build GRCh38
rs16980091 Chr.19: 45872521-45872521 on Build GRCh38
rs867687500 Chr.19: 45872501-45872501 on Build GRCh38
rs954700998 Chr.19: 45872400-45872400 on Build GRCh38
rs758978324 Chr.19: 45872708-45872708 on Build GRCh38
rs781697700 Chr.19: 45872766-45872766 on Build GRCh38
rs12983057 Chr.19: 45872582-45872582 on Build GRCh38
rs140760356 Chr.19: 45872831-45872831 on Build GRCh38
rs865905914 Chr.19: 45872395-45872395 on Build GRCh38
rs539294853 Chr.19: 45872797-45872797 on Build GRCh38
rs755697325 Chr.19: 45872453-45872453 on Build GRCh38
rs560929375 Chr.19: 45872430-45872430 on Build GRCh38
rs752376903 Chr.19: 45872544-45872544 on Build GRCh38
rs868733101 Chr.19: 45872793-45872793 on Build GRCh38
rs1044654245 Chr.19: 45872448-45872448 on Build GRCh38

View Full Product Details