Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46016826-46017332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
UniGene
Hs.515479
Gene Chromosome Location
Chr.19: 45995457-46018616 on Build GRCh38
Transcripts
NM_001267723.1 XM_005259191.3 XM_005259192.4 XM_011527257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6307634 c.1404G>T p.T468T Chr.19: 46017283-46017283 on Build GRCh38
5100724 c.1393delC p.S466fs*10 Chr.19: 46017272-46017272 on Build GRCh38
3823455 c.1370G>A p.G457D Chr.19: 46017249-46017249 on Build GRCh38
3773073 c.1351G>A p.E451K Chr.19: 46017053-46017053 on Build GRCh38
6444518 c.1385G>C p.G462A Chr.19: 46017264-46017264 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897487347 Chr.19: 46016877-46016877 on Build GRCh38
rs746973754 Chr.19: 46016980-46016980 on Build GRCh38
rs761140439 Chr.19: 46017233-46017233 on Build GRCh38
rs748011816 Chr.19: 46017081-46017081 on Build GRCh38
rs745885230 Chr.19: 46017059-46017059 on Build GRCh38
rs910739844 Chr.19: 46017090-46017090 on Build GRCh38
rs758535693 Chr.19: 46017259-46017259 on Build GRCh38
rs778107894 Chr.19: 46017028-46017028 on Build GRCh38
rs61742389 Chr.19: 46017248-46017248 on Build GRCh38
rs750017734 Chr.19: 46017162-46017162 on Build GRCh38
rs373150690 Chr.19: 46017257-46017257 on Build GRCh38
rs767126781 Chr.19: 46017145-46017145 on Build GRCh38
rs1047898902 Chr.19: 46017256-46017256 on Build GRCh38
rs753973936 Chr.19: 46017029-46017029 on Build GRCh38
rs1034992200 Chr.19: 46016987-46016987 on Build GRCh38
rs891736367 Chr.19: 46017283-46017283 on Build GRCh38
rs772724071 Chr.19: 46017001-46017001 on Build GRCh38
rs777055979 Chr.19: 46017291-46017291 on Build GRCh38
rs760739400 Chr.19: 46017216-46017217 on Build GRCh38
rs539339728 Chr.19: 46017015-46017015 on Build GRCh38
rs756954672 Chr.19: 46017096-46017096 on Build GRCh38
rs528219154 Chr.19: 46016901-46016901 on Build GRCh38
rs574809072 Chr.19: 46017146-46017146 on Build GRCh38
rs753128185 Chr.19: 46017026-46017026 on Build GRCh38
rs1053444201 Chr.19: 46017282-46017282 on Build GRCh38
rs773859574 Chr.19: 46017105-46017105 on Build GRCh38
rs896651083 Chr.19: 46016946-46016946 on Build GRCh38
rs937707774 Chr.19: 46017169-46017169 on Build GRCh38
rs748070579 Chr.19: 46017278-46017278 on Build GRCh38
rs938257536 Chr.19: 46016902-46016902 on Build GRCh38
rs151196475 Chr.19: 46016963-46016963 on Build GRCh38
rs768715127 Chr.19: 46017062-46017062 on Build GRCh38
rs752032657 Chr.19: 46016855-46016855 on Build GRCh38
rs1047765505 Chr.19: 46016955-46016955 on Build GRCh38
rs756033518 Chr.19: 46017284-46017284 on Build GRCh38
rs781325604 Chr.19: 46017272-46017272 on Build GRCh38
rs376206073 Chr.19: 46017218-46017218 on Build GRCh38
rs962669754 Chr.19: 46016873-46016874 on Build GRCh38
rs761630220 Chr.19: 46016989-46016989 on Build GRCh38
rs765485023 Chr.19: 46017258-46017258 on Build GRCh38
rs375059660 Chr.19: 46017083-46017083 on Build GRCh38
rs866429310 Chr.19: 46016881-46016881 on Build GRCh38
rs976750147 Chr.19: 46017110-46017110 on Build GRCh38
rs200834056 Chr.19: 46016975-46016975 on Build GRCh38
rs189121953 Chr.19: 46017174-46017174 on Build GRCh38
rs769554095 Chr.19: 46016995-46016995 on Build GRCh38
rs1049283308 Chr.19: 46016923-46016923 on Build GRCh38
rs944973267 Chr.19: 46016847-46016847 on Build GRCh38
rs773319743 Chr.19: 46016948-46016957 on Build GRCh38
rs781563570 Chr.19: 46016942-46016942 on Build GRCh38
rs948467422 Chr.19: 46017195-46017195 on Build GRCh38
rs369391848 Chr.19: 46017186-46017186 on Build GRCh38
rs779155038 Chr.19: 46017049-46017049 on Build GRCh38
rs565327478 Chr.19: 46016886-46016886 on Build GRCh38
rs372980641 Chr.19: 46017194-46017194 on Build GRCh38
rs377175335 Chr.19: 46016983-46016983 on Build GRCh38
rs1038222144 Chr.19: 46017288-46017288 on Build GRCh38
rs1040016332 Chr.19: 46016868-46016868 on Build GRCh38
rs112695593 Chr.19: 46017076-46017076 on Build GRCh38
rs929038487 Chr.19: 46016889-46016889 on Build GRCh38
rs140257655 Chr.19: 46016981-46016981 on Build GRCh38
rs541395930 Chr.19: 46017097-46017097 on Build GRCh38
rs57007585 Chr.19: 46016953-46016953 on Build GRCh38
rs555115930 Chr.19: 46017107-46017107 on Build GRCh38
rs764396237 Chr.19: 46017235-46017235 on Build GRCh38
rs755440583 Chr.19: 46016880-46016880 on Build GRCh38
rs766930102 Chr.19: 46017230-46017230 on Build GRCh38
rs556774508 Chr.19: 46017187-46017187 on Build GRCh38
rs770563011 Chr.19: 46017099-46017099 on Build GRCh38
rs919556862 Chr.19: 46017131-46017131 on Build GRCh38
rs369963014 Chr.19: 46017018-46017018 on Build GRCh38
rs775955966 Chr.19: 46017232-46017232 on Build GRCh38
rs111751953 Chr.19: 46016969-46016969 on Build GRCh38
rs756423049 Chr.19: 46017027-46017027 on Build GRCh38
rs368392086 Chr.19: 46016967-46016967 on Build GRCh38
rs61745745 Chr.19: 46017030-46017030 on Build GRCh38
rs765840978 Chr.19: 46017005-46017005 on Build GRCh38
rs1022319465 Chr.19: 46017021-46017021 on Build GRCh38
rs751025101 Chr.19: 46017011-46017011 on Build GRCh38
rs781267025 Chr.19: 46017077-46017077 on Build GRCh38

View Full Product Details