Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46610164-46610671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM3
Entrez Gene ID
808
Gene Name
calmodulin 3
Gene Aliases
HEL-S-72, PHKD, PHKD3
UniGene
Hs.515487
Gene Chromosome Location
Chr.19: 46601255-46610782 on Build GRCh38
Transcripts
NM_005184.2

SNP Detail

SNP ID Chromosome Location
rs576220714 Chr.19: 46610506-46610506 on Build GRCh38
rs545083458 Chr.19: 46610550-46610550 on Build GRCh38
rs569035039 Chr.19: 46610264-46610264 on Build GRCh38
rs757732447 Chr.19: 46610344-46610344 on Build GRCh38
rs973361122 Chr.19: 46610265-46610265 on Build GRCh38
rs993013464 Chr.19: 46610282-46610282 on Build GRCh38
rs1039666487 Chr.19: 46610208-46610208 on Build GRCh38
rs945192894 Chr.19: 46610210-46610210 on Build GRCh38
rs1056202235 Chr.19: 46610259-46610259 on Build GRCh38
rs898225911 Chr.19: 46610480-46610480 on Build GRCh38
rs371134338 Chr.19: 46610441-46610441 on Build GRCh38
rs925674214 Chr.19: 46610327-46610327 on Build GRCh38
rs1039359957 Chr.19: 46610411-46610411 on Build GRCh38
rs1056342 Chr.19: 46610533-46610533 on Build GRCh38
rs887205902 Chr.19: 46610482-46610482 on Build GRCh38
rs963310110 Chr.19: 46610292-46610302 on Build GRCh38
rs975654944 Chr.19: 46610296-46610296 on Build GRCh38
rs954821813 Chr.19: 46610241-46610241 on Build GRCh38
rs917370352 Chr.19: 46610288-46610288 on Build GRCh38
rs904224496 Chr.19: 46610198-46610198 on Build GRCh38
rs1029889385 Chr.19: 46610238-46610238 on Build GRCh38
rs192964295 Chr.19: 46610189-46610189 on Build GRCh38
rs886978440 Chr.19: 46610507-46610507 on Build GRCh38
rs986536235 Chr.19: 46610359-46610359 on Build GRCh38
rs1028511570 Chr.19: 46610583-46610583 on Build GRCh38
rs920961769 Chr.19: 46610313-46610313 on Build GRCh38
rs977614767 Chr.19: 46610317-46610317 on Build GRCh38
rs1043676074 Chr.19: 46610220-46610220 on Build GRCh38
rs945099633 Chr.19: 46610409-46610409 on Build GRCh38
rs536431447 Chr.19: 46610242-46610242 on Build GRCh38
rs549239108 Chr.19: 46610263-46610263 on Build GRCh38
rs867386761 Chr.19: 46610402-46610402 on Build GRCh38
rs12978611 Chr.19: 46610495-46610495 on Build GRCh38
rs942551075 Chr.19: 46610467-46610467 on Build GRCh38
rs933864400 Chr.19: 46610417-46610417 on Build GRCh38
rs1017525532 Chr.19: 46610493-46610493 on Build GRCh38
rs3209098 Chr.19: 46610488-46610488 on Build GRCh38
rs537917821 Chr.19: 46610267-46610267 on Build GRCh38
rs1017076590 Chr.19: 46610280-46610281 on Build GRCh38
rs911022883 Chr.19: 46610464-46610464 on Build GRCh38
rs779565546 Chr.19: 46610413-46610413 on Build GRCh38
rs959188896 Chr.19: 46610277-46610277 on Build GRCh38
rs577571062 Chr.19: 46610314-46610314 on Build GRCh38
rs1004029999 Chr.19: 46610564-46610564 on Build GRCh38
rs776131232 Chr.19: 46610199-46610199 on Build GRCh38
rs997544640 Chr.19: 46610556-46610556 on Build GRCh38
rs998044031 Chr.19: 46610229-46610229 on Build GRCh38
rs994352552 Chr.19: 46610487-46610487 on Build GRCh38
rs935682696 Chr.19: 46610358-46610358 on Build GRCh38
rs1055044328 Chr.19: 46610463-46610463 on Build GRCh38
rs1004278784 Chr.19: 46610486-46610486 on Build GRCh38
rs1038506637 Chr.19: 46610475-46610475 on Build GRCh38
rs1002103511 Chr.19: 46610251-46610251 on Build GRCh38
rs955068795 Chr.19: 46610638-46610638 on Build GRCh38
rs899224750 Chr.19: 46610637-46610637 on Build GRCh38
rs376180231 Chr.19: 46610615-46610615 on Build GRCh38
rs563042406 Chr.19: 46610468-46610468 on Build GRCh38
rs1004777961 Chr.19: 46610275-46610275 on Build GRCh38
rs1014989405 Chr.19: 46610570-46610570 on Build GRCh38
rs554435949 Chr.19: 46610418-46610418 on Build GRCh38
rs534269672 Chr.19: 46610377-46610377 on Build GRCh38
rs557741221 Chr.19: 46610269-46610269 on Build GRCh38
rs28678916 Chr.19: 46610431-46610431 on Build GRCh38
rs1005344789 Chr.19: 46610243-46610243 on Build GRCh38
rs899738926 Chr.19: 46610211-46610211 on Build GRCh38
rs898653996 Chr.19: 46610541-46610541 on Build GRCh38

View Full Product Details