Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46649086-46649620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACT3
Entrez Gene ID
147906
Gene Name
dishevelled binding antagonist of beta catenin 3
Gene Aliases
DAPPER3, RRR1
UniGene
Hs.515490
Gene Chromosome Location
Chr.19: 46647564-46661138 on Build GRCh38
Transcripts
NM_001301046.1 NM_145056.2 XM_011526499.2 XM_011526500.2 XM_017026334.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612447 c.833G>T p.G278V Chr.19: 46649539-46649539 on Build GRCh38
5795262 c.577G>A p.A193T Chr.19: 46649120-46649120 on Build GRCh38
4410271 c.328G>A p.E110K Chr.19: 46649369-46649369 on Build GRCh38
1612450 c.832G>T p.G278C Chr.19: 46649540-46649540 on Build GRCh38
5967092 c.797G>A p.G266D Chr.19: 46649575-46649575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903120598 Chr.19: 46649286-46649286 on Build GRCh38
rs528739876 Chr.19: 46649516-46649516 on Build GRCh38
rs189187210 Chr.19: 46649570-46649570 on Build GRCh38
rs1023715359 Chr.19: 46649365-46649365 on Build GRCh38
rs936761454 Chr.19: 46649387-46649398 on Build GRCh38
rs951686456 Chr.19: 46649501-46649501 on Build GRCh38
rs924429086 Chr.19: 46649534-46649534 on Build GRCh38
rs944553758 Chr.19: 46649262-46649262 on Build GRCh38
rs1045440321 Chr.19: 46649590-46649590 on Build GRCh38
rs920236273 Chr.19: 46649177-46649177 on Build GRCh38
rs780961523 Chr.19: 46649364-46649364 on Build GRCh38
rs920286982 Chr.19: 46649500-46649500 on Build GRCh38
rs188835431 Chr.19: 46649431-46649431 on Build GRCh38
rs998704536 Chr.19: 46649334-46649334 on Build GRCh38
rs766383316 Chr.19: 46649319-46649319 on Build GRCh38
rs928242049 Chr.19: 46649134-46649134 on Build GRCh38
rs761762456 Chr.19: 46649276-46649276 on Build GRCh38
rs971739715 Chr.19: 46649163-46649163 on Build GRCh38
rs755804135 Chr.19: 46649466-46649466 on Build GRCh38
rs986324823 Chr.19: 46649503-46649503 on Build GRCh38
rs777240402 Chr.19: 46649478-46649478 on Build GRCh38
rs748858897 Chr.19: 46649498-46649498 on Build GRCh38
rs1050170339 Chr.19: 46649191-46649191 on Build GRCh38
rs944424605 Chr.19: 46649519-46649519 on Build GRCh38
rs751477896 Chr.19: 46649351-46649351 on Build GRCh38
rs752423035 Chr.19: 46649432-46649432 on Build GRCh38
rs940890909 Chr.19: 46649148-46649148 on Build GRCh38
rs191886465 Chr.19: 46649537-46649537 on Build GRCh38
rs184739971 Chr.19: 46649556-46649556 on Build GRCh38
rs1040577259 Chr.19: 46649279-46649279 on Build GRCh38
rs930403897 Chr.19: 46649179-46649179 on Build GRCh38
rs962309470 Chr.19: 46649462-46649462 on Build GRCh38
rs17851379 Chr.19: 46649557-46649557 on Build GRCh38
rs1040224631 Chr.19: 46649530-46649530 on Build GRCh38
rs780144023 Chr.19: 46649211-46649227 on Build GRCh38
rs868568685 Chr.19: 46649536-46649536 on Build GRCh38
rs910412071 Chr.19: 46649198-46649198 on Build GRCh38
rs754826442 Chr.19: 46649355-46649355 on Build GRCh38

View Full Product Details